Buffer solution pH 7, Phosphate buffer, traceable to NIST, ready to use
Buffer solution pH 7, Phosphate buffer, traceable to NIST, ready to use
Buffer solution pH 7, Phosphate buffer, traceable to NIST, ready to use
Buffer solution pH 7, Phosphate buffer, traceable to NIST, ready to use
Thermo Scientific Chemicals

Buffer solution pH 7, Phosphate buffer, traceable to NIST, ready to use

CAS: 7558-79-4
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
Catalog number 383855000
Price (USD)
39.50
Each
In stock
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (USD)
39.50
Each
Add to cart
Chemical Identifiers
CAS7558-79-4
IUPAC Namedisodium hydrogen phosphate
Molecular FormulaHNa2O4P
InChI KeyBNIILDVGGAEEIG-UHFFFAOYSA-L
SMILES[Na+].[Na+].OP([O-])([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
pH6.98 to 7.02 (at 20°C)
Appearance (Form)Solution
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
2540915Certificate of AnalysisJun 13, 2025383855000, 383850010
2540914Certificate of AnalysisJun 13, 2025383855000, 383850010
2536235Certificate of AnalysisMay 24, 2025383855000, 383850010
2536234Certificate of AnalysisMay 24, 2025383855000, 383850010
2526118Certificate of AnalysisFeb 11, 2025383855000, 383850010
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection