Cyanocobalamin, 96%
Cyanocobalamin, 96%
Cyanocobalamin, 96%
Thermo Scientific Chemicals

Cyanocobalamin, 96%

CAS: 68-19-9 | C63H88CoN14O14P | 1355.39 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mg
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 405920010
Price (USD)
136.65
Special offer
Online exclusive
161.00
Save 24.35 (15%)
Each
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
136.65
Special offer
Online exclusive
161.00
Save 24.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS68-19-9
IUPAC Name(10S,12R,13R,24S,35R,41R)-30,35,40-tris(2-carbamoylethyl)-24,36,41-tris(carbamoylmethyl)-1-cyano-46-hydroxy-12-(hydroxymethyl)-5,6,17,23,28,31,31,36,38,41,42-undecamethyl-15,20-dioxo-11,14,16-trioxa-2λ⁵,9,19,26λ⁵,43,44λ⁵,45λ⁵-heptaaza-15λ⁵-phospha-1-cobaltadodecacyclo[27.14.1.1¹,³⁴.1²,⁹.1¹⁰,¹³.0¹,²⁶.0³,⁸.0²³,²⁷.0²⁵,⁴².0³²,⁴⁴.0³⁹,⁴³.0³⁷,⁴⁵]heptatetraconta-2(47),3,5,7,26,28,32(44),33,37(45),38-decaene-1,2,26,44,45-pentakis(ylium)-15-olate
Molecular FormulaC63H88CoN14O14P
InChI KeyYUWVGNQGDAPKCX-BVWPOUIRNA-L
SMILES[C@@H]1(C2=CC3=[N+]4C(C(C3(C)C)CCC(N)=O)=C(C3=[N+]5C6C7(N8C(C([C@@]7(C)CC(N)=O)CCC(N)=O)=C(C(=[N+]2[Co+]458([N+]2=CN(C4=CC(=C(C)C=C24)C)[C@@]2([H])O[C@@H]([C@]([H])(OP([O-])(=O)OC(C)CNC(=O)CCC3(C)[C@@H]6CC(N)=O)C2O)CO)C#N)C1(C)CC(N)=O)C)C)C)CCC(N)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
Loss on drying=<12 % (20 mg, 105°C, 2 hrs) (vacuum)
Appearance (Color)Red to dark red
Infrared spectrumConforms
UV spectrophotometry>=95 % (on dried substance)
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0467767Certificate of AnalysisJan 31, 2025405920010, 405920050, 405920250, 405925000
A0463384Certificate of AnalysisMay 31, 2024405920010, 405920050, 405920250, 405925000
A0450663Certificate of AnalysisMay 02, 2023405920010, 405920050, 405920250, 405925000
A0440053Certificate of AnalysisFeb 27, 2023405920010, 405920050, 405920250, 405925000
A0408177Certificate of AnalysisDec 13, 2022405920010, 405920050, 405920250, 405925000
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link