Ammonium phosphate, dibasic, 98+%, ACS reagent
Ammonium phosphate, dibasic, 98+%, ACS reagent
Ammonium phosphate, dibasic, 98+%, ACS reagent
Ammonium phosphate, dibasic, 98+%, ACS reagent
Thermo Scientific Chemicals

Ammonium phosphate, dibasic, 98+%, ACS reagent

CAS: 7783-28-0 | H9N2O4P | 132.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 423375000
Price (USD)
103.65
Special offer
Online exclusive
122.00
Save 18.35 (15%)
Each
In stock
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
103.65
Special offer
Online exclusive
122.00
Save 18.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS7783-28-0
IUPAC Namephosphoric acid diamine
Molecular FormulaH9N2O4P
InChI KeyMNNHAPBLZZVQHP-UHFFFAOYSA-N
SMILESN.N.OP(O)(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystalline powder or crystals and/or chunks
Titration with HCl>=98.0 %
Heavy metals (as Pb)=<0.001 %
Insoluble matter=<0.005 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection