TaqMan™ Gene Expression Assay (FAM)
TaqMan™ Gene Expression Assay (FAM)
Applied Biosystems™

TaqMan™ Gene Expression Assay (FAM)

Green features
Applied Biosystems TaqMan Gene Expression assays are used for quantitative real-time PCR analysis of gene expression and consist of aRead more
Promo ImagePromotion
Promo code:P5780456
Gene Expression Promo
From RNA Isolation kits, reverse transcriptases to analysis of your gene with qPCR - upto 35% on all Gene Expression workflow productsLearn More
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of Reactions
4351368Promo ImageL (2900 x 967 μL Reactions), made to order
4331182Promo ImageS (250 x 250 μL Reactions), inventoried
4351370Promo ImageM (750 x 750 μL Reactions), made to order
4351372Promo ImageS (360 x 360 μL Reactions), made to order
4448892Promo ImageXS (75 x 75 μL Reactions), made to order
4453320Promo ImageXS (75 x 75 μL Reactions), inventoried
Catalog number 4351368
Price (USD)
1,460.65
Online Exclusive
1,560.00
Save 99.35 (6%)
Each
Order now
No. of Reactions:
L (2900 x 967 μL Reactions), made to order
Ask our AI about this Product

Applied Biosystems TaqMan Gene Expression assays are used for quantitative real-time PCR analysis of gene expression and consist of a pair of unlabeled PCR primers and a TaqMan probe with a dye label (FAM) on the 5' end and a minor groove binder (MGB) and non-fluorescent quencher (NFQ) on the 3' end.

Features of TaqMan Gene Expression assays include:

  • Easy to use—just add cDNA and master mix and run the qPCR—no melt curves required
  • Specific—TaqMan assays use proprietary MGB-containing probes that can be up to 15 bases shorter than non-MGB probes, improving the specificity of the assay
  • Sensitive—TaqMan assays are ideal for measuring low levels of expression or low-abundance targets
  • Accurate—identify small fold-changes with high accuracy of quantitation
  • Extensive content—Over 1.8 million predesigned assays available for over 25 different species
  • Gold-standard TaqMan qPCR chemistry—TaqMan assays draw on Thermo Fisher Scientific's bioinformatics assay design pipeline to help ensure high specificity and minimal cross-reactivity, even for gene variants with high sequence homology
  • Fast delivery—your assays shipped in as little as 1–2 days (see below)

Approximate ship time

  • Inventoried: 1–2 days in North America, 3–5 days in Europe*
  • Made to order: 4–6 days in North America, 6–10 days in Europe

TaqMan Gene Expression assays are the gold standard in real-time PCR gene expression studies, built on more than 20 years of experience. Each assay includes target primers and a sequence-specific probe optimized for the best functional performance. No additional design, optimization, or melt curve analysis is needed. Available in a wide variety of formats and species, new assay designs are constantly added to help meet your research needs. TaqMan assays have been cited in over 40,000 publications and are backed by more than 350 patents.

Pre-formulated assay

  • 2 unlabeled PCR primers (900 nM each final 1X concentration)
  • 1 FAM dye-labeled TaqMan MGB probe (250 nM final 1X concentration)

All of our predesigned TaqMan Gene Expression assays are covered by the TaqMan Assays qPCR Guarantee.**

Recommended master mix: TaqMan Fast Advanced Master Mix (Cat. No. 4444556)

*Our most popular assays are stocked in North America and Europe for overnight delivery where possible. If an assay is out of stock, it may take an additional 3–5 business days for delivery.

**Subject to terms and conditions. For complete details, go to www.thermofisher.com/taqmanguarantee.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration60X
Fidelity (vs. Taq)60 X
For Use With (Equipment)7500 System, 7500 Fast System, 7900HT System, StepOne™, StepOnePlus™, ViiA™ 7 System, QuantStudio™ 3, QuantStudio™ 5, QuantStudio™ 6 Flex, QuantStudio™ 7 Flex, QuantStudio 6 Pro, QuantStudio 7 Pro, QuantStudio™ 12k Flex, QuantStudio™ Absolute Q Digital PCR System
FormatTube
Green FeaturesSustainable packaging
No. of ReactionsL (2900 x 967 μL Reactions), made to order
Product LineTaqMan™
Product TypeGene Expression Assay
QuantityL (2900 reactions/967 μL), made to order
Shipping ConditionRoom Temperature
Detection MethodPrimer-probe
GC-Rich PCR PerformanceHigh
PCR MethodqPCR
Reaction SpeedFast
Unit SizeEach
Contents & Storage
1 tube containing 20X (XS, S, and M sizes) or 60X (L size) mix of pre-formulated assay (1 probe and 2 primers).

Store at -15°C to -25°C.

Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Frequently asked questions (FAQs)

Can I multiplex my assays if my amplicons are not all the same length?

Yes, it is possible. The amplicons do not have to be the same length in order to duplex the assays. You need to use different reporter dyes on the probes in multiplex - they should have a substantial difference in maximum emission wavelength to be effective. A classic example is to use FAM and VIC for the 2 probes. Depending on the expression of the targets, you may also need to limit the primer concentration for one of the assays. This is common when duplexing a target and endogenous control (the control probe is VIC labeled and the primer concentration is decreased so the target assay can properly compete for use of enzyme and dNTPs).

I want to order a TaqMan Gene Expression Assay that you have discontinued. Is there a way do that?

Yes, you can order it as a custom assay. Since the assay is discontinued, it can not be ordered on-line. You will have to call or email the order to genomics@appliedbiosystems.com, and include the assay ID. The assay will be synthesized once the order is received. It will be designed and priced as a Custom TaqMan Gene Expression Assay so you will need to choose the scale.

Can I use TaqMan Gene Expression Assays for a gene copy number study?

TaqMan Gene Expression Assays with “_s1” as assay ID suffix are the assays whose primers and probes are designed within a single exon, and therefore, will detect genomic DNA. However, the application of these assays in gene copy number studies has not been validated. Other gene expression assays will not detect gDNA, let alone the region of variance.

I am working on both mouse and human samples for gene expression studies. Do you have one endogenous control that I can use with both species?

The 18S rRNA is highly conserved among eukaryotes. We have several configurations of assays for this gene: P/N 4331182, Assay ID: Hs99999901_s1; PN 4319413E, 4310893E, 4333760T, 4333760F, 4308329. You can use any of these for both human and mouse samples, as well as samples from any other eukaryotic species.

Can I check the end product of a TaqMan Gene Expression Assay by running an agarose gel?

Yes, you can check the end product on an agarose gel. If you use the TaqMan Universal PCR Master Mix (including AmpErase UNG, Cat. No. 4304437), we recommend running the gel immediately after the PCR, since the trace amount of AmpErase UNG may digest the end product that contains double-stranded DNA with dUTP incorporated. The average length of amplicon ranges from 50-150 bp. The exact amplicon length is indicated on the detailed page of each assay on our website. You can get to that page by clicking on the Assay ID; the amplicon information is listed in the section "Assay Details".

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
7189900Certificate of AnalysisMar 11, 20204331182
1 result displayed, search above for a specific certificate

Safety Data Sheets

Scientific Resources

Product Information

Citations & References (6702)

Citations & References
Abstract
KIF14 messenger RNA expression is independently prognostic for outcome in lung cancer.
Authors:Corson TW,Zhu CQ,Lau SK,Shepherd FA,Tsao MS,Gallie BL
Journal:Clinical cancer research : an official journal of the American Association for Cancer Research
PubMed ID:17545527
Cloning and functional characterization of human sodium-dependent organic anion transporter (SLC10A6).
Authors:Geyer J,Döring B,Meerkamp K,Ugele B,Bakhiya N,Fernandes CF,Godoy JR,Glatt H,Petzinger E
Journal:The Journal of biological chemistry
PubMed ID:17491011
Large-scale chemical dissection of mitochondrial function.
Authors:Wagner BK,Kitami T,Gilbert TJ,Peck D,Ramanathan A,Schreiber SL,Golub TR,Mootha VK
Journal:Nature biotechnology
PubMed ID:18297058
Mitochondrial oxidative phosphorylation (OXPHOS) is central to physiology and disease pathogenesis. To systematically investigate its activity and regulation, we performed a wide range of assays of OXPHOS physiology and nuclear and mitochondrial gene expression across 2490 chemical perturbations in muscle cells. Through mining of the resulting compendium, we discovered that: ... More
Human embryonic stem cell microenvironment suppresses the tumorigenic phenotype of aggressive cancer cells.
Authors:Postovit LM,Margaryan NV,Seftor EA,Kirschmann DA,Lipavsky A,Wheaton WW,Abbott DE,Seftor RE,Hendrix MJ
Journal:Proceedings of the National Academy of Sciences of the United States of America
PubMed ID:18334633
Embryonic stem cells sustain a microenvironment that facilitates a balance of self-renewal and differentiation. Aggressive cancer cells, expressing a multipotent, embryonic cell-like phenotype, engage in a dynamic reciprocity with a microenvironment that promotes plasticity and tumorigenicity. However, the cancer-associated milieu lacks the appropriate regulatory mechanisms to maintain a normal cellular ... More
Authors:
Journal:
PubMed ID:11114349
6702 total citations

Other products to consider



Share catalog number, name or link

1x1 image pixel for data collection