Potassium phosphate, monobasic, for HPLC
Potassium phosphate, monobasic, for HPLC
Thermo Scientific Chemicals

Potassium phosphate, monobasic, for HPLC

CAS: 7778-77-0 | H2KO4P | 136.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1 kg
Catalog number 447370010
Price (USD)
640.00
Each
In stock
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (USD)
640.00
Each
Add to cart
Chemical Identifiers
CAS7778-77-0
IUPAC Namepotassium dihydrogen phosphate
Molecular FormulaH2KO4P
InChI KeyGNSKLFRGEWLPPA-UHFFFAOYSA-M
SMILES[K+].OP(O)([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
UVat 280 nm A: =<0.05
Gradient analysispasses test
Titration with NaOH>=99.4 %
UVat 250 nm A: =<0.05
Appearance (Color)White
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0463588Certificate of AnalysisJun 27, 2024447370010, 447372500, 447370500
A0450269Certificate of AnalysisMay 02, 2023447370010, 447372500, 447370500
A0444102Certificate of AnalysisMay 02, 2023447370010, 447370500, 447372500
A0444102Certificate of AnalysisJul 14, 2022447370010, 447370500, 447372500
A0441804Certificate of AnalysisMay 03, 2022447370010, 447372500, 447370500
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection