Thermo Scientific™

SMART Digest™ Trypsin Kits

Catalog number: 60109-101-LPH
Have Questions?
Thermo Scientific™

SMART Digest™ Trypsin Kits

Catalog number: 60109-101-LPH
Have Questions?

Obtain high quality analytical results from protein digests using Thermo Scientific™ SMART Digest™ Kits. A significant advance in sample preparation for biopharmaceutical protein research, the kits provide fast and simple protein digestion with high reproducibility, high sensitivity, and high levels of data quality in a format that's compatible with automation. The SMART Digest Kits are a significant improvement over current in-solution protein digestion technologies, which are not reproducible, have poor sensitivity, and employ protracted methodologies not amenable to automation.

 
Catalog Number
60109-101-LPH
Unit Size
Each
Description
Smart digest kit with collection plate, low pH
Format
SMART Digest Low pH Resin Pre-filled into PCR Tubes
Includes
SMART Digest Low pH Resin Pre-filled into 96 PCR Tubes, Collection Plate
Price (USD)
Full specifications
Quantity
-
TypeProtein Digestion, MAM
DescriptionSmart digest kit with collection plate, low pH
FormatSMART Digest Low pH Resin Pre-filled into PCR Tubes
EnzymeTrypsin
CleavageC-terminal side of lysine and arginine amino acids
CaptureNone
Product LineSMART Digest
For Use With (Application)Peptide Quantitation, Peptide Mapping
IncludesSMART Digest Low pH Resin Pre-filled into 96 PCR Tubes, Collection Plate
Unit SizeEach
Catalog Number
Unit Size
Each
Description
SMART Digest Trypsin Kit, with Collection plate
Format
SMART Digest Resin Pre-filled into PCR Tubes
Includes
SMART Digest Resin Pre-filled into 96 PCR Tubes, Collection Plate
Price (USD)
Full specifications
Quantity
TypeProtein Digestion
DescriptionSMART Digest Trypsin Kit, with Collection plate
FormatSMART Digest Resin Pre-filled into PCR Tubes
EnzymeTrypsin
CleavageC-terminal side of lysine and arginine amino acids
CaptureNone
Product LineSMART Digest
For Use With (Application)Peptide Quantitation, Peptide Mapping
IncludesSMART Digest Resin Pre-filled into 96 PCR Tubes, Collection Plate
Unit SizeEach
Catalog Number
Unit Size
Each
Description
SMART Digest Trypsin Kit, with SOLAμ/collection plate
Format
SMART Digest Resin Pre-filled into PCR Tubes
Includes
SMART Digest Resin Pre-filled into 96 PCR Tubes, Collection Plate, SOLAμ SPE Plate
Price (USD)
Full specifications
Quantity
TypeProtein Digestion
DescriptionSMART Digest Trypsin Kit, with SOLAμ/collection plate
FormatSMART Digest Resin Pre-filled into PCR Tubes
EnzymeTrypsin
CleavageC-terminal side of lysine and arginine amino acids
CaptureNone
Product LineSMART Digest
For Use With (Application)Peptide Quantitation, Peptide Mapping
IncludesSMART Digest Resin Pre-filled into 96 PCR Tubes, Collection Plate, SOLAμ SPE Plate
Unit SizeEach
Catalog Number
Unit Size
Each
Description
SMART Digest Trypsin Kit, with Filter/Collection plate
Format
SMART Digest Resin Pre-filled into PCR Tubes
Includes
SMART Digest Resin Pre-filled into 96 PCR Tubes, Collection Plate, Filter Plate
Price (USD)
Full specifications
Quantity
TypeProtein Digestion
DescriptionSMART Digest Trypsin Kit, with Filter/Collection plate
FormatSMART Digest Resin Pre-filled into PCR Tubes
EnzymeTrypsin
CleavageC-terminal side of lysine and arginine amino acids
CaptureNone
Product LineSMART Digest
For Use With (Application)Peptide Quantitation, Peptide Mapping
IncludesSMART Digest Resin Pre-filled into 96 PCR Tubes, Collection Plate, Filter Plate
Unit SizeEach
Catalog Number
Unit Size
Each
Description
SMART Digest Trypsin Kit, Bulk Resin option
Format
SMART Digest Resin, Bulk Non-magnetic
Includes
Bulk Non-magnetic SMART Digest Resin, SMART Digest Buffer
Price (USD)
Full specifications
Quantity
TypeProtein Digestion
DescriptionSMART Digest Trypsin Kit, Bulk Resin option
FormatSMART Digest Resin, Bulk Non-magnetic
EnzymeTrypsin
CleavageC-terminal side of lysine and arginine amino acids
CaptureNone
Product LineSMART Digest
For Use With (Application)Peptide Quantitation, Peptide Mapping
IncludesBulk Non-magnetic SMART Digest Resin, SMART Digest Buffer
Unit SizeEach
Catalog Number
Unit Size
Each
Description
SMART Digest Trypsin Kit, Magnetic Bulk Resin option
Format
SMART Digest Resin, Bulk Magnetic
Includes
Bulk Magnetic SMART Digest Resin, SMART Digest Buffer
Price (USD)
Full specifications
Quantity
TypeProtein Digestion
DescriptionSMART Digest Trypsin Kit, Magnetic Bulk Resin option
FormatSMART Digest Resin, Bulk Magnetic
EnzymeTrypsin
CleavageC-terminal side of lysine and arginine amino acids
CaptureNone
Product LineSMART Digest
For Use With (Application)Peptide Quantitation, Peptide Mapping
IncludesBulk Magnetic SMART Digest Resin, SMART Digest Buffer
Unit SizeEach
Catalog Number
Unit Size
Each
Description
SMART Digest Trypsin Kit, Bulk Resin option with Filter/Collection plate
Format
SMART Digest Resin, Bulk Non-magnetic
Includes
Bulk Non-magnetic SMART Digest Resin, SMART Digest Buffer, Collection Plate, Filter Plate
Price (USD)
Full specifications
Quantity
TypeProtein Digestion
DescriptionSMART Digest Trypsin Kit, Bulk Resin option with Filter/Collection plate
FormatSMART Digest Resin, Bulk Non-magnetic
EnzymeTrypsin
CleavageC-terminal side of lysine and arginine amino acids
CaptureNone
Product LineSMART Digest
For Use With (Application)Peptide Quantitation, Peptide Mapping
IncludesBulk Non-magnetic SMART Digest Resin, SMART Digest Buffer, Collection Plate, Filter Plate
Unit SizeEach
Catalog Number
Unit Size
Each
Description
SMART Digest Trypsin Kit, Magnetic Bulk Resin option with Filter/Collection plate
Format
SMART Digest Resin, Bulk Magnetic
Includes
Bulk Magnetic SMART Digest Resin, SMART Digest Buffer, Collection Plate, Filter Plate
Price (USD)
Full specifications
Quantity
TypeProtein Digestion
DescriptionSMART Digest Trypsin Kit, Magnetic Bulk Resin option with Filter/Collection plate
FormatSMART Digest Resin, Bulk Magnetic
EnzymeTrypsin
CleavageC-terminal side of lysine and arginine amino acids
CaptureNone
Product LineSMART Digest
For Use With (Application)Peptide Quantitation, Peptide Mapping
IncludesBulk Magnetic SMART Digest Resin, SMART Digest Buffer, Collection Plate, Filter Plate
Unit SizeEach
Catalog Number
Unit Size
Each
Description
SMART Digest Trypsin Kit, Bulk Resin option with SOLAμ/collection plate
Format
SMART Digest Resin, Bulk Non-magnetic
Includes
Bulk Non-magnetic SMART Digest Resin, SMART Digest Buffer, Collection Plate, SOLAμ SPE Plate
Price (USD)
Full specifications
Quantity
TypeProtein Digestion
DescriptionSMART Digest Trypsin Kit, Bulk Resin option with SOLAμ/collection plate
FormatSMART Digest Resin, Bulk Non-magnetic
EnzymeTrypsin
CleavageC-terminal side of lysine and arginine amino acids
CaptureNone
Product LineSMART Digest
For Use With (Application)Peptide Quantitation, Peptide Mapping
IncludesBulk Non-magnetic SMART Digest Resin, SMART Digest Buffer, Collection Plate, SOLAμ SPE Plate
Unit SizeEach
Catalog Number
Unit Size
Each
Description
SMART Digest Trypsin Kit, Soluble
Format
SMART Digest Soluble Trypsin
Includes
SMART Digest Soluble Trypsin, SMART Digest Buffer, Collection Plate
Price (USD)
Full specifications
Quantity
TypeProtein Digestion
DescriptionSMART Digest Trypsin Kit, Soluble
FormatSMART Digest Soluble Trypsin
EnzymeTrypsin
CleavageC-terminal side of lysine and arginine amino acids
CaptureNone
Product LineSMART Digest
For Use With (Application)Peptide Quantitation, Peptide Mapping
IncludesSMART Digest Soluble Trypsin, SMART Digest Buffer, Collection Plate
Unit SizeEach
Showing 10 of 11Show more
Catalog NumberSpecificationsUnit SizeDescriptionFormatIncludesPrice (USD)Quantity
60109-101-LPHFull specifications
EachSmart digest kit with collection plate, low pHSMART Digest Low pH Resin Pre-filled into PCR TubesSMART Digest Low pH Resin Pre-filled into 96 PCR Tubes, Collection PlateRequest A Quote-
TypeProtein Digestion, MAM
DescriptionSmart digest kit with collection plate, low pH
FormatSMART Digest Low pH Resin Pre-filled into PCR Tubes
EnzymeTrypsin
CleavageC-terminal side of lysine and arginine amino acids
CaptureNone
Product LineSMART Digest
For Use With (Application)Peptide Quantitation, Peptide Mapping
IncludesSMART Digest Low pH Resin Pre-filled into 96 PCR Tubes, Collection Plate
Unit SizeEach
60109-101Full specifications
EachSMART Digest Trypsin Kit, with Collection plateSMART Digest Resin Pre-filled into PCR TubesSMART Digest Resin Pre-filled into 96 PCR Tubes, Collection Plate
TypeProtein Digestion
DescriptionSMART Digest Trypsin Kit, with Collection plate
FormatSMART Digest Resin Pre-filled into PCR Tubes
EnzymeTrypsin
CleavageC-terminal side of lysine and arginine amino acids
CaptureNone
Product LineSMART Digest
For Use With (Application)Peptide Quantitation, Peptide Mapping
IncludesSMART Digest Resin Pre-filled into 96 PCR Tubes, Collection Plate
Unit SizeEach
60109-103Full specifications
EachSMART Digest Trypsin Kit, with SOLAμ/collection plateSMART Digest Resin Pre-filled into PCR TubesSMART Digest Resin Pre-filled into 96 PCR Tubes, Collection Plate, SOLAμ SPE Plate
TypeProtein Digestion
DescriptionSMART Digest Trypsin Kit, with SOLAμ/collection plate
FormatSMART Digest Resin Pre-filled into PCR Tubes
EnzymeTrypsin
CleavageC-terminal side of lysine and arginine amino acids
CaptureNone
Product LineSMART Digest
For Use With (Application)Peptide Quantitation, Peptide Mapping
IncludesSMART Digest Resin Pre-filled into 96 PCR Tubes, Collection Plate, SOLAμ SPE Plate
Unit SizeEach
60109-102Full specifications
EachSMART Digest Trypsin Kit, with Filter/Collection plateSMART Digest Resin Pre-filled into PCR TubesSMART Digest Resin Pre-filled into 96 PCR Tubes, Collection Plate, Filter Plate
TypeProtein Digestion
DescriptionSMART Digest Trypsin Kit, with Filter/Collection plate
FormatSMART Digest Resin Pre-filled into PCR Tubes
EnzymeTrypsin
CleavageC-terminal side of lysine and arginine amino acids
CaptureNone
Product LineSMART Digest
For Use With (Application)Peptide Quantitation, Peptide Mapping
IncludesSMART Digest Resin Pre-filled into 96 PCR Tubes, Collection Plate, Filter Plate
Unit SizeEach
60109-101-BFull specifications
EachSMART Digest Trypsin Kit, Bulk Resin optionSMART Digest Resin, Bulk Non-magneticBulk Non-magnetic SMART Digest Resin, SMART Digest Buffer
TypeProtein Digestion
DescriptionSMART Digest Trypsin Kit, Bulk Resin option
FormatSMART Digest Resin, Bulk Non-magnetic
EnzymeTrypsin
CleavageC-terminal side of lysine and arginine amino acids
CaptureNone
Product LineSMART Digest
For Use With (Application)Peptide Quantitation, Peptide Mapping
IncludesBulk Non-magnetic SMART Digest Resin, SMART Digest Buffer
Unit SizeEach
60109-101-MBFull specifications
EachSMART Digest Trypsin Kit, Magnetic Bulk Resin optionSMART Digest Resin, Bulk MagneticBulk Magnetic SMART Digest Resin, SMART Digest Buffer
TypeProtein Digestion
DescriptionSMART Digest Trypsin Kit, Magnetic Bulk Resin option
FormatSMART Digest Resin, Bulk Magnetic
EnzymeTrypsin
CleavageC-terminal side of lysine and arginine amino acids
CaptureNone
Product LineSMART Digest
For Use With (Application)Peptide Quantitation, Peptide Mapping
IncludesBulk Magnetic SMART Digest Resin, SMART Digest Buffer
Unit SizeEach
60109-102-BFull specifications
EachSMART Digest Trypsin Kit, Bulk Resin option with Filter/Collection plateSMART Digest Resin, Bulk Non-magneticBulk Non-magnetic SMART Digest Resin, SMART Digest Buffer, Collection Plate, Filter Plate
TypeProtein Digestion
DescriptionSMART Digest Trypsin Kit, Bulk Resin option with Filter/Collection plate
FormatSMART Digest Resin, Bulk Non-magnetic
EnzymeTrypsin
CleavageC-terminal side of lysine and arginine amino acids
CaptureNone
Product LineSMART Digest
For Use With (Application)Peptide Quantitation, Peptide Mapping
IncludesBulk Non-magnetic SMART Digest Resin, SMART Digest Buffer, Collection Plate, Filter Plate
Unit SizeEach
60109-102-MBFull specifications
EachSMART Digest Trypsin Kit, Magnetic Bulk Resin option with Filter/Collection plateSMART Digest Resin, Bulk MagneticBulk Magnetic SMART Digest Resin, SMART Digest Buffer, Collection Plate, Filter Plate
TypeProtein Digestion
DescriptionSMART Digest Trypsin Kit, Magnetic Bulk Resin option with Filter/Collection plate
FormatSMART Digest Resin, Bulk Magnetic
EnzymeTrypsin
CleavageC-terminal side of lysine and arginine amino acids
CaptureNone
Product LineSMART Digest
For Use With (Application)Peptide Quantitation, Peptide Mapping
IncludesBulk Magnetic SMART Digest Resin, SMART Digest Buffer, Collection Plate, Filter Plate
Unit SizeEach
60109-103-BFull specifications
EachSMART Digest Trypsin Kit, Bulk Resin option with SOLAμ/collection plateSMART Digest Resin, Bulk Non-magneticBulk Non-magnetic SMART Digest Resin, SMART Digest Buffer, Collection Plate, SOLAμ SPE Plate
TypeProtein Digestion
DescriptionSMART Digest Trypsin Kit, Bulk Resin option with SOLAμ/collection plate
FormatSMART Digest Resin, Bulk Non-magnetic
EnzymeTrypsin
CleavageC-terminal side of lysine and arginine amino acids
CaptureNone
Product LineSMART Digest
For Use With (Application)Peptide Quantitation, Peptide Mapping
IncludesBulk Non-magnetic SMART Digest Resin, SMART Digest Buffer, Collection Plate, SOLAμ SPE Plate
Unit SizeEach
60113-101Full specifications
EachSMART Digest Trypsin Kit, SolubleSMART Digest Soluble TrypsinSMART Digest Soluble Trypsin, SMART Digest Buffer, Collection Plate
TypeProtein Digestion
DescriptionSMART Digest Trypsin Kit, Soluble
FormatSMART Digest Soluble Trypsin
EnzymeTrypsin
CleavageC-terminal side of lysine and arginine amino acids
CaptureNone
Product LineSMART Digest
For Use With (Application)Peptide Quantitation, Peptide Mapping
IncludesSMART Digest Soluble Trypsin, SMART Digest Buffer, Collection Plate
Unit SizeEach
Showing 10 of 11Show more
Trypsin Protein Digestion

SMART Digest kits enable you to characterize and quantitate proteins faster, easier, and quicker than ever before. This version of the kit uses trypsin, which cleaves peptides on the C-terminal side of lysine and arginine amino acid residues.

Easy and simple
  • Significantly increased speed of sample processing
  • Simple process
  • Easily automate entire protocol for high-throughput processing
Reliable Results
  • Increased reproducibility over existing protocols
  • Higher sensitivity
  • Higher data confidence
Significantly Reduces Sample Preparation Time

Typically it takes less than 60 minutes (dependent on sample complexity) to achieve full digestion, whereas an in-solution digest can take up to 24 hours.

Cleaner Samples

Reduction in the number of chemicals used and the immobilized trypsin design helps mitigate against chemically induced post translational modifications (PTMs) and autolysis. This results in results which are easier to interpret.

Couple with SOLAμ SPE Plates for Desalting

When used with the SMART Digest kits for desalting, Thermo Scientific™ SOLAμ™ SPE solid phase extraction plates provide a fast, easy-to-use, robust, and easily automatable sample preparation solution for analysis of biomolecules.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text