Obtain high quality analytical results from protein digests using Thermo Scientific™ SMART Digest™ Kits. A significant advance in sample preparation for biopharmaceutical protein research, the kits provide fast and simple protein digestion with high reproducibility, high sensitivity, and high levels of data quality in a format that's compatible with automation. The SMART Digest Kits are a significant improvement over current in-solution protein digestion technologies, which are not reproducible, have poor sensitivity, and employ protracted methodologies not amenable to automation.
Catalog Number
60109-101-MB
Unit Size
Each
Description
SMART Digest Trypsin Kit, Magnetic Bulk Resin option
Format
SMART Digest Resin, Bulk Magnetic
Includes
Bulk Magnetic SMART Digest Resin, SMART Digest Buffer
Price (USD)
903.19
Full specifications
Quantity
Type
Protein Digestion
Description
SMART Digest Trypsin Kit, Magnetic Bulk Resin option
Format
SMART Digest Resin, Bulk Magnetic
Enzyme
Trypsin
Cleavage
C-terminal side of lysine and arginine amino acids
Capture
None
Product Line
SMART Digest
For Use With (Application)
Peptide Quantitation, Peptide Mapping
Includes
Bulk Magnetic SMART Digest Resin, SMART Digest Buffer
SMART Digest kits enable you to characterize and quantitate proteins faster, easier, and quicker than ever before. This version of the kit uses trypsin, which cleaves peptides on the C-terminal side of lysine and arginine amino acid residues.
Easy and simple
Significantly increased speed of sample processing
Simple process
Easily automate entire protocol for high-throughput processing
Reliable Results
Increased reproducibility over existing protocols
Higher sensitivity
Higher data confidence
Significantly Reduces Sample Preparation Time
Typically it takes less than 60 minutes (dependent on sample complexity) to achieve full digestion, whereas an in-solution digest can take up to 24 hours.
Cleaner Samples
Reduction in the number of chemicals used and the immobilized trypsin design helps mitigate against chemically induced post translational modifications (PTMs) and autolysis. This results in results which are easier to interpret.
Couple with SOLAμ SPE Plates for Desalting
When used with the SMART Digest kits for desalting, Thermo Scientific™ SOLAμ™ SPE solid phase extraction plates provide a fast, easy-to-use, robust, and easily automatable sample preparation solution for analysis of biomolecules.