Inclusion Body Solubilization Reagent
Inclusion Body Solubilization Reagent
Thermo Scientific™

Inclusion Body Solubilization Reagent

The Thermo Scientific Inclusion Body Solubilization Reagent effectively retrieves recombinant proteins that form insoluble aggregates during bacterial overexpression.Features of InclusionRead more
Have Questions?
Catalog NumberQuantity
78115100 mL
Catalog number 78115
Price (USD)
262.65
Online Exclusive
284.00
Save 21.35 (8%)
Each
-
Add to cart
Quantity:
100 mL
Recurring order eligible. Learn more »
Request bulk or custom format
Price (USD)
262.65
Online Exclusive
284.00
Save 21.35 (8%)
Each
Add to cart
Ask our AI about this Product
The Thermo Scientific Inclusion Body Solubilization Reagent effectively retrieves recombinant proteins that form insoluble aggregates during bacterial overexpression.

Features of Inclusion Body Solubilization Reagent :

• Achieves the essential first step for subsequent insoluble protein refolding procedures
• One easy- and ready-to-use solution; no preparation needed
• Compatible with Thermo Scientific Pierce Coomassie Plus Protein Assay Kit (Part No. 23236) to determine degree of expression
• Proprietary denaturant can be easily removed by dialysis so that SDS-PAGE can be performed
• Can accommodate additional components such as reducing agents or chelating agents

The Inclusion Body Solubilization Reagent is a ready-to-use aqueous solution of denaturant that helps to dissolve aggregated proteins (inclusion bodies) that have been separated from the soluble fraction of bacterial cell lysates. The reagent is designed to retrieve expressed protein in soluble form after lysis and extraction with reagents such as Thermo Scientific B-PER Bacterial Protein Extraction Reagent. Bacteria are widely used for recombinant protein expression. However, 70%-80% of proteins produced by recombinant techniques in E. coli form inclusion bodies (i.e., protein aggregates). Once these aggregates are formed, it is very difficult to solubilize them. A proprietary denaturant contained in this reagent provides the most effective means for solubilizing aggregated proteins. Additional components, such as a reducing agent and a chelating agent, may be added to the reagent, depending on the particular application.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity100 mL
Reagent TypeChaotrope, Denaturant
FormLiquid
Product TypeSolubilization Reagent
Unit SizeEach
Contents & Storage
Upon receipt store at room temperature.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Los clientes que vieron este artículo también vieron



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
3213943Certificate of AnalysisJun 06, 202578115
ZJ400276Certificate of AnalysisOct 23, 202478115
ZB374895Certificate of AnalysisFeb 14, 202478115
YE365001Certificate of AnalysisJun 08, 202378115
XJ359193Certificate of AnalysisNov 08, 202278115
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection