Neuronal Culture Media Supplement (50X) for Pierce™ Primary Cell Isolation Kits
Neuronal Culture Media Supplement (50X) for Pierce™ Primary Cell Isolation Kits
Thermo Scientific™

Neuronal Culture Media Supplement (50X) for Pierce™ Primary Cell Isolation Kits

Neuronal Culture Media Supplement is a proprietary reagent that has been formulated for optimal growth of primary mouse and ratRead more
Have Questions?
Catalog NumberQuantity
8828610 mL
Catalog number 88286
Price (EUR)
207,00
Each
-
Add to cart
Quantity:
10 mL
Recurring order eligible. Learn more »
Request bulk or custom format
Price (EUR)
207,00
Each
Add to cart
Ask our AI about this Product
Neuronal Culture Media Supplement is a proprietary reagent that has been formulated for optimal growth of primary mouse and rat neurons. The supplement has been formulated for use with Neuronal Culture Medium (Product No. 88283) and is a component of the Thermo Scientific™ Pierce™ Primary Neuron Isolation Kit (Product No. 88280).

More Product Data
An efficient method for the isolation of highly functional primary neurons

Related Products
Pierce™ Primary Neuron Isolation Kit
Neuronal Culture Medium for Pierce™ Primary Cell Isolation Kits
Neuron Isolation Enzyme (with papain) for Pierce™ Primary Cell Isolation Kits
N-PER™ Neuronal Protein Extraction Reagent
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeNeuronal Cells (Pre-Natal), Primary Cells
Quantity10 mL
Target SpeciesRat, Mouse
FormLiquid
Product LinePierce™
Product TypePrimary Cell Isolation Media Supplement
Unit SizeEach
Contents & Storage
Stability: 1 year at -20°C
Avoid repeated freeze-thaws. Aliquot after initial thaw and store at -20°C.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
79511910Certificate of AnalysisJun 04, 202588286
ZL4590442Certificate of AnalysisJan 01, 202588286
ZI4464911Certificate of AnalysisSep 12, 202488286
ZH4444311Certificate of AnalysisAug 27, 202488286
ZC4264001Certificate of AnalysisMar 31, 202488286
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection