Finntip™ Stepper Pipette Tips
Due to high demand, order processing and delivery times for non-filtered tips may be longer than usual. Compare options for filtered tips »
Finntip™ Stepper Pipette Tips
Thermo Scientific™
Finntip™ Stepper Pipette Tips
Integrate Thermo ScientificTM FinntipTM Stepper Pipette Tips with the Thermo ScientificTM FinnpipetteTM Stepper Pipetter.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Sterility
94041831250 μLSterile
9404170500 μLNon-sterile
9404173500 μLSterile
94041801250 μLNon-sterile
94041902.5 mLNon-sterile
94041932.5 mLSterile
94042005 mLNon-sterile
94042035 mLSterile
940421012.5 mLNon-sterile
940421312.5 mLSterile
940422025 mLNon-sterile
940422325 mLSterile
940423050 mLNon-sterile
940423350 mLSterile
Catalog number 9404183
Price (USD)
113.40
Online Exclusive
126.00
Save 12.60 (10%)
Case of 50
In stock
Add to cart
Volume (Metric):
1250 μL
Sterility:
Sterile
Recurring order eligible. Learn more »
Price (USD)
113.40
Online Exclusive
126.00
Save 12.60 (10%)
Case of 50
Add to cart
Ask our AI about this Product
Integrate Thermo Scientific™ Finntip™ Stepper Pipette Tips with the Thermo Scientific™ Finnpipette™ Stepper Pipetter.
  • Nonsterile tips are bulk packaged; sterile tips are individually wrapped
Specifications
Certifications/ComplianceISO 9001, ISO 14001
Volume (Metric) Dispensing25, 50, 75, 100, 125 μL
FilteredNon-filtered
For Use With (Equipment)Thermo Scientific FinnPipette Stepper
FormatIndividually Wrapped
Product LineFinnTip
SterilitySterile
Volume (Metric)1250 μL
Unit SizeCase of 50
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Customers who viewed this item also viewed
Documents & Downloads
Certificates
Lot #Certificate TypeDateCatalog Number(s)
2410616Certificate of ComplianceJun 06, 20259404203
2330015Certificate of ComplianceMay 22, 20259404203
2202619Certificate of ComplianceMay 22, 20259404173
2410917Certificate of ComplianceMay 22, 20259404180
2202619Certificate of ComplianceMay 22, 20259404170
5 results displayed, search above for a specific certificate
Share catalog number, name or link