CTS™ GlutaMAX™ Supplement
CTS™ GlutaMAX™ Supplement
CTS™ GlutaMAX™ Supplement
CTS™ GlutaMAX™ Supplement
CTS™ GlutaMAX™ Supplement
Gibco™

CTS™ GlutaMAX™ Supplement

Green features
CTS™ GlutaMAX™ Supplement is an animal origin free (AOF) dipeptide alternative to L-glutamine, with increased stability that improves cell health. CTS™ GlutaMAX™ Supplement can be used as a direct substitute for L-glutamine at equimolar concentrations in mammalian and stem cell culture with minimal or no adaptation.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityFormat
A1286001100 mLBottle
A4737001100 mLBag - BioProcess Container (BPC)
A47370031 LBag - BioProcess Container (BPC)
Catalog number A1286001
Price (USD)
-
Quantity:
100 mL
Format:
Bottle
Ask our AI about this Product

CTS GlutaMAX Supplement is an animal origin free (AOF) dipeptide alternative to L-glutamine, with increased stability that improves cell health. CTS GlutaMAX Supplement can be used as a direct substitute for L-glutamine at equimolar concentrations in mammalian and stem cell culture with minimal or no adaptation.

Gibco CTS products are designed for use in manufacturing of cell or gene products, supporting your transition from research to clinical applications.

Compared to L-glutamine, CTS GlutaMAX Supplement:

  • Minimizes toxic ammonia build-up - Unlike L-glutamine, CTS GlutaMAX Supplement does not spontaneously break down to form ammonia. This prevents waste build-up and maintains a fresh supply of L-glutamine during long-term culture.
  • Improves growth efficiency and performance of mammalian cell cultures - Lower levels of ammonia result in improved cell viability. In addition, culture conditions are more controlled due to the consistent concentration of CTS GlutaMAX Supplement that is not possible with L-glutamine.
  • Remains stable across a wide range of temperatures - CTS GlutaMAX Supplement retains full functionality at room temperature or 2 to 8°C, for convenient storage and handling.

Closed systems compatible

This size of CTS GlutaMAX Supplement is provided in bioprocess container format with the chamber constructed from Aegis 5-14 film. Sterile, weldable C-Flex™ tubing with a female luer lock provides versatile connection options for integrating into a closed system manufacturing process.

CTS quality

The Gibco CTS product line enables you to reduce your burden in qualifying reagents during your transition from research to clinical applications.

Products are manufactured at a site that uses methods and controls that conform to cGMP for medical devices, 21 CFR Part 820. Our FDA-registered manufacturing sites are ISO 13485- and ISO 9001 to certified. Gibco CTS products follow USP <1043> ancillary materials for cell, gene, and tissue-engineered products within the responsibilities applicable to a supplier.

Gibco CTS products are of high quality and are supplied with harmonized documentation such as certificates of analysis, certificates of origin, and convenient access to our drug master file (DMF).

For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.
Specifications
Cell TypeMammalian and stem cell culture
Concentration100X
For Use With (Application)Cell and Gene Therapy Research, Development and Manufacturing
FormatBottle
Green FeaturesSustainable packaging
Manufacturing QualityISO 13485, MDSAP, FDA-registered, 21 CFR 820
Quantity100 mL
Shelf Life24 months from date of manufacture
Shipping ConditionRoom Temperature
ClassificationXeno-free
FormLiquid
Product TypeSupplement
SterilitySterile-filtered
Unit SizeEach
Contents & Storage
Storage Conditions: Room Temperature
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Figures

Kunden, die diesen Artikel ansahen, interessierten sich auch für



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
2898937Certificate of AnalysisJun 29, 2025A4737001
3221703Certificate of AnalysisJun 01, 2025A4737001
3161124Certificate of AnalysisMay 01, 2025A1286001
3046103Certificate of AnalysisApr 04, 2025A1286001
2876394Certificate of AnalysisMar 30, 2025A4737001
5 results displayed, search above for a specific certificate

Safety Data Sheets

Limited Use Label Licenses (LULL)

'

Notice to Purchaser: The purchase of this product conveys to the purchaser the limited, non- transferable right to use the purchased amount of the product to (a) perform internal research for the sole benefit of the purchaser; (b) manufacture protein (or other biological material) for resale; and (c) perform research or manufacturing services conducted by the purchaser on a fee for service or contract basis for or on behalf of third parties. However, the purchaser may transfer this product, its components, or materials made using this product to a third party (including contract research/manufacturing organizations), provided that each such third party agrees in writing to use such product, components, or materials solely on behalf of the purchaser, and such third party is restricted from further transferring any such product, components, or materials to any individual or entity other than the purchaser. No additional rights are granted. By purchasing this product, the purchaser agrees not to: (1) resell the product in any form; (2) use the product as a therapeutic agent or diagnostics test component; (3) reverse engineer the product or cause the product to be reverse engineered; or (4) use the product for purposes other than what is indicated in this Limited Use Label License. Life Technologies is not aware of Intellectual Property ("IP") that would be infringed by the manufacture or sale of its products. Customers are urged to perform an IP search and analysis specific to their manufacturing processes and target biologic products. Should that analysis require information about Life Technologies products, Life Technologies will provide to the customer's IP counsel the information necessary to analyze any relevant IP in a manner that protects Life Technologies proprietary information. The purchaser is responsible for obtaining all regulatory approvals necessary for any therapeutic or diagnostic use of the protein (or biological material) manufactured using this product. For information on obtaining additional rights, please contact outlicensing@thermofisher.com, Licensing and Commercial Supply, Thermo Fisher Scientific, 5823 Newton Drive, Carlsbad, CA, 92008, United States.

'

Frequently asked questions (FAQs)

The Gibco Cell Therapy Systems (CTS) portfolio of cell and gene therapy products are GMP manufactured, safety tested, and backed by regulatory documentation to support your transition from discovery through clinical and commercial manufacturing. Through our CTS solutions, we are committed to helping customers streamline therapeutic development, minimize risk, and ease the burden on their quality systems. Learn more here.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

The formulation and manufacturing process are the same for all three products. The main difference is in the “intended use” statement. CTS GlutaMAX-I Supplement (Cat. No. A1286001) and CTS GlutaMAX Supplement (Cat. No. A4737001) have an intended use statement of “For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals”. GlutaMAX Supplement (Cat. No. 35050061) has an intended use statement of “For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals”. The catalog number to use will depend on the specific application.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Yes. If you suspect that this is the case, remove the medium and add fresh medium. Alternatively, you can supplement medium with growth-promoting components. It is also possible to substitute GlutaMax I or II for glutamine in the medium to prevent glutamine exhaustion.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

In all media containing GlutaMAX supplement dipeptides as a substitute for L-glutamine, concentration is equimolar with the L-glutamine in the original formulation.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Citations & References (1)

Citations & References
Abstract
Different culture conditions modulate the immunological properties of adipose stem cells.
Authors:Patrikoski M, Sivula J, Huhtala H, Helminen M, Salo F, Mannerström B, Miettinen S
Journal:
PubMed ID:25122689
The potential of human adipose stem cells (ASCs) for regenerative medicine has received recognition owing to their ease of isolation and their multilineage differentiation capacity. Additionally, low immunogenicity and immunosuppressive properties make them a relevant cell source when considering immunomodulation therapies and allogeneic stem cell treatments. In the current study, ... More
1 total citations

Andere Produkte



Share catalog number, name or link

1x1 image pixel for data collection