7-AAD (7-Aminoactinomycin D)
7-AAD (7-Aminoactinomycin D)
Invitrogen™

7-AAD (7-Aminoactinomycin D)

7-aminoactinomycin D (7-AAD) is a fluorescent intercalator that undergoes a spectral shift upon association with DNA. 7-AAD/DNA complexes can beRead more
Have Questions?
Catalog NumberQuantity
A13101 mg
Catalog number A1310
Price (KRW)
250,000
Online offer
Ends: 30-Jun-2025
294,000
Save 44,000 (15%)
Each
-
Add to cart
Quantity:
1 mg
Recurring order eligible. Learn more »
Price (KRW)
250,000
Online offer
Ends: 30-Jun-2025
294,000
Save 44,000 (15%)
Each
Add to cart
Ask our AI about this Product

7-aminoactinomycin D (7-AAD) is a fluorescent intercalator that undergoes a spectral shift upon association with DNA. 7-AAD/DNA complexes can be excited by the 488 nm laser and has an emission maxima of 647 nm, making this nucleic acid stain useful for multicolor fluorescence microscopy and flow cytometry. 7-AAD appears to be generally excluded from live cells, but can be used with cells that have been fixed and permeabilized. 7-AAD has been used for cell cycle analysis by flow cytometry. 7-AAD also binds selectively to GC regions of DNA yielding a distinct banding pattern in polytene chromosomes and chromatin for use in chromosome banding studies.

To make a concentrated stock solution from 1–2 mM, dissolve 7-AAD in either high quality anhydrous DMSO or DMF. Store frozen, desiccated and protected from light.

View a selection guide for all nonfixable viability dyes for flow cytometry.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection MethodFluorescence
Dye TypeCell-Impermeant
Emission647 nm
Excitation Wavelength Range488 nm
For Use With (Equipment)Fluorescence Microscope, Flow Cytometer
FormSolid
Molecular Weight (g/mol)1270.45
Quantity1 mg
Shipping ConditionRoom Temperature
SolubilityDMSO (Dimethylsulfoxide)
Label TypeFluorescent Dye
Product TypeNuclear Labeling
SubCellular LocalizationNucleic Acids, Nucleus
Unit SizeEach
Contents & Storage
Contains 1 vial of 7-AAD (1 mg).

Store in freezer (-5°C to -30°C) and protect from light.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Figures

Fluorescence spectra

Fluorescence spectra

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
2942311Certificate of AnalysisJul 12, 2024A1310
2729835Certificate of AnalysisSep 19, 2023A1310
2557916Certificate of AnalysisNov 11, 2022A1310
2525788Certificate of AnalysisOct 06, 2022A1310
2383850Certificate of AnalysisSep 24, 2021A1310
5 results displayed, search above for a specific certificate

Safety Data Sheets

Frequently asked questions (FAQs)

The following cell health and viability assays can be performed by flow cytometry :

-Apoptosis Assays:
Membrane Asymmetry: Annexin V is a member of a family of structurally related proteins that bind phospholipids in the presence of Ca2+. Annexin V binds several phospholipids, but shows highest affinity for phosphatidylserine.
Phosphatidylserine is normally found in the inner leaflet of the cell membrane; however, in the early stages of apoptosis, phosphatidylserine is observed to translocate to the outer leaflet. This translocation makes phosphatidylserine available for annexin V binding in the presence of Ca2+ containing incubation buffer. Cells undergoing apoptosis will stain with annexin V, while normal cells will not. annexin V is available conjugated with a wide range of fluorophores.

Mitochondrial Health: A distinctive feature of the early stages of apoptosis is the disruption of the mitochondria, including changes in membrane and redox potential. We exclusively offer a number of fluorescent probes for analyzing mitochondrial activity in live cells by flow cytometry, with minimal disruption of cellular function.

The MitoProbe family of mitochondrial stains (MitoProbe DiOC2(3) Assay Kit, Cat. No. M34150, MitoProbe JC-1 Assay Kit, Cat. No. M34152, and MitoProbe DiIC1(5) Assay Kit, Cat. No. M34151) provides quick, easy, and reliable flow cytometric detection of the loss of mitochondrial membrane potential that occurs during apoptosis.

Caspase Activity: The CellEvent Caspase-3/7 Green Flow Cytometry Assay Kit (Cat. No. C10427) enables flow cytometric detection of activated caspase-3 and caspase-7 in apoptotic cells. The kit includes the novel fluorogenic substrate CellEvent Caspase-3/7 Green Detection Reagent which targets the recognition sequence for activated caspase-3 and caspase-7, as well as SYTOX AADvanced Dead Cell Stain.

DNA Fragmentation: The later stages of apoptosis are characterized by changes in nuclear morphology, including DNA fragmentation, chromatin condensation, degradation of nuclear envelope, nuclear blebbing, and DNA strand breaks. DNA fragmentation that occurs during apoptosis produces DNA strand breaks, and can be analyzed using TUNEL (terminal deoxynucleotidyl transferase dUTP nick end labeling) assays. The APO-BrdU TUNEL assay (Cat. No. A23210) is a two-color assay for labeling DNA breaks and total cellular DNA to detect apoptotic cells by imaging or flow cytometry.

Nuclear Chromatin Condensation: The later stages of apoptosis are characterized by changes in nuclear morphology, including DNA fragmentation, chromatin condensation, degradation of nuclear envelope, nuclear blebbing, and DNA strand breaks. Cells undergoing apoptosis display an increase in nuclear chromatin condensation. As the chromatin condenses, cell-permeable nucleic acid stains becomes hyperfluorescent, thus enabling the identification of apoptotic cells when combined with a traditional dead-cell stain. The Vybrant Apoptosis Assay Kit #5, Hoechst 33342/Propidium Iodide (Cat. No. V13244) provides a rapid and convenient assay for apoptosis based on fluorescence detection of the compacted state of the chromatin in apoptotic cells. The Chromatin Condensation & Membrane Permeability Dead Cell Apoptosis Kit with Hoechst 33342, YO-PRO-1, and PI dyes, for flow cytometry (Cat. No. V23201) detects apoptotic cells with changes in nuclear chromatin condensation and plasma membrane permeability.

-Cell Cycle Analysis:
Live cell assays: The Vybrant DyeCycle family of dyes offers robust fluorescent dyes for live-cell cycle analysis with limited cytotoxicity using 405 nm (Cat. No. V35003), 488 nm (Cat. No. V35004), 532 nm (Cat. No. V35005), or 633 nm (Cat. Nos. V10309 and V10273) excitation. The dyes have low cytotoxicity, allowing stained cells to be sorted and otherwise cultured or assessed with functional assays after staining.

Fixed cell assays: Analyzing cell cycle using FxCycle Violet Stain (Cat. No. F10347), SYTOX AADvanced Dead Cell Stain Kit (Cat. No. S10349) or FxCycle Far Red Stain (Cat. No. F10348) allows for multiple color options for simplified fixed cell cycle analysis.

-Cell Proliferation:
Dye dilution assays for cell proliferation: Dye dilution assays for cell proliferation rely on cell membrane–permeant fluorescent molecules. Upon entry into the cell, the dye will covalently bind to amine groups on proteins, resulting in long-term dye retention within the cell. Through subsequent cell divisions, each daughter cell receives approximately half the fluorescence of the parent. Analysis of the fluorescence intensities of cell populations by flow cytometry enables determination of the number of generations through which a cell or population has progressed since the label was applied. CellTrace fluorescent stains can be used without affecting morphology or physiology to trace generations in vivo or in vitro. There is no known effect on proliferative ability or biology of cells and they are well retained in cells for several days post-stain. Available kits for flow cytometry include CellTrace CFSE Cell Proliferation Kit (Cat. No. C34554), CellTrace Violet Cell Proliferation Kit (Cat. No. C34557), and CellTrace Far Red Cell Proliferation Kit (Cat. No. C34564).

DNA Synthesis Assays: Measuring the synthesis of new DNA is a precise way to assay cell proliferation in individual cells or in cell populations. DNA synthesis–based cell proliferation assays measure the rate of new DNA synthesis based on incorporation of modified nucleosides. The Click-iT Plus EdU cell proliferation assay utilizes the power of click chemistry and the modified nucleoside EdU to provide a superior alternative to BrdU staining for detecting and quantitating newly synthesized DNA. The Click-iT Plus EdU cell proliferation assay is available with Pacific Blue (Cat. No. C10636), Alexa Fluor 488 (Cat. Nos. C10632 and C10633), and Alexa Fluor 647 (Cat. Nos. C10634 and C10635).

-Viability Assays:
Dead cells often give false positive results, as they tend to bind non-specifically to many reagents. Therefore, removing dead cells from your flow cytometry data is a critical step to help ensure accurate results and analysis.

Non-fixable Membrane Permeability Stains: SYTOX Dead Cell Stains (Cat. Nos. S34857, S34860, S34861, S34859, and S34862) do not cross intact cell membranes, and they exhibit increased fluorescence upon dsDNA binding, making them some of our most brilliant dead cell stains. Cell-impermeant classic DNA-binding dyes include propidium iodide (Cat. No. P21493) and 7-AAD (Cat. No. A1310). Both of these dyes have been used extensively for viability assays in flow cytometry. CellTrace Calcein AM dyes can be passively loaded into adherent and nonadherent cells. These cell-permeant esterase substrates serve as viability probes that measure both enzymatic activity, which is required to activate their fluorescence, and cell membrane integrity, which is required for intracellular retention of their fluorescent products. Available with blue (Cat. No. C34853), violet (Cat. No. C34858), and green (Cat. No. C34852) fluorescence, these dyes are ideal for short-term staining of live cells and can be used in multiplexed flow cytometry experiments.

Fixable Viability Stains: The LIVE/DEAD Fixable Dead Cell Stains are fixable viability dyes that help to ensure accurate assessment of cell viability in samples after fixation and/or permeabilization. LIVE/DEAD Fixable Dead Cell Stain Kits are based on the reaction of a fluorescent reactive dye with cellular proteins (amines). These dyes cannot penetrate live-cell membranes, so only cell-surface proteins are available to react with the dye, resulting in dim staining. The reactive dye can permeate the damaged membranes of dead cells and stain both the interior and exterior amines, resulting in more intense staining. LIVE/DEAD Fixable Dead Cell Stain Kits are available in eight single-channel colors available for UV, 405, 488, 532, 561, or 633 nm lasers in three packaging sizes to match your experiment.



Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Citations & References (269)

Citations & References
Abstract
Authors:
Journal:
PubMed ID:18258751
A Glu-496 to Ala polymorphism leads to loss of function of the human P2X7 receptor.
Authors:Gu BJ,Zhang W,Worthington RA,Sluyter R,Dao-Ung P,Petrou S,Barden JA,Wiley JS
Journal:The Journal of biological chemistry
PubMed ID:11150303
Strategies for phenotyping apoptotic peripheral human lymphocytes comparing ISNT, annexin-V and 7-AAD cytofluorometric staining methods.
Authors:Lecoeur H,Ledru E,Prévost MC,Gougeon ML
Journal:Journal of immunological methods
PubMed ID:9461328
The present article compares the reliability of four previously described cytofluorometric methods of apoptosis quantification for phenotyping apoptotic human lymphocytes. Each of these assays detects distinct cellular alterations of the apoptotic process. Alteration in plasma membrane integrity can be evaluated following 7-AAD incorporation and the translocation of phosphatidylserine from the ... More
Features of medullary thymic epithelium implicate postnatal development in maintaining epithelial heterogeneity and tissue-restricted antigen expression.
Authors:Gillard GO, Farr AG
Journal:J Immunol
PubMed ID:16670287
'Although putative thymic epithelial progenitor cells have been identified, the developmental potential of these cells, the extent of medullary thymic epithelium (mTEC) heterogeneity, and the mechanisms that mediate the expression of a wide range of peripheral tissue-restricted Ags (TRAs) by mTECs remain poorly defined. Here we have defined several basic ... More
A human gene coding for a membrane-associated nucleic acid-binding protein.
Authors:Siess DC, Vedder CT, Merkens LS, Tanaka T, Freed AC, McCoy SL, Heinrich MC, Deffebach ME, Bennett RM, Hefeneider SH
Journal:J Biol Chem
PubMed ID:10938276
'Studies to clone a cell-surface DNA-binding protein involved in the binding and internalization of extracellular DNA have led to the isolation of a gene for a membrane-associated nucleic acid-binding protein (MNAB). The full-length cDNA is 4.3 kilobases with an open reading frame of 3576 base pairs encoding a protein of ... More
269 total citations

Other products to consider



Share catalog number, name or link