High Resolution Melt (HRM) App
Try our HRM App for free by visiting our Apps Page (opens a new window) on Thermo Fisher Cloud.
High Resolution Melt (HRM) App
Applied Biosystems™
High Resolution Melt (HRM) App
The High Resolution Melt (HRM) App hosted on Thermo Fisher Cloud is a tool for HRM analysis, which is aRead more
Have Questions?
Catalog NumberQuantity
A295131 license
Catalog number A29513
Price (USD)
-
Quantity:
1 license
Ask our AI about this Product
The High Resolution Melt (HRM) App hosted on Thermo Fisher Cloud is a tool for HRM analysis, which is a post-PCR analysis method used to identify variation in nucleic acid sequences. The method is based on detecting small differences in PCR melting (dissociation) curves. It is enabled by high-brightness, dsDNA-binding dyes used in conjunction with real-time PCR instrumentation that has precise temperature ramp control, advanced data capture capabilities, and access to software designed specifically for HRM analysis.
Key product features:
Minimal user input—app automatically applies default/saved analysis settings upon opening an experiment file
High accuracy—clustering algorithm accurately distinguishes control and variant genotypes
High sensitivity—app allows input of expected number of clusters for difficult-to-detect SNPs (class 4)
Efficiency—analyze multiple runs simultaneously and visualize the data
Minimize user input and easily recall analysis settings
The HRM App automatically applies default or saved analysis settings upon opening an experiment file, which allows you to achieve results with minimal input.
Realize high accuracy and sensitivity
The HRM App uses the same clustering algorithm as the desktop version, which helps to accurately distinguish control and variant genotypes, and allows the user to input the expected number of clusters for increased sensitivity for difficult SNP genotyping. It also offers data compatibility with the desktop version.
Achieve more with less work
With the HRM App, you can analyze multiple plates simultaneously. Unlike the desktop version, you can access an additional set of data review tools, such as those for amplification plots and multicomponent data.
App subscription
The following subscription plans are available at rates for 1 user or 10 users. (Please contact us if our available plans do not meet your needs.) A free trial is available for first-time users. The Terms and Conditions for Software Application Subscription Plans on the Thermo Fisher Cloud apply to each subscription plan described below:
Auto-renewal Monthly:This plan is charged month-to-month. Each month is paid up front and the subscription auto-renews every month. This plans provides the most flexibility with the lowest up-front costs.
Auto-renewal Annual: This plan is charged once a year. Each year is paid up front and auto-renews every year. This plan is more cost-effective than the month-to-month plan.
Termed Contract for One Year or Five Years: With this plan the entire term of the contract is paid up front and does not renew. This plan is most suitable when a capital expenditure is more desirable than smaller recurring payments.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)QuantStudio™
Quantity1 license
Shipping ConditionRoom Temperature
Software TypeWeb Based Software
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Customers who viewed this item also viewed
Documents & Downloads
Certificates
0 results displayed, search above for a specific certificate
Frequently asked questions (FAQs)
Continuous melt curve: increases the temperature by the ramp increment (degrees C/sec) so that the temperature will change at a constant rate set in the protocol.
Step and Hold melt curve: increases the temperature by the ramp increment (degrees C) and then holds at that temperature for the time specified by the user.
The amount of data collected will depend on the filter(s) selected. The more filters you use, the longer the acquisition will take and the fewer data points will be collected.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.
To create a new variant call group in the HRM app on Thermo Fisher Connect, follow these steps:
  1. Select the 'Quality Control & Results' tab.
  2. In the 'Quality Control & Results' pane, select your experiment of interest.
  3. Select the 'Derivative melt plot'.
  4. Click 'Actions', and then select 'Manual Call'.
  5. From the 'Manual Call' dialog box, you have two options:
    1. To assign the sample to an existing variant call: click 'Select Existing', choose the appropriate call from the 'Group' drop-down menu, and then click 'OK'.
    2. To create a new variant call: click 'Create New', enter a name for the new call in the 'Group' field, select a color, and then click 'OK'.
  6. Repeat the above steps to assign more manual calls.
  7. Click 'Analyze' to reanalyze the data using the manual calls.
Find additional tips, troubleshooting help, and resources within our High Resolution Melt Support Center.
Other products to consider
Share catalog number, name or link
1x1 image pixel for data collection