BenchStable™ DMEM/F12
BenchStable™ DMEM/F12
Gibco™

BenchStable™ DMEM/F12

Green features
Gibco DMEM/F-12 (Dulbecco's Modified Eagle Medium/Nutrient Mixture F-12) is a widely used basal medium for supporting the growth of manyRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
A419200210 x 500 mL
A4192001500 mL
Catalog number A4192002
Price (USD)
550.65
Online Exclusive
596.00
Save 45.35 (8%)
Each
-
Add to cart
Quantity:
10 x 500 mL
Recurring order eligible. Learn more »
Price (USD)
550.65
Online Exclusive
596.00
Save 45.35 (8%)
Each
Add to cart
Ask our AI about this Product

Gibco DMEM/F-12 (Dulbecco's Modified Eagle Medium/Nutrient Mixture F-12) is a widely used basal medium for supporting the growth of many different mammalian cells. Cells successfully cultured in DMEM/F-12 include MDCK, glial cells, fibroblasts, human endothelial cells, and rat fibroblasts. We offer a variety of DMEM/F-12 modifications for a range of cell culture applications. Find the right formulation using the media selector tool.

BenchStable DMEM/F-12 has been engineered for storage at room temperature while supporting cellular proliferation and maximum cell densities comparable to conventional DMEM/F12 when supplemented with ≥10% fetal bovine serum (FBS).

This DMEM/F-12 is modified as follows:
• Optimized for room temperature stability
• With GlutaMAX, Phenol Red
• Without HEPEPS

Using BenchStable DMEM/F-12
DMEM/F-12 is a 1:1 mixture of DMEM and Ham's F-12. This formulation combines DMEM's high concentrations of glucose, amino acids, and vitamins with F-12's wide variety of components. DMEM/F-12 uses a sodium bicarbonate buffer system and therefore requires a 5–10% CO2 environment to maintain physiological pH. DMEM/F-12 with GlutaMAX supplement minimizes toxic ammonia build-up and improves cell viability and growth in an easy-to-use format. BenchStable DMEM/F-12 contains no proteins, lipids, or growth factors and so requires supplementation with ≥10% fetal bovine serum (FBS).

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell LineMDCK, glial cells, fibroblasts, human endothelial cells, and rat fibroblasts
Concentration1 X
Green FeaturesEnergy efficient, Fewer resources used, Sustainable packaging
Manufacturing QualitycGMP-compliant under the ISO 9001: 2015 standard
Product LineBenchStable™
Product TypeDMEM (Dulbecco's Modified Eagle Medium)/F-12
Quantity10 x 500 mL
Shelf Life12 Months From Date of Manufacture
Shipping ConditionRoom Temperature
ClassificationAnimal Origin-free
FormLiquid
Serum LevelStandard Serum Supplementation
SterilitySterile-filtered
With AdditivesHigh Glucose, GlutaMAX, Phenol Red, Sodium Pyruvate
Without AdditivesNo HEPES
Unit SizeEach
Contents & Storage
Store at room temperature. Protect from light.
four-plex-image-COS7-live-cell promo pod

Follow the 5 steps to publication-quality fixed-cell imaging.
Download your free 5-step guide today ›

Have questions about this product? Ask our AI assisted search.
Will the shelf-life of BenchStable media improve if stored at lower temperature (2-8 degrees C)?
My BenchStable medium arrived partially/completely frozen. Is it still stable?
What is the recommended usage timeframe for BenchStable media after serum supplementation?
Is degradation of BenchStable media due to light exposure only limited to fluorescent light?
How should I store BenchStable media? Can they be stored at 4 degrees C?
+Show more FAQs for this product
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Figures

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
3102901Certificate of AnalysisApr 01, 2025A4192001
3002241Certificate of AnalysisNov 06, 2024A4192001
2975765Certificate of AnalysisAug 07, 2024A4192001
2911106Certificate of AnalysisJun 03, 2024A4192001
2854367Certificate of AnalysisMar 14, 2024A4192001
5 results displayed, search above for a specific certificate

Safety Data Sheets

Frequently asked questions (FAQs)

We recommended supplementing BenchStable media with at least 10% FBS for best results. If your culture method calls for greater than 10% FBS supplementation using our other existing media, we recommend that you use the same concentration with BenchStable media.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

We recommend supplementing BenchStable media with at least 10% FBS for best results. We have run tests on limited cell types using 5% FBS concentration and have not observed compromised performance. Please contact Tech Support at techsupport@thermofisher.com to ensure that your specific application allows for 5% serum supplementation.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

We have tested our BenchStable media for growth and morphology on the following cell lines: A549, CHO-K1, HEK293, HeLa, HL-60, Jurkat, MCF7, MDA-MB-231, Neuro2A, SH-SY5Y, and THP-1.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

At this time, we do not offer BenchStable media without phenol red.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

The shelf life of BenchStable media is unchanged when stored cold (2-4 degrees C) or ambient (15-30 degrees C).

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Citations & References (3)

Citations & References
Abstract
Non-viral-mediated gene transfer of OX40 ligand for tumor immunotherapy.
Authors:Rakitina OA,Kuzmich AI,Bezborodova OA,Kondratieva SA,Pleshkan VV,Zinovyeva MV,Didych DA,Sass AV,Snezhkov EV,Kostina MB,Koksharov MO,Alekseenko IV
Journal:Frontiers in immunology
PubMed ID:39007148
BACKGROUND: Immune checkpoint blockade (ICB) is rapidly becoming a standard of care in the treatment of many cancer types. However, the subset of patients who respond to this type of therapy is limited. Another way to promote antitumoral immunity is the use of immunostimulatory molecules, such as cytokines or T ... More
Adaptation of the Kirkstall QV600 LLI Microfluidics System for the Study of Gastrointestinal Absorption by Mass Spectrometry Imaging and LC-MS/MS.
Authors:Clench MR
Journal:Pharmaceutics
PubMed ID:35214096
Impact of Metformin Treatment on Human Placental Energy Production and Oxidative Stress.
Authors:Aiken CE
Journal:Frontiers in cell and developmental biology
PubMed ID:35784487
3 total citations

Other products to consider



Share catalog number, name or link

1x1 image pixel for data collection