Pierce™ Pyrogen-Free Plate Strips with Frame, 12 x 8 well
Pierce™ Pyrogen-Free Plate Strips with Frame, 12 x 8 well
Thermo Scientific™

Pierce™ Pyrogen-Free Plate Strips with Frame, 12 x 8 well

These endotoxin-free 12 x 8-well plates are supplied with one frame and containRead more
Have Questions?
Catalog NumberQuantity
A438885 plates, 1 frame
Catalog number A43888
Price (USD)
52.25
Each
In stock
Add to cart
Quantity:
5 plates, 1 frame
Recurring order eligible. Learn more »
Price (USD)
52.25
Each
Add to cart
Ask our AI about this Product
These endotoxin-free 12 x 8-well plates are supplied with one frame and contain <0.0005 EU/well endotoxin. The individually wrapped 8-well strips are ideal for assays of less than 96 tests at a time.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
DescriptionEndotoxin level <0.0005 EU/well. 12 strip x 8 well 96-well plate, strips are individual wrapped.
For Use With (Application)Chromogenic LAL Endotoxin Assay, Kinetic Chromogenic LAL Assay and Kinetic Turbidimetric LAL Assay
Quantity5 plates, 1 frame
FormatStrip Plate
SterilitySterile
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
0A20T52207Certificate of AnalysisJun 19, 2025A43888
0B30F01807Certificate of AnalysisApr 17, 2024A43888
0C91T52807Certificate of AnalysisNov 03, 2023A43888
0DC1F82507Certificate of AnalysisJul 29, 2023A43888
SPECSCertificate of AnalysisSep 20, 2022A43888
5 results displayed, search above for a specific certificate

Other products to consider



Share catalog number, name or link

1x1 image pixel for data collection