1x1 image pixel for data collection
Quant-iT™ Endotoxin Detection Assay Kit
Quant-iT™ Endotoxin Detection Assay Kit
Quant-iT™ Endotoxin Detection Assay Kit
Quant-iT™ Endotoxin Detection Assay Kit
Invitrogen™
Quant-iT™ Endotoxin Detection Assay Kit
The Invitrogen Quant-iT Endotoxin Detection Assay Kit is a sensitive and easy-to-use fluorescent endpoint assay for use on fluorescence microplateRead more
Promo ImagePromotion
Promo code:P5780456
Gene Expression Promo
From RNA Isolation kits, reverse transcriptases to analysis of your gene with qPCR - upto 35% on all Gene Expression workflow productsLearn More
Have Questions?
Catalog NumberQuantity
Q32892Promo Image160 reactions
Catalog number Q32892
Price (USD)
802.00
Each
Estimated availability date 05-Aug-2025
Add to cart
Quantity:
160 reactions
Recurring order eligible. Learn more »
Price (USD)
802.00
Each
Add to cart
Ask our AI about this Product
The Invitrogen Quant-iT Endotoxin Detection Assay Kit is a sensitive and easy-to-use fluorescent endpoint assay for use on fluorescence microplate readers that uses amebocyte lysates to quantify endotoxin in protein, peptide, antibody, or nucleic acid samples. Amebocyte lysates are widely used as a simple and sensitive assay for the detection of endotoxin lipopolysaccharide (LPS), the membrane component of gram-negative bacteria.
Features of the Quant-iT Endotoxin Detection Assay Kit include:
Highly sensitive with a broad range—detect as little as 0.01 EU/mL to 10.0 EU/mL
Flexible—suitable for wide range of samples, including DNA, RNA, plasmids, antibodies, and more
Easy to use—the simple workflow requires one incubation prior to measurement
Endotoxin levels in the samples are accurately determined using the included endotoxin standard of known concentration that is derived from E. coli strain O111: B4. Determining endotoxin levels is important to assess the efficiency of endotoxin removal methods and prevent endotoxic shock, inflammation, and/or sepsis in tissue culture cells and animals injected with endotoxin-contaminated proteins.
When endotoxin encounters the amebocyte lysate, a series of enzymatic reactions results in the activation of Factor C, Factor B, and pro-clotting enzyme. The activated enzyme catalyzes a cleavage event in the substrate to produce a strong fluorescent signal. After stopping the reaction, the resulting signal is measured on a fluorescence microplate reader (such as Varioskan LUX). The correlation between fluorescent signal is proportional to the endotoxin concentration in the sample.
Notes
• Compatible with Pierce Endotoxin-Free Plate Strips (Cat. No. A43888)
For Research Use Only. Not for use in diagnostic procedures.
Specifications
DetectsEndotoxin
FormatKit
Quantity160 reactions
Specimen TypeProtein, Antibody, Plasmid, DNA, RNA
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
How do I store the Quant-iT Endotoxin Detection Assay Kit?
What is considered an acceptable correlation coefficient, r value for the Quant-iT Endotoxin Detection Assay?
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Customers who viewed this item also viewed
Documents & Downloads
Certificates
Lot #Certificate TypeDateCatalog Number(s)
3142720Certificate of AnalysisMar 31, 2025Q32892
3021921Certificate of AnalysisMar 31, 2025Q32892
3110708Certificate of AnalysisMar 30, 2025Q32892
2978481Certificate of AnalysisAug 26, 2024Q32892
2916404Certificate of AnalysisMay 30, 2024Q32892
5 results displayed, search above for a specific certificate
Safety Data Sheets
Frequently asked questions (FAQs)
The US Pharmacopeia recommends performing this analysis using log transformed concentration and RFU data and then performing a linear regression fit to verify that the correlation coefficient, r, is greater than or equal to 0.980.
The entire kit can be stored at 2-8 degrees C for 6 months. Reconstituted Endotoxin standard solutions are stable for 4 weeks at 2-8 degrees C and reconstituted dye can be stored for 4 weeks at 2-8 degrees C. Reconstituted amebocyte lysate solution is stable for 1 week at -20 degrees C or colder if frozen immediately after reconstitution.
Share catalog number, name or link
1x1 image pixel for data collection