1x1 image pixel for data collection
FastAP Thermosensitive Alkaline Phosphatase (1 U/μL)
FastAP Thermosensitive Alkaline Phosphatase (1 U/μL)
Thermo Scientific™

FastAP Thermosensitive Alkaline Phosphatase (1 U/μL)

Thermo Scientific FastAP Thermosensitive Alkaline Phosphatase catalyzes the release of 5'- and 3'-phosphate groups from DNA, RNA, and nucleotides. ThisRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
EF0654300 units
EF06511,000 units
EF06525 x 1,000 units
Catalog number EF0654
Price (USD)
52.65
Online Exclusive
58.25
Save 5.60 (10%)
Each
In stock
Add to cart
Quantity:
300 units
Recurring order eligible. Learn more »
Request bulk or custom format
Price (USD)
52.65
Online Exclusive
58.25
Save 5.60 (10%)
Each
Add to cart
Ask our AI about this Product
Thermo Scientific FastAP Thermosensitive Alkaline Phosphatase catalyzes the release of 5'- and 3'-phosphate groups from DNA, RNA, and nucleotides. This enzyme also removes phosphate groups from proteins.

FastAP is a novel alkaline phosphatase, which is active in all Thermo Scientific restriction enzyme buffers as well as in PCR buffers. It dephosphorylates all types of DNA ends (blunt, 5'- and 3'-overhangs) in 10 minutes at 37°C. The enzyme is inactivated in 5 minutes at 75°C (see Figure 1 in Supporting Data). Therefore, removal of alkaline phosphatase is not required prior to ligation.

Highlights

Recombinant enzyme
Fast dephosphorylation—10 minutes at 37°C
Fast and complete inactivation—5 minutes at 75°C
Simultaneous digestion and dephosphorylation of vector DNA
100% active in restriction enzyme and PCR buffers
PCR clean-up in conjunction with Exo I
Protein dephosphorylation

One protocol for all types of DNA ends:
• 5'-overhangs
• 3'-overhangs
• blunt-ends
• single nucleotides

Applications

• Dephosphorylation of cloning vector DNA to prevent recircularization during ligation
• Simultaneous digestion and dephosphorylation of vector DNA
• PCR product clean-up: nucleotide degradation prior to sequencing of PCR product
• Dephosphorylation of nucleic acid 5'-termini prior to labeling with T4 Polynucleotide Kinase
• Other applications where dephosphorylation of DNA and RNA substrates is necessary
• Protein dephosphorylation

Note

• Binding of FastAP Thermosensitive Alkaline Phosphatase to DNA may result in a band shift in agarose gels. To avoid this, incubate samples with 6X DNA Loading Dye & SDS Solution at 65°C for 10 minutes, and chill on ice prior to electrophoresis.
• FastAP Thermosensitive Alkaline Phosphatase is active in all restriction enzyme buffers and may be added directly to digested DNA. Heat inactivation of the restriction enzyme before dephosphorylation reaction is not necessary.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Compatible BufferPCR Buffer, Enzyme Buffer
Product TypeAlkaline Phosphatase
Quantity300 units
Concentration1 U/μL
EnzymePhosphatase
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Frequently asked questions (FAQs)

Do I need to remove the FastAP Thermosensitive Alkaline Phosphatase prior to ligation?

No, removal is not necessary. The enzyme can be inactivated in 5 mins at 75 degrees C.

How long does the dephosphorylation take when using your FastAP Thermosensitive Alkaline Phosphatase?

Dephosphorylation takes place in approximately 10 mins at 37 degrees C.

Figures

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
3274593Certificate of AnalysisJul 07, 2025EF0652
3262849Certificate of AnalysisJun 17, 2025EF0652
3262553Certificate of AnalysisJun 17, 2025EF0651
3255904Certificate of AnalysisJun 09, 2025EF0651
3250744Certificate of AnalysisMay 30, 2025EF0654
5 results displayed, search above for a specific certificate

Safety Data Sheets

Other products to consider



Share catalog number, name or link