SUPERase·In™ RNase Inhibitor (20 U/μL)
SUPERase·In™ RNase Inhibitor (20 U/μL)
Invitrogen™

SUPERase·In™ RNase Inhibitor (20 U/μL)

SUPERase•In RNase Inhibitor is a protein-based inhibitor of non-human origin that noncovalently binds and inhibits the most common and troublesomeRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
AM26942500 U
AM269610,000 U
Catalog number AM2694
Price (EUR)
255,65
Online Exclusive
288,00
Save 32,35 (11%)
Each
-
Add to cart
Quantity:
2500 U
Request bulk or custom format
Price (EUR)
255,65
Online Exclusive
288,00
Save 32,35 (11%)
Each
Add to cart
Ask our AI about this Product
SUPERase•In RNase Inhibitor is a protein-based inhibitor of non-human origin that noncovalently binds and inhibits the most common and troublesome RNases, including RNase A, B, C, 1, and T1.

SUPERase•In RNase Inhibitor can be used in any application where RNase contamination could be problematic. Because it inhibits a broader range of RNases than traditional RNase inhibitors, SUPERase•In is the most effective RNase inhibitor available, providing a higher level of protection against degradation.

SUPERase•In does not interfere with other enzymes such as RNA polymerases, reverse transcriptase, or Taq DNA polymerase. Additionally, SUPERase•In is active up to 65°C and over a pH range of 5.5 to 8.5.

If you are plannning to use this product with standard antibody purification methods, please contact our technical support to discuss experimental design.

Unit definition
SUPERase•In RNase Inhibitor at 1 U/μL will block the degradation of 0.1 μg/μL labeled RNA by 2.5 pg/μL of RNase A, 2.5 pg/μL of RNase I, and 0.0075 U/μL of RNase T1, for 4 hours at 37°C, in 20 mM Tris-HCl, pH 7.5, 50 mM NaCl, 1 mM EDTA. Analysis is by denaturing PAGE. SUPERase•In is currently the only ribonuclease inhibitor for which the unit activity is defined by such a functional assay.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration50 mM
Product LineAmbion™, SUPERase•In™
Quantity2500 U
Shipping ConditionDry Ice
Unit SizeEach
Contents & Storage
Supplied in a single tube that should be stored at -20°C.

Storage buffer: 2 mM KH2PO4, 8 mM Na2HPO4,2.7 mM KCl, 137 mM NaCl, pH 7.4 in 50% glycerol
Have questions about this product? Ask our AI assisted search.
Can beta-mercaptoethanol (BME) be used rather than DTT as the reducing agent in the NuPAGE LDS Sample Buffer?
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Figures

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
3184887Certificate of AnalysisJun 19, 2025AM2694
3249592Certificate of AnalysisMay 30, 2025AM2696
3221366Certificate of AnalysisMay 30, 2025AM2694
3246723Certificate of AnalysisMay 29, 2025AM2696
3242199Certificate of AnalysisMay 29, 2025AM2696
5 results displayed, search above for a specific certificate

Safety Data Sheets

Frequently asked questions (FAQs)

SUPERase·In RNase Inhibitor (20 U/µL) (Cat. No. AM2694, AM2696) is active up to 65 degrees C and over a pH range of 5.5 to 8.5.

Find additional tips, troubleshooting help, and resources within our Nucleic Acid Purification and Analysis Support Center.

SUPERase.In RNase Inhibitor is a protein-based inhibitor of nonhuman origin that noncovalently binds and inhibits the most common and troublesome RNases, including RNase A, B, C, 1, and T1.

The storage buffer we recommend is as follows: 2 mM KH2PO4, 8 mM Na2HPO4, 2.7 mM KCl, 137 mM NaCl, pH 7.4 in 50% glycerol.

Share catalog number, name or link

1x1 image pixel for data collection