Fetal Hemoglobin Test Kit, FITC
Product Image
Invitrogen™

Fetal Hemoglobin Test Kit, FITC

The Fetal Hemoglobin (HbF) Test is based on a monoclonal antibody directed to fetal hemoglobin (hemoglobin F). This antibody isRead more
Promo ImagePromotion
Promo code:P5906365
Don't miss out: buy 3, only pay for 2
Promo ImagePromotion
Promo code:RPUZZ25
Stock up on essentials to piece your discovery together
Have Questions?
Catalog NumberQuantity
HFH01Promo Image100 Tests
Catalog number HFH01
Price (USD)
562.65
Online Exclusive
750.00
Save 187.35 (25%)
Each
In stock
Add to cart
Quantity:
100 Tests
Recurring order eligible. Learn more »
Price (USD)
562.65
Online Exclusive
750.00
Save 187.35 (25%)
Each
Add to cart
Ask our AI about this Product

The Fetal Hemoglobin (HbF) Test is based on a monoclonal antibody directed to fetal hemoglobin (hemoglobin F). This antibody is conjugated with fluorescein (FITC). Commonly used, the fluorescein conjugates provide relatively high absorptivity, excellent fluorescence quantum yield, and good water solubility.

This reagent is intended for the identification followed by enumeration of fetal red blood cells. Fetal cells are identified by the presence of fetal hemoglobin using a flow cytometric method. The Fetal Hemoglobin Test may be used as an aid in identifying fetal-maternal hemorrhage and determining the need for immunoprophylaxis with immune globulin.

Antibody Specifications

  • Description: Fetal Hemoglobin F (HbF), mouse IgG1 monoclonal
  • Label: Fluorescein (FITC)
  • Validated Application: Flow Cytometry
  • Species Reactivity: Human
  • Ex/Em and laser configuration: 488/519 nm, blue laser
  • Clone ID: HbF-1

Clinical Relevance

The Fetal Hemoglobin Test contains a monoclonal antibody directed to HbF that specifically recognizes fetal hemoglobin found in fetal erythrocytes. The test permits the enumeration of fetal erythrocytes in the maternal circulation as a result of a fetal-maternal hemorrhage, or leakage, of fetal cells into the maternal circulation. Hemorrhage of cells from an Rh+ fetus into the circulation of an Rh- mother may result in the formation of Rh-reactive antibodies in the mother. Rh hemolytic anemia in a subsequent Rh+ fetus may result from placental transfer of antibodies formed in the mother to the fetus. Although the volume of fetal erythrocytes found in the maternal circulation during pregnancy and immediately post-partum is reported to be small and of uncertain clinical significance in many cases, substantial hemorrhage may result from a number of causes including fetal or maternal trauma and placental defects.

Erythrocytes containing fetal hemoglobin may be found in individuals of any age, but with lower amounts of fetal hemoglobin compared to fetal red cells. These cells have been termed F cells. High levels of F cells may also exist in adults with a heterogeneous group of genetic disorders of uncertain etiology, referred to as Hereditary Persistence of Fetal Hemoglobin. Other clinical conditions causing significant levels of anemia may also result in elevated levels of F cells. Several clinical conditions have been described with increased levels of F cells. These conditions include hereditary anemic diseases such as sickle cell anemia and thalassemia major.

Additional Information

Use the Spectraviewer to help you plan your experiments. Additional general information about Life Technologies flow cytometry products and support is also available.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Antibody ConjugateClassic Dyes (e.g. FITC, TRITC), FITC (Fluorescein)
Detection MethodFluorescence
For Use With (Application)Flow Cytometry Antibodies and Secondary Detection
FormatKit
Label or DyeFITC (Fluorescein)
Quantity100 Tests
Shipping ConditionWet Ice
SpeciesMouse
TypeHemoglobin Test Kit
CE MarkerYes
Unit SizeEach
Contents & Storage
Store in refrigerator (2°C to 8°C).
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
2533936Certificate of AnalysisJan 26, 2025HFH01
2995097Certificate of AnalysisJul 06, 2024HFH01
2979609Certificate of AnalysisMay 25, 2024HFH01
3 results displayed, search above for a specific certificate

Limited Use Label Licenses (LULL)

'

Notice to Purchaser: Resale of this product is expressly prohibited. No right to resell this product or any of its components is conveyed expressly, by implication, or by estoppel. For information on obtaining additional rights, please contact outlicensing@thermofisher.com, Licensing and Commercial Supply, Thermo Fisher Scientific, 5823 Newton Drive, Carlsbad, CA, 92008, United States.

'
'

Notice to Purchaser: This product is licensed from Trillium Diagnostics, LLC, Bloomfield Hills, MI.

'

Citations & References (6)

Citations & References
Abstract
Detection of fetomaternal hemorrhage.
Authors:Kim YA, Makar RS,
Journal:Am J Hematol
PubMed ID:22231030
'The prevention of Rhesus D alloimmunization through Rh immune globulin (RhIg) administration is the major indication for the accurate detection and quantification of fetomaternal hemorrhage (FMH). In the setting of D incompatibility, D-positive fetal cells can sensitize the D-negative mother, resulting in maternal anti-D alloantibody production. These anti-D alloantibodies may ... More
Site-specific gene correction of a point mutation in human iPS cells derived from an adult patient with sickle cell disease.
Authors:Zou J, Mali P, Huang X, Dowey SN, Cheng L,
Journal:Blood
PubMed ID:21881051
'Human induced pluripotent stem cells (iPSCs) bearing monogenic mutations have great potential for modeling disease phenotypes, screening candidate drugs, and cell replacement therapy provided the underlying disease-causing mutation can be corrected. Here, we report a homologous recombination-based approach to precisely correct the sickle cell disease (SCD) mutation in patient-derived iPSCs ... More
Estimation of
Authors:Roys JL, Warzynski MJ,
Journal:Cytometry B Clin Cytom
PubMed ID:16059876
Improving laboratory practice: looking at fetal hemoglobin histograms.
Authors:Warzynski MJ, Roys JL,
Journal:Cytometry B Clin Cytom
PubMed ID:14994377
Tracking donor RBC survival in premature infants: agreement of multiple populations of biotin-labeled RBCs with Kidd antigen-mismatched RBCs.
Authors:Widness JA, Nalbant D, Matthews NI, Strauss RG, Schmidt RL, Cress GA, Zimmerman MB, Mock DM,
Journal:
PubMed ID:24108188
Anemia, a common condition among critically ill premature infants, is affected by red blood cell (RBC) survival (RCS). We hypothesized that transfused allogeneic Kidd antigen-mismatched RBCs would demonstrate the same concurrent RCS tracking as RBCs multilabeled at separate, discrete low densities with biotin (BioRBCs). Allogeneic RBCs from adult donors were ... More
6 total citations

Other products to consider



Share catalog number, name or link

1x1 image pixel for data collection