Triton X-100 (1%)
Product Image
Invitrogen™

Triton X-100 (1%)

This reagent, a component of the Fetal Hemoglobin (HbF) test, comes as a 1% solution that can be diluted inRead more
Have Questions?
Catalog NumberQuantity
HFH1010 mL
Catalog number HFH10
Price (USD)
118.00
Each
-
Add to cart
Quantity:
10 mL
Recurring order eligible. Learn more »
Price (USD)
118.00
Each
Add to cart
Ask our AI about this Product
This reagent, a component of the Fetal Hemoglobin (HbF) test, comes as a 1% solution that can be diluted in phosphate-buffered saline (PBS) containing bovine serum albumin (BSA) and sodium azide to create a 0.1% working solution.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity10 mL
Reagent TypeTriton X-100
Shipping ConditionWet Ice
FormLiquid
Product TypeDetergent
Unit SizeEach
Contents & Storage
Store in refrigerator (2–8°C).

Shelf life: 24 months from date of manufacture
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
3160959Certificate of AnalysisMar 16, 2025HFH10
2362224Certificate of AnalysisJan 29, 2025HFH10
2209541ACertificate of AnalysisJan 29, 2025HFH10
2527642BCertificate of AnalysisJan 29, 2025HFH10
2527642Certificate of AnalysisJan 29, 2025HFH10
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection