LB Agar plate with 50μg/mL Carbenicillin
This product is animal origin and can only be shipped within the United States
LB Agar plate with 50μg/mL Carbenicillin
Thermo Scientific Chemicals

LB Agar plate with 50μg/mL Carbenicillin

 
Have Questions?
Catalog NumberQuantity
J62338.EQF
also known as J62338-=F
20 Plates
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J62338.EQF
also known as J62338-=F
Price (USD)
-
Quantity:
20 Plates
Request bulk or custom format
Specifications
Diameter (Metric) Outside10 cm
For Use With (Application)For growth of Escherichia coli, plasmid and other bacterial cultures
Height (Metric)15 mm
Media TypeLB Agar
Packaging TypePlate
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Used for growth of Escherichia coli, plasmid and other bacterial cultures
RUO – Research Use Only
Contents & Storage
2°C to 8°C
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
S06L988Certificate of AnalysisJun 22, 2025J62338.EQF
Q01L991Certificate of AnalysisMar 28, 2025J62338.EQF
Z02K973Certificate of AnalysisJan 31, 2025J62338.EQF
M03L979Certificate of AnalysisJan 27, 2025J62338.EQF
Z02K972Certificate of AnalysisDec 02, 2024J62338.EQF
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection