Lectin GS-II From Griffonia simplicifolia, Alexa Fluor™ 594 Conjugate
Lectin GS-II From <i>Griffonia simplicifolia</i>, Alexa Fluor&trade; 594 Conjugate
Invitrogen™

Lectin GS-II From Griffonia simplicifolia, Alexa Fluor™ 594 Conjugate

Lectin GS-II, isolated from the seeds of the tropical African legume Griffonia simplicifolia is a 113,000-dalton compoased of identical subunits.Read more
Promo ImagePromotion
Promo code:RIRP25
Save on Imaging Essentials to Create Your Next Masterpiece
14% off your purchase of $499 or more on Invitrogen cell stains and mounting products Learn More
Have Questions?
Catalog NumberQuantity
L21416Promo Image500 μg
Catalog number L21416
Price (USD)
213.00
Each
Add to cart
Quantity:
500 μg
Recurring order eligible. Learn more »
Price (USD)
213.00
Each
Add to cart
Ask our AI about this Product
Lectin GS-II, isolated from the seeds of the tropical African legume Griffonia simplicifolia is a 113,000-dalton compoased of identical subunits. Each subunit contains a single binding site specific for terminal non-reducing a- or ß-linked N-acetyl-D-glucoasemine. Unlike many other lectins, GS-II does not react with any specific blood group. The red-fluorescent lectin GS-II Alexa Fluor™ 594 conjugate can be used in fluorescence microscopy..

View complete list of fluorescent dye-conjugated lectins ›
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Label TypeAlexa Fluor Dyes
Product LineAlexa Fluor™
Protein SubtypeLectins
Quantity500 μg
Shipping ConditionRoom Temperature
SourceAfrica Shrub legume
ConjugateAlexa Fluor 594
FormLyophilized
Unit SizeEach
Contents & Storage
Store in freezer (-5 to -30°C) and protect from light.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Fluorescence spectra

Fluorescence spectra

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
2978752Certificate of AnalysisNov 05, 2024L21416
2411569Certificate of AnalysisDec 10, 2021L21416
2160052Certificate of AnalysisJan 13, 2020L21416
2062940Certificate of AnalysisJan 22, 2019L21416
2014022Certificate of AnalysisAug 14, 2018L21416
5 results displayed, search above for a specific certificate

Safety Data Sheets

Citations & References (10)

Citations & References
Abstract
Increased apical insertion of the multidrug resistance protein 2 (MRP2/ABCC2) in renal proximal tubules following gentamicin exposure.
Authors:Notenboom S, Wouterse AC, Peters B, Kuik LH, Heemskerk S, Russel FG, Masereeuw R,
Journal:J Pharmacol Exp Ther
PubMed ID:16757538
'Multidrug resistance protein (MRP) 2 (MRP2; ABCC2), an organic anion transporter apically expressed in liver, kidney, and intestine, plays an important protective role through facilitating the efflux of potentially toxic compounds. We hypothesized that upon a toxic insult, MRP2 is up-regulated in mammalian kidney, thereby protecting the tissue from damage. ... More
The maturation of mucus-secreting gastric epithelial progenitors into digestive-enzyme secreting zymogenic cells requires Mist1.
Authors:Ramsey VG, Doherty JM, Chen CC, Stappenbeck TS, Konieczny SF, Mills JC,
Journal:Development
PubMed ID:17164426
Continuous regeneration of digestive enzyme (zymogen)-secreting chief cells is a normal aspect of stomach function that is disrupted in precancerous lesions (e.g. metaplasias, chronic atrophy). The cellular and genetic pathways that underlie zymogenic cell (ZC) differentiation are poorly understood. Here, we describe a gene expression analysis of laser capture microdissection ... More
Plasma membrane topology of syncytial domains of herpes simplex virus type 1 glycoprotein K (gK): the UL20 protein enables cell surface localization of gK but not gK-mediated cell-to-cell fusion.
Authors:Foster TP, Alvarez X, Kousoulas KG
Journal:J Virol
PubMed ID:12477855
Most spontaneously occurring mutations that cause extensive herpes simplex virus type 1 (HSV-1)-induced cell fusion are single amino acid changes within glycoprotein K (gK). Despite the strong genetic association of gK with virus-induced cell fusion, its direct involvement in cellular membrane fusion has been controversial, largely due to previously unsuccessful ... More
Signalling codes for the maintenance and lineage commitment of embryonic gastric epithelial progenitors.
Authors:
Journal:Development
PubMed ID:32878924
Apolipoprotein M expression increases the size of nascent pre beta HDL formed by ATP binding cassette transporter A1.
Authors:
Journal:J Lipid Res
PubMed ID:19767535
10 total citations

Other products to consider



Share catalog number, name or link

1x1 image pixel for data collection