μDrop™ and μDrop Duo Plates
μDrop™ and μDrop Duo Plates
μDrop™ and μDrop Duo Plates
μDrop™ and μDrop Duo Plates
μDrop™ and μDrop Duo Plates
μDrop™ and μDrop Duo Plates
Thermo Scientific™
μDrop™ and μDrop Duo Plates
Thermo Scientific™ μDrop and μDrop Duo plates are used for low volume (2-10 μl) photometric DNA, RNA and protein quantitation and purity analysis. The μDrop plates are compatible with Thermo Scientific Varioskan LUX and Multiskan SkyHigh microplate readers. The ready-made protocols in SkanIt software or Multiskan SkyHigh user interface make quantification fast and easy.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct Type
N12391μDrop Plate
N12391M2μDrop Duo Plate
Catalog number N12391
Price (EUR)
-
Request A Quote
Product Type:
μDrop Plate
Ask our AI about this Product
Thermo Scientific™ μDrop™ and μDrop™ Duo Plates are ideal tools for quick and easy photometric low-volume (2–10 μl) concentration measurements of DNA, RNA, protein samples, or spectral scanning. The low volume area is designed to be used with 8-channel pipette for fast and accurate multiple sample pipetting. The plates also contain a separate area for cuvette measurement. μDrop Plates are compatible with Thermo Scientific Varioskan™ LUX, Multiskan™ Sky and Multiskan SkyHigh microplate readers. The intuitive Thermo Scientific SkanIt™ software and Multiskan SkyHigh user interface offer ready-made measurement protocols for quick sample measurement and analysis.
  • dsDNA detection range from a few micrograms to a few milligrams per milliliter
  • Up to 16 (μDrop Plate) or 32 (μDrop Duo Plate) sample positions for quick and easy measurement of sample volumes down to 2 μl
  • Compatible with an eight-channel pipette for sample addition
  • Easy to clean after measurement
  • Measurement slot for a rectangular cuvette
  • Ready-made Multiskan SkyHigh user interface and SkanIt Software protocols for nucleic acid and protein analysis
  • Compatible with Multiskan Sky, Multiskan SkyHigh and Varioskan LUX microplate readers
Specifications
Capacity (Metric)2 to 10μL (low-volume function)
No. of Positions16
Plate TypeμDrop Duo Plate
Product TypeμDrop Plate
Unit SizeEach
Our new Varioskan ALF Multimode Microplate Reader for absorbance, fluorescence and luminescence measurements is fast, flexible & easy-to-use. We offer intuitive SkanIt software editions for Research or 21 CFR Part 11 compliance.
Learn more now
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Customers who viewed this item also viewed
Documents & Downloads
Certificates
0 results displayed, search above for a specific certificate
Frequently asked questions (FAQs)
µDrop and µDrop Duo Plates (Cat. No. N12391, N12391M2) are designed for low-volume (2 to 10 µL) photometric concentration measurements of DNA, RNA, protein, or spectral scanning and can be used without a cuvette. The plates feature a low volume area that allows for direct sample application, eliminating the need for a traditional cuvette. This design facilitates quick and efficient sample preparation and measurement, making the process more streamlined.
The blanking process for µDrop and µDrop Duo Plates (Cat. No. N12391, N12391M2) involves using a designated well under the cuvette for blanking purposes. Specifically, the blank sample is added to the G11 well of the first plate in the layout. This blank sample is used to calibrate the measurements and help ensure accuracy by accounting for any background noise or interference. Each measurement should contain a blank subtraction and we recommend including at least one blank sample in each measurement to maintain the integrity of the results. There are different ways to perform the blank subtraction:
- Blanks on the same µDrop or µDrop Duo Plate
- Sample-specific blanks
- Previously made blanks
Yes, the µDrop Plate can be used with the Multiskan Sky Microplate Spectrophotometer. Prewritten protocols are available in the SkanIt Software Cloud Library.
Yes, the µDrop Plate can be used. Prewritten protocols are available in the SkanIt Software Cloud library.
Share catalog number, name or link
1x1 image pixel for data collection