Multiskan™ FC Microplate Photometer
Build your own complete bundle and get personalized pricing today! Start Building
Save 51% on plasticware essentials and stock your lab to culture with confidence. View promotion details >
Multiskan™ FC Microplate Photometer
Thermo Scientific™

Multiskan™ FC Microplate Photometer

Multiskan FC microplate photometer is a reliable instrument for a wide variety of research and routine applications. It reads 96- and 384-well plates and is equipped with shaking and incubation capabilities for temperature critical assays. It can be used as a stand-alone instrument or under PC control with intuitive Thermo Scientific SkanIt™ software.
Promo ImagePromotion
Promo code:P5780456
Gene Expression Promo
From RNA Isolation kits, reverse transcriptases to analysis of your gene with qPCR - upto 35% on all Gene Expression workflow productsLearn More
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionPlate Type
51119000Promo ImageMultiskan FC96-well Plate
1410101Promo ImageAccessory - Lamp 6V/10W
51119100Promo ImageMultiskan FC with incubator96 and 384-well Plates
Catalog number 51119000
Price (USD)
8,070.00
Each
In stock
Add to cart
Description:
Multiskan FC
Plate Type:
96-well Plate
Price (USD)
8,070.00
Each
Add to cart
Ask our AI about this Product
Multiskan FC brings together over 40 years experience of the well-known Multiskan product brand, with features for enhanced usability. The Multiskan FC has a 340–850 nm wavelength range, enabling a wide variety of applications from enzyme kinetic studies to Lowry assays. The instrument provides fast and accurate measurements enabling complete 96-well plate reading in less than seven seconds. The instrument is also equipped with linear shaking as standard. A model with an incubator enables incubation temperatures up to 50°C and the capacity to read 384-well plates. SkanIt software supports optimal use of the instrument with a visual workflow, effortless data analysis and exporting capabilities.
  • Proven performance and reliable day-to-day results through patented optical design and in-built self-diagnostics
  • A broad wavelength range of 340–850 nm for a wide variety of research and routine applications such as ELISA immunoassays, protein quantification, endotoxin, cytotoxicity and proliferation assays, enzyme assays and growth curves
  • Equipped with an eight-position filter wheel with three standard filters, 405 nm, 450 nm and 620 nm, preinstalled
  • A comprehensive range of easy-to install additional filters available for order
  • Fast and accurate measurement of both 96- and 384-well plates for various throughput requirements
  • Shaking and incubation up to 50°C for temperature critical assays
  • Ease of use through the large color screen and a variety of language versions
  • Visual and logical SkanIt software for comprehensive instrument control and data handling
  • Multiskan verification plate available for instrument performance verification
  • Robot compatibility for high-throughput environments
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Accuracy± 1% (0 to 3 Abs) or ± 0.003 Abs, which ever is greater at 405 nm
CompatibilityRobot
Depth (English)15.7 in.
Depth (Metric)40 cm
DescriptionMultiskan FC
Filters8-position filter wheel; filters installed: 405nm, 450nm, and 620nm
Height (English)8.7 in.
Height (Metric)22 cm
Hertz50/60 Hz
Linearity0 to 3 Abs, ±2% at 405 nm, 96-well plate
Measurement Speed7 s for 96-well Plate (Fast Mode), 13 s for 96-well Plate (Normal Mode)
PrecisionCV ≤ 0.2% (0.3 to 3 Abs) at 405 nm
Resolution0.001 Abs
Voltage100/240 V
Warranty1 Year
Wavelength Range340 to 850 nm
Width (English)11.4 in.
Width (Metric)29 cm
Detector TypePhotodetector
InterfaceOn-board or PC Control
Light SourceQuartz Halogen Lamp
Measurement TechnologyAbsorbance
Plate Type96-well Plate
Product LineMultiskan
ShakingLinear
Software TypeSkanIt Software
Wavelength SelectionFilters
Unit SizeEach

Our new Varioskan ALF Multimode Microplate Reader for absorbance, fluorescence and luminescence measurements is fast, flexible & easy-to-use. We offer intuitive SkanIt software editions for Research or 21 CFR Part 11 compliance.

Learn more now

software-resource

Thermo Scientific SkanIt Software comes free with your microplate reader purchase and can be installed on as many computers as you like. This easy-to-use program is so intuitive, it requires little-to-no training. SkanIt Software has features that help boost productivity and prevent common errors in the lab.
Learn more ›

Have questions about this product? Ask our AI assisted search.
I think my Multiskan FC isn't working right, but there's no error message. How can I check?
Can I read my DNA / RNA / protein at 260 / 280 in Multiskan FC?
What are the electrical specifications for Multiskan FC?
Do I have to use a PC with software to read plates with Multiskan FC?
Does the Multiskan FC include shaking?
+Show more FAQs for this product
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

0 results displayed, search above for a specific certificate

Scientific Resources

Product Information

Frequently asked questions (FAQs)

The Multiskan FC Microplate Photometer (same instrument as the Fisherbrand accuSkan FC Filter-Based Microplate Photometer) is factory-calibrated and performs self-diagnostics on start-up to ensure consistent results over time. If you need to verify that the instrument is working properly, one of the options below can be utilized:

- Performance can be user-verified using Multiskan Verification Plate (Cat. No. 24072800, for 8 wavelengths or Cat. No. 24072805, for 16 wavelengths).

- The other option for verification is by purchasing our qualification service as an IQ/OQ/PQ or IPV that is required for laboratories that operate within a quality management system, comply with global standards (e.g., GLP, GCP, GMP, ISO 17025, and ISO 15189), and/or follow country-specific regulations (e.g., US FDA 21 CFR Part 11, CLIA, and CAP). Our qualification service provides documented verification that laboratory instruments are installed and operating according to the manufacturer’s performance specifications. More information about this service can be found here.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

The Multiskan FC Microplate Photometer offers filters with the following wavelengths for separate purchase. The filter SKU is 142xxx5, where xxx = filter wavelength.
340 nm
375 nm
414 nm
492 nm
520 nm
530 nm
540 nm
550 nm
560 nm
570 nm
595 nm
630 nm
650 nm
690 nm
740 nm
750 nm
820 nm

Yes, the Multiskan FC is intergrateable with robotics.

Yes, the Halogen lamp 6V/10W is Cat. No. 1410101. You can replace the lamp yourself using instructions in the Multiskan FC User Manual: https://www.thermofisher.com/order/catalog/product/51119100

If the results are not as expected, performance can be verified using Multiskan Verification Plate (Cat. Nos. 24072800 (for 8 wavelengths, use with either model) or 24072805 (for 16 wavelengths, use with 51119100)).

Other products to consider



Share catalog number, name or link