NuPAGE™ LDS Sample Buffer (4X)
NuPAGE™ LDS Sample Buffer (4X)
Invitrogen™

NuPAGE™ LDS Sample Buffer (4X)

NuPAGE LDS Sample Buffer (4X) is used to prepare protein samples for denaturing gel electrophoresis with Bis-Tris or Tris-Acetate gels.Read more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
NP0008250mL
NP000710mL
Catalog number NP0008
Price (USD)
185.00
Each
In stock
Add to cart
Quantity:
250mL
Recurring order eligible. Learn more »
Price (USD)
185.00
Each
Add to cart
Ask our AI about this Product
NuPAGE LDS Sample Buffer (4X) is used to prepare protein samples for denaturing gel electrophoresis with Bis-Tris or Tris-Acetate gels. It contains lithium dodecyl sulfate, pH 8.4, which allows for maximum activity of the reducing agent.

See all available buffers and reagents available for SDS-PAGE

NuPAGE LDS Sample Buffer contains Coomassie G250 and Phenol Red as tracking dyes instead of bromophenol blue. Coomassie G250 gives a sharp dye front with both MES and MOPS SDS running buffers and migrates much closer to the moving ion front than bromophenol blue. Bromophenol blue runs more slowly than some peptides with MES SDS Running Buffer. This ensures that small peptides do not run off the gels.

To use: Heat the sample in a 1X dilution (reduced or non-reduced) at 70°C for 10 minutes for optimal results.

Note: NuPAGE LDS Sample Buffer should be brought to room temperature (25°C) prior to use. It is a highly viscous and concentrated solution containing twice the amount of LDS compared to the amount of SDS in typical sample buffers. NuPAGE LDS Sample Buffer also contains a higher concentration glycerol.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
BufferSample Loading Buffers
Concentration4X
Gel CompatibilityNuPAGE™ Gels
Product LineNuPAGE™
Product TypeLDS Sample Buffer
Quantity250mL
Shipping ConditionApproved for shipment at Room Temperature or on Wet Ice
Gel TypeNuPAGE
Unit SizeEach
Contents & Storage
Sample Buffer (4X) containing lithium dodecyl sulfate (LDS) at pH 8.5 with SERVA Blue G250 and phenol red.

Store at 4°C to 25°C.

iBlot 3 Western Blot Transfer System

Introducing iBlot 3 Western Blot Transfer System

Featuring higher throughput and built-in cooling for consistent protein transfer
Learn more

Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Frequently asked questions (FAQs)

Can I prepare my protein sample with the reducing agent and store it for future use?

DTT is not stable, so it must be added and the reduction performed just prior to loading your samples.

Find additional tips, troubleshooting help, and resources within our Protein Gel 1D Electrophoresis Support Center.

My LDS or SDS sample buffer precipitates when stored at 4 degrees C. Can I warm it up? Can I store it at room temperature?

Precipitation of the LDS or SDS at 4 degrees C is normal. Bring the buffer to room temperature and mix until the LDS/SDS goes into solution. If you do not want to wait for it to dissolve, you can store the sample buffer at room temperature.

Find additional tips, troubleshooting help, and resources within our Protein Gel 1D Electrophoresis Support Center.

How are Bolt gels different than NuPAGE gels?

While they are both Bis-Tris based gels, the chemistries are very different since Bolt gels are optimized for western blotting. Another key difference is the wedge well design of the Bolt gels, which allows larger sample volumes to be loaded.

Find additional tips, troubleshooting help, and resources within our Protein Gel 1D Electrophoresis Support Center.

What is the advantage of NuPAGE Gels over regular Tris-Glycine gels?

The neutral operating pH of the NuPAGE Gels and buffers provides following advantages over the Laemmli system:
-Longer shelf life of 8-12 months due to improved gel stability
-Improved protein stability during electrophoresis at neutral pH resulting in sharper band resolution and accurate results (Moos et al, 1998)
-Complete reduction of disulfides under mild heating conditions (70 degrees C for 10 min) and absence of cleavage of asp-pro bonds using the NuPAGE LDS Sample buffer (pH > 7.0 at 70 degrees C)
-Reduced state of the proteins maintained during electrophoresis and blotting of the proteins by the NuPAGE Antioxidant
Please refer to the following paper: Moos M Jr, Nguyen NY, Liu TY (1988) Reproducible High Yield Sequencing of Proteins Electrophoretically Separated and Transferred to an Inert Support. J Biol Chem 263:6005-6008.

Find additional tips, troubleshooting help, and resources within our Protein Gel 1D Electrophoresis Support Center.

What is the composition of the NuPAGE LDS Sample Buffer and what is the recipe for the 4X formulation?

The composition of the 1X NuPAGE LDS Sample Buffer is as follows:

141 mM Tris base
106 mM Tris HCl
2% LDS
10% Glycerol
0.51 mM EDTA
0.22 mM SERVA Blue G
0.175 mM Phenol Red
pH 8.5


To prepare 10 mL of 4 X NuPAGE LDS Sample Buffer, dissolve the following reagents to 8 mL ultrapure water:

Tris HCl 0.666 g
Tris Base 0.682 g
LDS 0.800 g
EDTA 0.006 g
Glycerol 4 g
SERVA Blue G (1 solution) 0.75 mL
Phenol Red (1 solution) 0.25 mL

Mix well and adjust the volume to 10 mL with ultrapure water. Store at +4. The buffer is stable for 6 months when stored at +4°C.

Find additional tips, troubleshooting help, and resources within our Protein Gel 1D Electrophoresis Support Center.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
3197456Certificate of AnalysisJun 04, 2025NP0007
3154062Certificate of AnalysisMay 16, 2025NP0007
3161011Certificate of AnalysisMay 09, 2025NP0008
3146929Certificate of AnalysisMar 21, 2025NP0008
3136399Certificate of AnalysisMar 10, 2025NP0007
5 results displayed, search above for a specific certificate

Safety Data Sheets

Citations & References (9)

Citations & References
Abstract
Characterization of four murine homologs of the human ov-serpin monocyte neutrophil elastase inhibitor MNEI (SERPINB1).
Authors: Benarafa Charaf; Cooley Jessica; Zeng Weilan; Bird Phillip I; Remold-O'Donnell Eileen;
Journal:J Biol Chem
PubMed ID:12189154
'The human ov-serpin monocyte neutrophil elastase inhibitor (MNEI) is encoded by a single gene SERPINB1. It is a highly efficient inhibitor of neutrophil granule proteases. Four murine genes with high sequence identity with MNEI were identified and fully sequenced, and these were named EIA, EIB, EIC, and EID. EIA, EIB ... More
Novel Modulation of Neuronal Nicotinic Acetylcholine Receptors by Association with the Endogenous Prototoxin lynx1.
Authors: Ibañez-Tallon Inés; Miwa Julie M; Wang Hai Long; Adams Niels C; Crabtree Gregg W; Sine Steven M; Heintz Nathaniel;
Journal:Neuron
PubMed ID:11906696
We previously identified lynx1 as a neuronal membrane molecule related to snake alpha-neurotoxins able to modulate nAChRs. Here, we show that lynx1 colocalizes with nAChRs on CNS neurons and physically associates with nAChRs. Single-channel recordings show that lynx1 promotes the largest of three current amplitudes elicited by ACh through alpha(4)beta(2) ... More
Apolipoprotein A-I is a selective target for myeloperoxidase-catalyzed oxidation and functional impairment in subjects with cardiovascular disease.
Authors:Zheng L, Nukuna B, Brennan ML, Sun M, Goormastic M, Settle M, Schmitt D, Fu X, Thomson L, Fox PL, Ischiropoulos H, Smith JD, Kinter M, Hazen SL,
Journal:J Clin Invest
PubMed ID:15314690
In recent studies we demonstrated that systemic levels of protein-bound nitrotyrosine (NO(2)Tyr) and myeloperoxidase (MPO), a protein that catalyzes generation of nitrating oxidants, serve as independent predictors of atherosclerotic risk, burden, and incident cardiac events. We now show both that apolipoprotein A-I (apoA-I), the primary protein constituent of HDL, is ... More
In vivo functional assay of a recombinant aquaporin in Pichia pastoris.
Authors:Daniels MJ, Wood MR, Yeager M,
Journal:Appl Environ Microbiol
PubMed ID:16461705
The water channel protein PvTIP3;1 (alpha-TIP) is a member of the major intrinsic protein (MIP) membrane channel family. We overexpressed this eukaryotic aquaporin in the methylotrophic yeast Pichia pastoris, and immunogold labeling of cellular cryosections showed that the protein accumulated in the plasma membrane, as well as vacuolar and other ... More
Spermidine but not spermine is essential for hypusine biosynthesis and growth in Saccharomyces cerevisiae: Spermine is converted to spermidine in vivo by the FMS1-amine oxidase.
Authors:Chattopadhyay MK, Tabor CW, Tabor H,
Journal:Proc Natl Acad Sci U S A
PubMed ID:14617780
In our earlier work we showed that either spermidine or spermine could support the growth of spe2Delta or spe3Delta polyamine-requiring mutants, but it was unclear whether the cells had a specific requirement for either of these amines. In the current work, we demonstrate that spermidine is specifically required for the ... More
9 total citations

Share catalog number, name or link

1x1 image pixel for data collection