Staphylococcus aureus (Wood strain without protein A) BioParticles™, unlabeled
<i>Staphylococcus aureus</i> (Wood strain without protein A) BioParticles&trade;, unlabeled
Invitrogen™

Staphylococcus aureus (Wood strain without protein A) BioParticles™, unlabeled

The Molecular Probes™ unlabeled BioParticles™ product line comprises heat- or chemically killed S. aureus (Wood strain, without protein A) andRead more
Promo ImagePromotion
Promo code:P5906365
Don't miss out: buy 3, only pay for 2
Promo ImagePromotion
Promo code:RPUZZ25
Stock up on essentials to piece your discovery together
Have Questions?
Catalog NumberQuantity
S2859Promo Image100 mg
Catalog number S2859
Price (USD)
196.00
Each
In stock
Add to cart
Quantity:
100 mg
Recurring order eligible. Learn more »
Price (USD)
196.00
Each
Add to cart
Ask our AI about this Product
The Molecular Probes™ unlabeled BioParticles™ product line comprises heat- or chemically killed S. aureus (Wood strain, without protein A) and zymosan (S. cerevisiae) cells. These nonfluorescent BioParticles™ products are useful in phagocytosis assays, both as controls and for custom-labeling with reactive dyes.

BioParticles Specifications:
• Label (Ex/Em): Unlabeled
• Particle: S. aureus (Wood strain, without protein A)
Opsonizing reagent available


Using BioParticles™ Products
BioParticles™ conjugates are provided as lyophilized powders. There are approximately 3 x 108 S. aureus particles per mg solid and approximately 2 x 107 zymosan particles per mg solid. BioParticles™ conjugates can be reconstituted in the buffer of your choice for use in phagocytosis assays.

Find More BioParticles™ Products
We offer a large range of dye-labeled and unlabeled E. coli (K-12 strain), S. aureus (Wood strain, without protein A), and zymosan (S. cerevisiae) BioParticles™ products. Find out about these products and their applications by reviewing Probes for Following Receptor Binding and Phagocytosis—Section 16.1 in the Molecular Probes™ Handbook.

For pH-sensitive endocytosis assays, see our fluorescent pHrodo™ BioParticles™ conjugates.

For Research Use Only. Not for human or animal therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection MethodFluorescence
Dye TypeUnconjugated
FormLyophilized Powder
Quantity100 mg
Shipping ConditionRoom Temperature
SpeciesS. aureus
For Use With (Equipment)Fluorescence Microscope
Product LineBioParticles
Product TypeBioparticle Unlabeled
Unit SizeEach
Contents & Storage
Store in freezer (-5 to -30°C).
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
3197492Certificate of AnalysisMay 13, 2025S2859
3132644Certificate of AnalysisMar 30, 2025S2859
3100013Certificate of AnalysisDec 23, 2024S2859
3017456Certificate of AnalysisSep 04, 2024S2859
2610337Certificate of AnalysisFeb 22, 2023S2859
5 results displayed, search above for a specific certificate

Safety Data Sheets

Frequently asked questions (FAQs)

While the bacteria have been attenuated with formaldehyde and alcohol desiccation, the BioParticles products are not considered sterile, and we do not recommend incubation of more than 4 hours. This applies to all of our dye-labeled (pHrodo, Alexa Fluor, etc.) and unlabeled BioParticles products.

Staphylococcus aureus (Wood strain without protein A) BioParticles, unlabeled (Cat. No. S2859) contains 3 x 108particles/mg.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

We use amine-reactive dyes to covalently attach fluorescent dyes to all of our BioParticles probes such as the Escherichia coli (K-12 strain) BioParticles probes, Staphylococcus aureus (Wood strain without protein A) BioParticles, and the Zymosan A (S. cerevisiae) BioParticles probes.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Fluorescent Bioparticles have been employed to study phagocytosis by fluorescence microscopy, quantitative spectrofluorometry, and flow cytometry. We offer E.Coli, S. aureus, and zymosan BioParticles conjugates covalently labeled with a variety of different fluorophores.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Citations & References (1)

Citations & References
Abstract
Dectin-2 is a pattern recognition receptor for fungi that couples with the Fc receptor gamma chain to induce innate immune responses.
Authors:Sato K, Yang XL, Yudate T, Chung JS, Wu J, Luby-Phelps K, Kimberly RP, Underhill D, Cruz PD, Ariizumi K
Journal:J Biol Chem
PubMed ID:17050534
'Antigen presenting cells recognize pathogens via pattern recognition receptors (PRR), which upon ligation transduce intracellular signals that can induce innate immune responses. Because some C-type lectin-like receptors (e.g. dectin-1 and DCSIGN) were shown to act as PRR for particular microbes, we considered a similar role for dectin-2. Binding assays using ... More
1 total citations

Other products to consider



Share catalog number, name or link

1x1 image pixel for data collection