Whole Allergen
e82 Rabbit epithelium
e82 Rabbit epithelium Scientific Information
Type:
Whole Allergen
Display Name:
Rabbit epithelium
Route of Exposure:
Inhalation
Family:
Leporidae
Species:
Oryctolagus cuniculus
Latin Name:
Oryctolagus cuniculus
Other Names:
Domestic Rabbit, European Rabbit, and Common European Rabbit
Summary
Oryctolagus cuniculus, referred as European rabbits, are the most successful, invasive, exotic mammals, found globally. These species prefer living in herds, in interconnected underground tunnels. Rabbits are domesticated and popular lab animals. For laboratory animal care workers, laboratory animal allergy (LAA) is an important health issue. Rabbit saliva is the most potent allergen source identified, however the allergens are also found in rabbit fur, urine, hair, and dander. Inhalation is the main route of exposure to rabbit allergen, and since the size of the airborne particle is very minute, it easily penetrates through the lower respiratory tract, causing allergic symptoms. In susceptible individuals, allergic sensitization to rabbit allergens is via rabbit contact, either through its ownership, or contact in the laboratory. Allergic symptoms may include asthma, allergic rhinitis, eczema, and urticaria. Rabbit allergen, Ory c 1, is a major allergen (lipocalin family) found in the extracts of saliva, urine and fur of rabbits, that can be used for investigations of sensitivity to rabbits. Ory c 2 belongs to the family of lipocalin, that is derived from rabbit hair, fur, dust and urine. Another important allergen, Ory c 3 has shown high structural homology with cat allergen (Fel d 1), however, at amino acid level, similarity is quite low. Thus, in spite of high similarity, no cross-reactivity has been reported with Fel d 1, thus making it as a potential marker for rabbit allergy. Ory c 4 has demonstrated a degree of cross-reactivity with those of other animals, such as cat, dog and horse.
Allergen
Nature
Oryctolagus cuniculus are European rabbits that belong to the family Leporidae and genus Oryctolagus. These rabbits are considered to be one of the most successful invasive exotic mammals (1). The characteristic features of these species include the body length measuring 35–50 cm and the bodyweight of around 1,500–3,000 g. The fur of the rabbit is generally brownish in color and the tail is white underside (2). Earlier, these rabbits were raised as a food source and for their fur, however, presently their demand as domestic pets have increased (3). 
Habitat
European rabbits are generally seen near brushy fields, shrubs and short grasses (4). They are nocturnal animals, however, prefer to feed at dawn (2). They feed upon dried grass, seeds, bark, buds, fruits, clover, and alfalfa (4). Further, since these species do not prefer cold and wet ecotypes, they generally are unable to survive above 1500 m (1). They are inhabitants of open grasslands that have loosely-compacted and well-drained soils, helping them build their burrows (2). Feral rabbits are generally seen across various habitats around Australia, that range from deserts to wet coastal plains — wherever the soil is appropriate for digging burrows. These rabbits are found to be less in the zones where there is clay soil, whereas they are found in abundance, in the areas where the soil is deep and sandy (5). Most of the rabbits prefer living in herds, in interconnected underground tunnels (6).        
Taxonomy
Taxonomic tree of rabbit (7)
Domain Eukaryotae
Kingdom Metazoa
Phylum Chordata
Subphylum Vertebrata
Class Mammalia
Family Leporidae
Genus Oryctolagus
Species Oryctolagus cuniculus
Taxonomic tree of rabbit (7)
Tissue
Various allergens, such as Ory c 1, Ory c 2, Ory c 3 and Ory c 4 have been identified in rabbit’s saliva, hair, urine, dander and dust (3, 8). Ory c 1 is a major rabbit allergen found in rabbit’s hair, urine, fur and saliva, whereas Ory c 2 is derived from its hair, fur, dust and urine (9). Further, the allergens Ory c 3 and Ory c 4 have been identified in rabbit’s hair, fur and dust, respectively. Of all the rabbit allergen extracts, saliva has been reported to be the most potent one (3).
Epidemiology
Worldwide distribution
Lately, rabbit has been reported to be a significant pet allergen in the development of severe respiratory allergic diseases, due to its exposure in the household. Moreover, rabbit ownership is also on the rise. Hence, it is essential to focus on rabbit sensitization in patients allergic to this species (10).
Rabbit exposure in domestic environment may trigger immunoglobulin E (IgE)-mediated bronchial asthma and/or rhinitis. The development of allergic diseases such as asthma, conjunctivitis, and allergic rhinitis associated with rabbit exposure is frequently seen among researchers, laboratory animal care workers and technicians (8). Further, laboratory animal allergy (LAA) is an important health issue for individuals exposed to experimental animals, since it may easily lead to asthma. In a cross-sectional survey conducted in Japan (n=136), the overall prevalence of LAA was reported be 17.6%. Furthermore, the most commonly reported symptoms were found to be allergic rhino-conjunctivitis (2.4%) and asthma (3%). Among the 69 rabbit handlers, LAA was reported in 16.3% (11).
Individuals who are in direct contact with rabbits in laboratory and domestic settings commonly experience IgE-mediated allergic sensitization. Choi and colleagues presented a case of three individuals who developed rabbit-induced bronchial asthma and/or rhinitis, as a result of keeping rabbit as a household pet. Serum-specific IgE and IgG4 antibodies to rabbit allergens were confirmed. Further, Ory c 1 (17-kDa) glycoprotein was reported to be the major rabbit allergen. With the increase in demand for rabbits as domestic pets, allergic diseases from rabbit exposure at home or workplace have become a common scenario. Physicians should take note that exposure to rabbits may develop extreme respiratory allergic reactions even in individuals who are non-atopic (8).
In Japan, a large cross-sectional study was conducted wherein over 5641 laboratory animal technicians were enrolled. The prevalence of LAA was found to be 29.7% in rabbit handlers. Allergic rhinitis was the most commonly reported LAA among laboratory animal workers (9, 12). In another study conducted in Korea on 401 patients with allergic symptoms, 9% were found to be sensitized to rabbits, whereas 16.7% had direct exposure to rabbits (6, 10).
In a study conducted in Italy, it was demonstrated that the prevalence of sensitization to rabbits ranged between 0.62 and 4.72%. Out of 39 patients who were sensitized to rabbits, 19 individuals experienced direct rabbit contact (10 for ownership and 9 for occasional contact), and 5 individuals reported only indirect exposure through the contact with rabbit owners. Furthermore, the last 15 individuals reported neither direct nor indirect exposure. It was observed that all the monosensitized individuals domesticated rabbits as pets. Also, they were having persistent (moderate-severe) respiratory symptoms. The four (non-smokers) patients with single sensitization to rabbits reported continuous allergic rhinitis, and asthma and had a rabbit as their household pet. Hence, from the study results it has been inferred that the increased prevalence of sensitization to rabbits should not be ignored (13).
Risk factors
In susceptible individuals, a risk factor for the development of allergic sensitization to rabbit allergens in contact with rabbits (rabbit ownership, occasional contact, or contact in the laboratory) (14, 15).
In a domestic environment, direct or even indirect exposure to rabbits in susceptible individuals may result into allergic sensitization. Further, an increase in rabbit ownership may increase the risk of sensitization. Atopy status and the degree of exposure to rabbits are important determinants of sensitization to rabbits in a household environment. Sensitization to cats/dogs has been considered as a risk factor for developing allergies to other furry animals, such as rabbits (16).
It is presumed that the salivary allergen can spread to the rabbit’s fur by their grooming activity. Extensive spreading of allergen containing dust particles is seen in areas where the rabbit is active. Aerosolized allergens related with rabbit activities or with human activities such as handling the rabbits or cleaning-out, may result into allergen levels likely to cause sensitization or trigger symptoms in individuals who are already sensitized (16).
Due to high percentage of rabbit ownership, a progressive increase in rabbit sensitization has been forecasted in the future. In a study conducted in Italy, skin tests were conducted on 2,329 patients, of which 1,602 had a respiratory allergy (asthma and/or rhino-conjunctivitis). Of 1,602 individuals, 2.43% were sensitized to rabbit dander (4 monosensitized) (13).
In laboratories, the laboratory animal workers are at risk of sensitization, and it has been reported that up to one-third will have symptomatic asthma (15). An atopic background and the intensity of exposure are further risk factors (15).
Environmental Characteristics
Worldwide distribution
The European rabbit is a species of rabbit with ancestors belonging to the Iberian range. This species has been introduced to all the continents (except Antarctica), and close to 800 different islands, globally (1). Post Iberian range, it was introduced in Northern Africa, and then Britain, and later expanded its roots throughout the central and northeast part of Europe. They further took their route into Australia, Chile, New Zealand, and Argentina (17).
Route of Exposure
Main
Airway inhalation is the main route of exposure to rabbit allergen (18). The characteristic feature of this allergen is that it can easily penetrate through the lower respiratory tract and cause allergic symptoms since the size of the airborne particle obtained from rabbits is very minute (19).
Secondary
Direct skin and eye contact are considered as a secondary route of allergen exposure (18). It has been reported, that, on rare occurrences, laboratory animal workers allergic to an experimental animal may experience an anaphylactic reaction from either an animal bite or through contaminated needle punctures (9).
Clinical Relevance
Exposure or contact to rodents, in general, have long been identified as a risk factor for the development of allergic symptoms in sensitized individuals (14, 15). In occupational settings, the association between laboratory animal allergy or occupational asthma and rodent exposure is well documented. It is estimated that between 11% and 44% of people working with laboratory animals will develop allergies related to their occupation. Symptoms may include rhinitis, eczema, urticaria, and asthma. Cases are often identified using disease surveillance methods, and exposure reduction may be achieved by the use of respiratory protection or by reassignment to a job where exposure is minimized. Medication may also be required (20).
Severe anaphylaxis
Two case studies were conducted that reported severe allergic reactions such as anaphylaxis among individuals who were exposed to rabbits. In one case study, an atopic girl experienced symptoms such as coughing, itching, rhinorrhea, breathlessness, reverse sneezing, and sensation of swelling in the throat, after inhalant exposure to a rabbit. Her serum-specific IgE to rabbit epithelium and fur was raised and her skin prick test was found to be positive to rabbit epithelium (21). In the other case study, a 32-year-old male physician incidentally got a minor injury from a needle that had been used on the rabbit tissue. A serious anaphylactic reaction developed within 15 minutes of its use, and the physician was hospitalized. Serum IgE antibodies were elevated (16.2 U/ml), however, other antibody titers were reduced (22).
Allergic rhinitis
In a case of a 6-year-old girl with a 3-year history of allergic rhinitis and asthma, worsening of both these symptoms was reported in-home setting and upon exposure to outdoor pollen. Upon interrogation, it was revealed that the family owned a pet rabbit. The laboratory findings revealed positive results for rabbit epithelium (8.43 kIU/L). The family was advised to avoid going out during pollination and keep the rabbit away from home, as it could be a potential trigger aggravating the child’s symptoms (23).
Asthma
It is often seen that rabbits are associated with allergic asthma, of insidious onset. However, it can sometimes result in severe, continuous asthma of rapid onset (24).
A cross-sectional study was conducted in school children in Kuwait that assessed the relationship between keeping rabbits as pets and the development of allergic symptoms. According to the results, it was reported that exposure to rabbits was statistically significantly related to asthma symptoms, such as current dry cough at night (adjusted prevalence ratios [aPRs] 1.18), ever-doctor diagnosed asthma (aPR 1.30), and asthma in the last 12 months (aPR 1.38) (25).
Atopic Dermatitis
In a cross-sectional study, significant association between rabbit keeping as a pet and atopic dermatitis was found. According to the results, it was reported that exposure to rabbits was statistically significantly associated with severe eczema symptoms (aPR 1.94) and ever-diagnosed eczema i.e. study-defined current eczema (aPR 1.45) (25).
Prevention and Therapy
Prevention strategies
Avoidance
In symptomatic animal care handlers, the use of personal protective equipment can help in reducing the exposure of allergen. Equipment such as laminar flow cage, and protective measures such as regular wet washing of containers and routine maintenance of ventilation systems can decrease the exposure of these allergens to a good extent (9).
All laboratory animal handlers should undergo regular medical screening with allergic skin testing. Further, they should also be given training on how to reduce personal exposure (9).
Molecular Aspects
Allergenic molecules
Rabbit saliva is the major source of allergy, besides dust, hair, fur and urine (3). A total of four allergenic molecules have been identified for rabbit epithelium and published in the literature. Among the four allergens, Ory c 1, 3 and 4 have been reported in WHO/IUIS, whereas Ory c 2  has been reported in published literatures, but is not fully characterized yet (26). Ory c 1 is a 17-18 kDa glycoprotein that belongs to the family lipocalin. It is found in rabbit’s fur, urine, hair and saliva (6, 8). Ory c 2 is a 21-kDa that belongs to lipocalin family (3). Ory c 3 is a 19­21 kDa lipophilin having high structural homology to Fel d 1 (cat allergen) (6, 27). Despite a high structural similarity, sequence identity between Ory c 3 and Fel d 1 is very low. Apart from hair and fur, Ory c 3 has been traced in dust procured from homes, where rabbits have been used as pets (3, 27). Ory c 4 is a 24-kDa lipocalin (6). It has been derived from rabbit’s fur. It has a high sequence similarity to dog (Can f 6), cat (Fel d 4) and horse (Equ c 1), and thus the cross-reactivity (3, 27). 
Rabbit allergens recognized by WHO/IUIS Allergen Nomenclature Sub-Committee (26)
Allergen Molecular Weight (kDa) Cross- reactivity/species specificity Protein family Allergenicity
Ory c 1 17-18 nd Lipocalin nd
Ory c 3 19-21 Species-specific Lipophilin Of all patients allergic to rabbits, prevalence of IgE to Ory c 3 was found to be 77% (27)
Ory c 4 24 Probable cross-reactivity with Can f 6, Fel d 4 and Equ c 1 Lipocalin In a group of rabbit-allergic patients, prevalence of specific IgE to Ory c 4 was reported to be 46% (27)
Allergen Molecular Weight (kDa) Cross- reactivity/species specificity Protein family Allergenicity
IUIS: International Union of Immunological Societies; kDa: Kilodaltons; MW: Molecular weight; nd: Not determined; WHO: World Health Organization
Biomarkers of severity
Ory c 1 and Ory c 2, initially referred to as antigen R1 (AgR1) and antigen 2 (Ag2) are important components found in the extracts of dust, urine, saliva and fur of rabbits. Ory c 1 is the major allergen, whose presence in specimens (urine, saliva and fur), used for diagnosis is important for adequate testing. The allergen Ory c 2 has been found in high quantities in the other extracts (dust) and has been found beneficial, while conducting quantitative research on rabbit inhalant allergens. Hence, it has been identified that extracts of rabbit dust, urine, saliva or fur could be considered as important allergens that can be used for investigations of sensitivity to rabbits (28, 29).
Another important rabbit allergen identified i.e. Ory c 3, has demonstrated high specific-IgE prevalence (77%). It has shown high structural homology with cat allergen, Fel d 1, however, at amino acid level, the similarity identified was quite low. Thus, in spite of high similarity, cross-reactivity was not seen between Ory c 3 (rabbit allergen) and Fel d 1 (cat allergen), thus making it as a potential marker for rabbit allergy (3, 27).
Cross-reactivity
The importance of cross-reactivity between furry animal sensitization is strengthened by the fact that greater numbers of patients having cat and dog sensitivity report rabbit sensitivity, with no obvious history of rabbit exposure (6).
Ory c 3 is a lipophilin having high structural homology to Fel d 1 (cat allergen). In a study including a group of patients sensitized to rabbit, the prevalence of IgE to Ory c 3 was found to be 77%. In spite of high structural homology, the sequence identity has been reported to be very low, and no cross-reactivity has been found between Ory c 3 (rabbit) and Fel d 1 (cat) allergen. In the case of Ory c 4, high sequence identity has been observed between Ory c 4 (rabbit) and dog (Can f 6), cat (Fel d 4) and horse (Equ c 1) allergens, suggesting high probability of IgE cross-reactivity (3, 27).
Further, a case of a 17-year-old female patient was reported who had sensitization to rabbit products (urine, serum, and epithelium) during childhood. The symptoms worsened during adolescence and the female patient developed anaphylaxis with severe bronchospasm, secondary to ingestion of rabbit meat. It was demonstrated that the 60-kDa albumin was accountable for the cross-reactivity between rabbit epithelium and meat (30).
Some allergens belonging to lipocalin family have reported high amino acid sequence identity among them (up to 60%), thus explaining the cross‐reactivity between these allergens. These include - Ory c 4 (rabbit), Fel d 4 (cat), Can f 6 (dog), Equ c 1 (horse), Mus m 1 (mouse), and Rat n 1 (rat) (31).
Although not dominant, however cross-reactivity has been observed between rabbit allergen, 15 kDa and IgE antibodies in deer-allergic individuals (29).
Compiled By
Author: Turacoz Healthcare Solutions
Reviewer: Dr. Christian Fischer
Last reviewed: January 2021
References
  1. Lees ACB, Diana J. A conservation paradox for the 21st century: the European wild rabbit Oryctolagus cuniculus, an invasive alien and an endangered native species. Mammal Review. 2008;38(4):304-20.
  2. Michael Hutchins DGK, Valerius Geist, and Melissa C. McDade. Grzimek’s Animal Life Encyclopedia, Mammals I-V.  .12-16. 2nd ed2003. p. 481.
  3. M. Curin CH. Allergy to pets and new allergies to uncommon pets. Allergol Select 2017;1(2):214-221. 2017;1:214-21.
  4. Blagojevich RR. Illinois Mammals. In: Resources IDoN, editor. Illinois2008. p. 8.
  5. Environment.gov.au. FERAL EUROPEAN RABBIT (ORYCTOLAGUS CUNICULUS) 2011 [cited 2020 29 October]. Available from: https://www.environment.gov.au/system/files/resources/7ba1c152-7eba-4dc0-a635-2a2c17bcd794/files/rabbit.pdf.
  6. Weber RW. Allergen of the month--rabbit. Ann Allergy Asthma Immunol. 2014;112(6):A13.
  7. CABI. Rabbit epithelium 2020 [29 October 2020]. Available from: https://www.ncbi.nlm.nih.gov/Taxonomy/Browser/wwwtax.cgi?mode=Info&id=9986&lvl=3&lin=f&keep=1&srchmode=1&unlock.
  8. Choi JH, Kim HM, Park HS. Allergic asthma and rhinitis caused by household rabbit exposure: identification of serum-specific IgE and its allergens. J Korean Med Sci. 2007;22(5):820-4.
  9. Bush RK, Wood RA, Eggleston PA. Laboratory animal allergy. J Allergy Clin Immunol. 1998;102(1):99-112.
  10. Park YB, Mo EK, Lee JY, Kim JH, Kim CH, Hyun IG, et al. Association between pet ownership and the sensitization to pet allergens in adults with various allergic diseases. Allergy Asthma Immunol Res. 2013;5(5):295-300.
  11. Muzembo BA EM, Inaoka Y. Prevalence of occupational allergy in medical researchers exposed to laboratory animals. Ind Health 2014;52(3):256-61. 2014.
  12. Aoyama K, Ueda A, Manda F, Matsushita T, Ueda T, Yamauchi C. Allergy to laboratory animals: an epidemiological study. Br J Ind Med. 1992;49(1):41-7.
  13. Liccardi G, Passalacqua G. Sensitization to rabbit allergens in Italy--a multicentre study in atopic subjects without occupational exposure. Int Arch Allergy Immunol. 2006;141(3):295-9.
  14. Liccardi G, Piccolo A, Dente B, Salzillo A, Noschese P, Gilder JA, et al. Rabbit allergens: a significant risk for allergic sensitization in subjects without occupational exposure. Respir Med. 2007;101(2):333-9.
  15. Phipatanakul W, Matsui E, Portnoy J, Williams PB, Barnes C, Kennedy K, et al. Environmental assessment and exposure reduction of rodents: a practice parameter. Ann Allergy Asthma Immunol. 2012;109(6):375-87.
  16. Willerton L, Mason HJ. The development of methods to measure exposure to a major rabbit allergen (Ory c 1). AIMS Public Health. 2018;5(2):99-110.
  17. Ferrand N, Branco M. The evolutionary history of the European rabbit (Oryctolagus cuniculus): major patterns of population differentiation and geographic expansion inferred from protein polymorphism.  Phylogeography of southern European refugia: Springer; 2007. p. 207-35.
  18. Muzembo BA, Eitoku M, Inaoka Y, Oogiku M, Kawakubo M, Tai R, et al. Prevalence of occupational allergy in medical researchers exposed to laboratory animals. Ind Health. 2014;52(3):256-61.
  19. Sartorelli P, Romeo R, Coppola G, Nuti R, Paolucci V. Angora wool asthma in textile industry. Case Reports Immunol. 2012;2012:358271.
  20. Matsui EC. Role of mouse allergens in allergic disease. Curr Allergy Asthma Rep. 2009;9(5):370-5.
  21. Prince E, Zacharisen MC, Kurup VP. Anaphylaxis to rabbit: a case report. Ann Allergy Asthma Immunol. 1998;81(3):272-3.
  22. Watt AD, McSharry CP. Laboratory animal allergy: anaphylaxis from a needle injury. Occup Environ Med. 1996;53(8):573-4.
  23. Kapur K, et al. Rabbit Anaphylaxis—An Interesting Case Report. Pediatric Asthma, Allergy & Immunology. 2008;21(1):31-4.
  24. Colloff MJ, Ayres J, Carswell F, Howarth PH, Merrett TG, Mitchell EB, et al. The control of allergens of dust mites and domestic pets: a position paper. Clin Exp Allergy. 1992;22 Suppl 2:1-28.
  25. AlShatti KA, Ziyab AH. Pet-Keeping in Relation to Asthma, Rhinitis, and Eczema Symptoms Among Adolescents in Kuwait: A Cross-Sectional Study. Front Pediatr. 2020;8:331.
  26. WHO/IUIS. Oryctolagus cuniculus 2019 [22.12.2020]. Available from: http://www.allergen.org/search.php?allergensource=Oryctolagus+cuniculus.
  27. C. Hilger JK-T, and M. van Hage. Molecular Diagnostics in Allergy to Mammals.  Molecular Allergy Diagnostics Innovation for a Better Patient Managemen. 2017;1-531.2017. p. 363-79.
  28. Warner JA, Longbottom JL. Allergy to rabbits. III. Further identification and characterisation of rabbit allergens. Allergy. 1991;46(7):481-91.
  29. Baker J, Berry A, Boscato L, Gordon S, Walsh B, Stuart M. Identification of some rabbit allergens as lipocalins. Clinical & Experimental Allergy. 2001;31(2):303-12.
  30. Gomez Galan C, Ferre Ybarz L, Sansosti Viltes A, de la Borbolla Moran JM, Pena Peloche MA, Duocastella Selvas P, et al. Rabbit meat anaphylaxis. J Investig Allergol Clin Immunol. 2014;24(2):136-8.
  31. Davila I, Dominguez-Ortega J, Navarro-Pulido A, Alonso A, Antolin-Amerigo D, Gonzalez-Mancebo E, et al. Consensus document on dog and cat allergy. Allergy. 2018;73(6):1206-22.