Whole Allergen
f204 Trout, Rainbow trout
f204 Trout, Rainbow trout Scientific Information
Type:
Whole Allergen
Display Name:
Trout, Rainbow trout
Family:
Salmonidae
Latin Name:
Oncorhynchus mykiss
Other Names:
Trout, Rainbow trout
Clinical Relevance
IgE-mediated reactions
Atopic dermatitis has been seen in patients with IgE antibodies to trout.
Other topics
All Pacific salmon belong to the genus Oncorhynchus, the same as the Rainbow trout. King, Coho, Pink and Chub are major species caught outside the North American west coast. Several species
A study found some cod-sensitive children to have IgE antibodies to trout.(1) IgE antibodies to trout were also measured in a group of patients suffering from atopic dermatitis using Pharmacia CAP System™ (2), and in food-allergic children (3)
Molecular Aspects
Cross-reactivity
Species within groups of fish, like Gadiformes (examples: codfish and hake) and Scombroid fishes (examples: mackerel and tuna) seem to share allergenic components. The overlap of allergen specificity between the groups seems to be moderate or even small. 
Compiled By
Last reviewed: June 2022
References
  1. de Martino, M; Novembre, E; Galli, L; de Marco, A; Botarelli, P; Marano, E; Vierucci, A. Allergy to different fish species in cod-allergic children: In vivo and in vitro studies. J Allergy Clin Immunol; 1990; 86: 909-914.
  2. Lindqvist, A; Ikezawa, Z.; Tanaka, A; Yman, L. Seafood specific IgE in atopic dermatitis. Ann Allergy; 1993;70:58.
  3. Matsumaru, S; Artia, M et al. Clinical evaluation of Pharmacia CAP System new allergens for fish, vegetables, fruits and grains. Jap Soc Ped Allergol; 1992.