Whole Allergen
f300 Goat's milk
f300 Goat's milk Scientific Information
Type:
Whole Allergen
Display Name:
Goat's milk
Other Names:
Goat's milk
Route of Exposure
Main 
Goat's milk, cheese and other dairy products.
Clinical Relevance
IgE-mediated reactions
IgE-mediated reactions have been reported.
Other topic
Goat’s milk often appears as a substitute for cow’s milk, particularly in diets administered by parents to children with atopic dermatitis (1). Some cow’s milk-allergic children may tolerate goat’s milk (2). However, there appears to be evidence for crossreactivity between goat’s milk and cow’s milk (3). In a study of crossreactivity of the casein fractions among different species, the IgE response to ewe and goat’s milk was weaker than that to cow and buffalo (4).
14/16 patients with cow’s milk allergy had specific IgE antibodies to goat’s milk (5). Serum albumin and caseins show homology to the corresponding proteins of cow and sheep (6).
Molecular Aspects
Cross-reactivity
Milk from related animals
Compiled By
Last Reviewed : June 2022
References
  1. Webber, SA; Graham-Brown, RAC; Hutchinson, PE; Burns, DA. Dietary manipulation in childhood atopic dermatitis. Br J Dermatol; 1989; 121: 91-98.
  2. Ellis, MH; Short, J; Heiner, DC. Anaphylaxis after ingestion of a recently introduced hydrolyzed whey protein formula. J Pediatr; 1991; 118: 74-77.
  3. Gjesing, B; Østerballe, O; Schwartz, B; Wahn, U; Løwenstein, H. Allergen-specific IgE antibodies against antigenic components in cow milk and milk substitutes. Allergy; 1986; 41: 51-56.
  4. Bernard, H; Wal, JM; Creminon, C; Grassi, J; Yvon, M; Houdebine, LM. Sensitivities of cow's milk allergic patients to casein fraction of milks from different species. Allergy; 1992; 47: 306.
  5. Dean, TP; Adler, BR; Ruge, F; Warner, JO. In vitro allergenicity of cow's milk substitutes. Clin and Exp Allergy; 1993; 23: 205-210.
  6. Spuergin, P; Walter, M; Schiltz, E; Deichman, K; Forster, J; Mueller. Allergenicity of a -caseins from cow, sheep, and goat. Allergy; 1997; 52: 293-298.