Whole Allergen
f88 Mutton
f88 Mutton Scientific Information
Type:
Whole Allergen
Display Name:
Mutton
Latin Name:
Ovis spp.
Other Names:
Mutton
Summary
Mutton is regarded as a weak allergen and preparations of lamb meat have been suggested as an alternative to hydrolyzed cow's milk formulas
Molecular Aspects
Cross-reactivity
>Sheep and goats belong to Caprines of the order Artiodactyla (even-toed hoofed animals). The true sheep which resembles the domesticated animal is Ovis. Many breeds exist and a characteristic feature is the animal’s climbing ability, making it appropriate for grazing in mountainous areas. Lamb is less than one year. Mutton is the meat from animals more than one year old. Beef and mutton share allergens and it cannot be excluded that mutton has some allergens in common with sheep wool. Mutton is regarded as a weak allergen and preparations of lamb meat have been suggested as an alternative to hydrolyzed cow’s milk formulas (1). The allergenicity of the meat is decreased by heating and pepsin treatment (2, 3).
Compiled By
Last Reviewed-  April 2022
References
  1. Weisselberg, B; Dayal, Y; Thompson, JF; Doule, MS; Senior, B; Grand, J. A lamb-meat-based formula for infants allergic to casein hydrolysate formulas. Clin Pediatr Phila; 1996; 35: 491-495.
  2. Restani, P; Fiocchi, A; Restelli, AR; Velona, T; Beretta, B; Giovannini, M; Galli, CL. Effect of technological treatment on digestability and allergenicity of meat-based baby foods. J Am Coll Nutr; 1997; 16: 376-382.
  3. Fiocchi, A; Restani, P; Riva, E; Restelli, AR; Biasucci, G; Galli, CL; Giovannini, M. Meat allergy: II – Effects of food processing and enzymatic digestion on the allergenicity of bovine and ovine meats. J Am Coll Nutr; 1995; 14: 245-250.