Whole Allergen
f92 Banana
f92 Banana Scientific Information
Type:
Whole Allergen
Display Name:
Banana
Route of Exposure:
Ingestion
Family:
Musaceae
Species:
M.acuminata/sapientum/paradisiaca
Latin Name:
Musa acuminata/sapientum/paradisiaca
Other Names:
Plantain
Summary
Banana (Musa acuminata/sapientum/paradisiaca) is a perennial herb majorly found in the tropical regions. India, China, Ecuador, Brazil, and the Philippines are the top producers of banana in the world. Banana is a common food in human diet worldwide due to its high nutritional content. However, banana is found to cause allergic reactions in some individuals. The prevalence of banana allergy among general population is considered uncommon, with an estimated to be as low as 0.04% to up to 1.2% globally; however, an increasing trend on the number of cases on hypersensitivity reactions to banana in infants as well as in adults are being reported till date. Ingestion of fresh, uncooked banana fruit is the main cause of allergy as most banana allergens are sensitive to heat. Clinical manifestation of banana allergy ranges from mild local symptoms, referred to as oral allergy syndrome to severe anaphylactic reactions, including asthma and gastrointestinal symptoms in rare cases. The identified banana allergens are profilin - actin binding protein (Mus a 1), class I chitinase (Mus a 2), non-specific lipid transfer protein (Mus a 3), thaumatin-like protein (Mus a 4), beta-1,3-glucanase (Mus a 5) and ascorbate peroxidase (Mus a 6). Extensive cross-reactivity of banana with latex, pollens (olive, birch, palm, ragweed, and hazelnut), and fruits (melons, peach, kiwi, avocado, and pineapple) has been observed.
Allergen
Nature
Banana (Musa acuminata/sapientum/paradisiaca) is a perennial herb with huge leaves (1). It has a pseudo stem (3-8 m long with 6-25 cm diameter) at base with brown or black blotches (2).
Banana plant is well known for its fruit, which is a good source of nutrition because of the presence of abundant minerals, antioxidants, vitamins, low glycemic carbohydrates, and fiber (1).
Bananas are usually divided into two types- dessert type and cooking type. On ripening, dessert types are consumed raw. Various processing methods like boiling, frying, brewing, powdering, or roasting can be applied to the cooking types (starchy) before consumption. Plantains are the most well-known cooking bananas; also there is no sharp distinction between the plantain and the banana neither genetically nor botanically (2).
Besides global consumption of banana (raw or cooked), it has other uses, such as a flavoring agent in processed foods and an ingredient of cosmetics (3).
Habitat
Banana is majorly found in the tropical regions and less commonly in sub-tropical regions (2). They grow well at an altitude less than 2000 m at a temperature of 28℃ and in humid, well-drained clayey soil (4). The wild bananas are found to grow in warm regions with a limited tolerance to wide temperature variation; however, a few species may grow in cool climates. Moreover, bananas are unable to survive in drought conditions (2).
Taxonomy
Bananas belong to the family Musaceae, which consists of two genera Musa and Ensete. The two common Musa species are M. paradisiaca and M. sapientum. M. pardisiaca was described from a plantain and M. sapientum from the desert banana (2). M. accuminata is another species belonging to the family Musaceae and is well known for fruit allergy for more than 50 years (3).
Taxonomic tree of Banana (5)
Domain Eukaryota
Kingdom Viridiplantae
Phylum Streptophyta
Subphylum Streptophytina
Class Magnoliopsida
Order Zingiberales
Family Musaceae
Genus Musa
Species M. acuminata/sapientum/paradisiaca
Taxonomic tree of Banana (5)
Tissue
The banana fruit is subcylindrical with a length of 8-13 cm and diameter of 1.5-3.0 cm. The seeds present in the fruit are dull black, often smooth with a minute round projection on the surface (2). Banana fruit has been considered to be one important source of food allergen. The amines that are present in the fleshy part of the banana fruit can lead to allergic reactions. Banana allergens are usually glycoproteins or immunoglobulin E (IgE) reactive proteins. Banana allergen Mus a 5 (beta-1,3-glucanase) is abundantly found in banana pulp (1).
Epidemiology
Worldwide distribution
Banana is a common food in human diet worldwide due to its high nutritional content. Nonetheless, banana is found to cause allergic reactions in some individuals (6). The prevalence of banana allergy among general population is considered uncommon, with an estimated to be as low as 0.04% to up to 1.2% globally in different studies. However, increasing number of cases on hypersensitivity reactions to banana in infants as well as in adults are being reported till date (3). A European Community Respiratory Health Survey analyzed food sensitization (testing up to 24 foods) among 4522 young adults from 13 countries across globe. Sensitization pattern to banana along with walnut, sunflower, poppy, melon, fish, egg, milk, and soy was found to be in the range of 0.2% to 2.5% (7).
A multicenter, cross-sectional EuroPrevall study conducted to detect the prevalence of self-reported food allergy and food sensitization in 16,935 European school-age children (mean age 8.9 years) found self-reported food allergy prevalence for banana to be 1.1% (8). A German study evaluating case histories of 380 food-allergic patients (mostly suffering from pollen-related food allergy, PFAS) found 16 patients showing immediate allergic reactions to banana (9).
In a case-control study on 80 allergic children (40 each with/without history of banana allergy) in Egypt, 7.5% (3/40) were diagnosed with banana allergy based on positive history, elevated serum specific IgE to banana, and positive skin prick test (SPT). The prevalence of banana allergy was higher in this study compared to the general population (0.04%-1.2%), which may due to the inclusion of only atopic individuals in the study (10).
Further, in a group of 689 allergic patients (15-73 years) from various allergy centers in Pakistan, 7.1% (49/689) patients showed a positive SPT to banana allergen (11). In a 5-year cross-sectional study testing for food allergy among 5161 patients of all age groups in Kolkata, India, the highest rate of sensitization (32.4%) was observed for banana among all other foods tested (12).
A cross-sectional retrospective study on 365 American children in Honduras (1 to 18 years) reported 2.2% children sensitive to banana (13). Further, another study on 160 Columbian children (of age < 2years) with food allergies reported 23% (47/160) to be sensitive to one food based on specific IgE measurement. Banana sensitivity was found in 10% of them  (14).
Risk factors
Banana, rich in vitamins, minerals, and fibers, is often given to infants at a very early age (3). It has been postulated that infants who are exposed to banana allergens at an early age may be at a risk for the development of banana sensitization. A case study reported an infant to be sensitized to banana. He was probably exposed to the banana allergens via the placenta or breast milk during pregnancy or lactation, respectively or may be even orally by sucking mother’s thumb (15).
Pediatric issues
Banana is a commonly used food for infants. In Spain and other European countries, non-reported symptomatic cases of banana hypersensitivity among infants is on rise (16). In accordance to this, a study by Dey et al (2014 ) with participants of all ages found higher prevalence of banana allergy among children below 15 years of age (12).
Environmental Characteristics
Worldwide distribution
Worldwide, bananas are one of the most important food crops grown and consumed in over 100 countries (1). Bananas are found in wide geographic span in the tropics, ranging from 175º E to 150º W longitude and from 30º N to 23º S latitude. India is the top producer of banana in the world with 15% of the total production, followed by China, Ecuador, Brazil, and the Philippines each producing 5-6% of the total. Moreover, south and southeast Asia, and the Pacific contribute to 37% of the worldwide production of bananas. Tropical Africa produces 30%, Central and South America and the Caribbean produce 26%, while about 7% is produced in other countries of the world (2). Further, plantain is considered as primary and secondary staple food in Africa, Pacific Ocean islands, Latin America, and the Caribbean (2).
Route of Exposure
Main
Ingestion of fresh, uncooked banana fruit is the main cause of allergy as most banana allergens are sensitive to heat (17).
Clinical Relevance
Oral allergy symptoms and Anaphylaxis
Clinical manifestation of banana allergy ranges from mild local symptoms, referred to as oral allergy syndrome (OAS) to severe anaphylactic reactions (1).
An open-label banana challenge test conducted on 6 adults with a history of banana hypersensitivity depicted systemic symptoms like OAS in all of them with skin manifestation (generalized urticaria) as the most common symptom seen in 83.33% of cases. Other systemic symptoms included hypotension (33.3%), dyspnea, and diarrhea (16.67% each). A severe reaction like anaphylaxis was also observed in all the patients (half of the patients showed grade III anaphylaxis as per Ring and Messmer classification) (3).
In a telephonic survey involving 47 banana allergic children in Turkey, urticaria and gastrointestinal (GI) symptoms were observed in 9% and 6% respectively along with other atopic symptoms while none of the children reported anaphylaxis (18). In a study by Palacin et al. (2011) on 51 children with severe reactions towards banana, OAS was observed in 53% of patients, and systemic reactions in 47% including urticaria and vomiting, as well as anaphylaxis (n=3) (16).
Anaphylaxis was also observed in 2.8% of 319 Japanese patients with self-reported cases of allergic anaphylaxis due to banana hypersensitivity (19). Further, individual case reports of anaphylaxis due to banana allergy in infants have been reported. A 4-month-old Hispanic baby with a history of eczema started vomiting with other allergic symptoms like urticaria and cyanosis within 5 minutes of banana consumption (20). Another 7-month-old boy reported generalized urticaria, vomiting, and respiratory symptoms like wheezing within 2 hours of ingestion of crushed banana. The patient was tested negative to commercial extract of banana but reacted positively in SPT to fresh banana fruit (21). Moreover, another case of severe angioedema up to the glottis requiring intubation was reported in a 16-month-old girl who underwent an orthotopic liver transplant 8 months ago and was continued tacrolimus immunosuppression since then. As a vitamin source, she used to consume banana daily. Hypersensitivity to banana during tacrolimus medication was thought to be the cause of angioedema in the patient (22).
Further oral pruritis was observed in 33.3% of 6 adults with a history of banana hypersensitivity during an open-label banana challenge test (3).
Allergic rhinitis and Asthma
A telephonic survey on 47 banana-allergic Turkish children (age 6.2 ± 2.1 years) reported wheezing, allergic rhinitis (AR), and asthma in 60%, 15%, and 13% of patients respectively (18). Also, rhinorrhea was reported in 50% of 6 adults with a history of banana hypersensitivity during an open-label banana challenge test (3).
Atopic Dermatitis
In a Turkish study with 47 banana-allergic children, atopy was found in 87% (41/47) cases while 23% complained of atopic dermatitis (18).
Other diseases
A 47-year-old woman having a history of AR to Japanese cedar experienced symptoms of acute pancreatitis like nausea, vomiting, and diarrhea after consuming a banana. She had three attacks of acute pancreatitis each time after consuming a banana. She also experienced migraines and pruritis within 2 hours of eating bananas (23).
Molecular Aspects
Allergenic molecules
Banana contains a large number of different potential allergens such as pathogenesis related (PR) proteins: PR5 thaumatin-like, PR2 endo-β-1,3-glucanase, PR8 class III chitinase in addition to jacalin-related lectin (24).
To date, 6 allergenic proteins from Musa acuminata has been listed officially in the database of the World Health Organization/International Union of Immunological Studies (WHO/IUIS) Allergen Nomenclature Sub-Committee (25).
The table below provides detailed information on the allergenic proteins identified by WHO/IUIS (25):
Allergen Biochemical Name Molecular Weight (kDa) Allergenicity
Mus a 1 Profilin 15
  • Specific IgE-binding on immunoblots was found in 44% (7/16 sera) of suspected banana allergic patients (1, 9).
  • Sensitive to heat and gastric digestion (1).
  • High cross-reactivity with Bet v 2 (birch profilin) and Hev b 8 (latex profilin) (1, 9).
Mus a 2 Class I chintinase 33
  • Major banana allergen (1).
  • Two identified isoforms with hevein-like domain (1, 26).
  • The two purified isoforms (32 kDa and 34 kDa) gave a positive SPT in more than 50% of banana allergic patients (1, 26).
Mus a 3 Non-specific lipid transfer protein type 1 (nsLTP1) 9
  • PR-14 family of proteins (1).
  • IgE immunodetection seen in 20% of 36 sera of banana allergic patients (1, 16).
Mus a 4 Thaumatin-like protein 20
  • Major allergen in pediatric patients (16).
  • PR-5 family of proteins and is non-glycosylated (1).
  • Specific IgE reactivity seen in 72% of 51 sera and positive SPT in 50% of 12 sera of banana-allergic children (16).
Mus a 5 Beta-1,3-glucanase 30
  • Specific IgE reactivity was seen in 84% of 51 sera while positive SPT in only 20% of 12 sera of banana-allergic children (16).
  • Reactivity on ELISA immunoblots was found in 74% of 23 banana-sensitized patients’ sera (27). 
Mus a 6 Ascorbate peroxidase 27
  •  Most recent reported banana allergen.
  •  IgE reactivity was seen in 10 of 11 and 7 of 11 sera of banana allergic patients on immunoblots to natural and recombinant protein respectively (1, 25)
Allergen Biochemical Name Molecular Weight (kDa) Allergenicity
Biomarkers of severity
Mus a 5 can be used for component-based diagnosis of banana allergy in patients with negative IgE test with banana extract as reported by a study in 23 banana-allergic patients (27).
Cross-reactivity
Banana allergy has been correlated with latex and/or pollen suggesting that banana is responsible for latex-fruit, pollen-fruit, and latex-pollen-fruit syndrome (6).  
Latex-food syndrome is observed in about half of the latex allergic patients and linked to various fruits and vegetables including banana, kiwi, avocado, peach, pineapple, chestnut, tomato, potato, and pepper (16, 29). Individuals developing ‘latex-fruit’ syndrome (commonly seen in health care workers who frequently wear natural rubber latex gloves) are initially sensitized to latex and later become allergic to food due to cross-reacting IgE against allergenic proteins (9, 30). This latex sensitization results in allergic reactions like urticaria, GI, and in severe cases, anaphylaxis or even loss of consciousness (1).
Cross-reactivity of banana with pollens of different trees and weeds including palm, birch, plane tree, olive, ragweed, and hazelnut has also been observed (1, 6, 8, 28, 31, 32). It can lead to the development of banana allergic reactions, such as oral allergy symptoms like itching and swelling of the mouth and throat (1).
The possible cross-reactive molecules responsible for latex-pollen-fruit syndrome are the gluconases such as Hev b 2 from rubber tree latex (Hevea brasilianis), the Mus a 5 from banana (M. acuminata), and Ole e 9 from olive pollen. These molecules showed high sequence homology and have common IgE epitopes (1). Besides, cross reactivity also occurs between profilin of banana (Mus xp 1, M. parasiadica), and profilins of rubber tree latex (Hev b 8), olive pollen (Ole e 2), and fruits like kiwi (Act d 9), and ananas (Ana c1) (28). Furthermore, there is an association between ragweed-melon-banana and the probable cross reacting molecules are either profilins or LTPs, such as Mus xp 1 (profilin) of banana, Amb a 6 (LTP), Amb a 8 (profilin) of ragweed (Ambrosia artemisilifolia), Cit la 2 of watermelon (Citrullus lanatus), Cuc m 2 of muskmelon/cantaloupe (Cucumis melo), Cuc p 2 of zucchini (Cucurbita pepo), and Cuc s 2 of cucumber (Cucumis sativus) (28). 
Additionally, the cross-reactivity between banana and fruits like peach and pineapple was studied and it was observed that more than 75% of 36 banana allergic patients recognized the major allergen of peach (Pru p 3, nsLTP) on IgE immunoblots in addition to pineapple bromelain (a marker of cross-reactive carbohydrate determinants, CCD) (16).
Compiled By
Author: Turacoz Healthcare Solutions
Reviewer: Dr. Fabio Iachetti
Last reviewed: January 2021
References
  1. Nikolić JP, Milica & Gavrovic-Jankulovic, Marija. Banana as a food allergen source. . 2016.
  2. Nayar. The Bananas: Botany, Origin, Dispersal.  . Horticultural Reviews, Volume 36 (pp117 - 164). 2010.
  3. Thongkhom R, Oncham S, Sompornrattanaphan M, Laisuan W. Banana anaphylaxis in Thailand: case series. Asia Pac Allergy. 2020;10(1):e4.
  4. Umuhoza Karemera N. Banana production. Lambert academic publishingEditor: Cornelia Schidu. 2014.
  5. NCBI. Taxonomic browser- Musa acuminata. 2020 [ December 1st, 2020.]. Available from: https://www.ncbi.nlm.nih.gov/Taxonomy/Browser/wwwtax.cgi?mode=info&id=4641.
  6. Nikolic J, Mrkic I, Grozdanovic M, Popovic M, Petersen A, Jappe U, et al. Protocol for simultaneous isolation of three important banana allergens. J Chromatogr B Analyt Technol Biomed Life Sci. 2014;962:30-6.
  7. Sicherer SH. Epidemiology of food allergy. J Allergy Clin Immunol. 2011;127(3):594-602.
  8. Lyons SA, Clausen M, Knulst AC, Ballmer-Weber BK, Fernandez-Rivas M, Barreales L, et al. Prevalence of Food Sensitization and Food Allergy in Children Across Europe. J Allergy Clin Immunol Pract. 2020;8(8):2736-46 e9.
  9. Reindl J, Rihs HP, Scheurer S, Wangorsch A, Haustein D, Vieths S. IgE reactivity to profilin in pollen-sensitized subjects with adverse reactions to banana and pineapple. Int Arch Allergy Immunol. 2002;128(2):105-14.
  10. El-Sayed ZA, El-Ghoneimy DH, El-Shennawy D, Nasser MW. Evaluation of banana hypersensitivity among a group of atopic egyptian children: relation to parental/self reports. Allergy Asthma Immunol Res. 2013;5(3):150-4.
  11. Inam M, Shafique RH, Roohi N, Irfan M, Abbas S, Ismail M. Prevalence of sensitization to food allergens and challenge proven food allergy in patients visiting allergy centers in Rawalpindi and Islamabad, Pakistan. Springerplus. 2016;5(1):1330.
  12. Dey D, Ghosh N, Pandey N, Gupta Bhattacharya S. A hospital-based survey on food allergy in the population of Kolkata, India. Int Arch Allergy Immunol. 2014;164(3):218-21.
  13. Gonzales-Gonzalez VA, Diaz AM, Fernandez K, Rivera MF. Prevalence of food allergens sensitization and food allergies in a group of allergic Honduran children. Allergy Asthma Clin Immunol. 2018;14:23.
  14. Hossny E, Ebisawa M, El-Gamal Y, Arasi S, Dahdah L, El-Owaidy R, et al. Challenges of managing food allergy in the developing world. World Allergy Organ J. 2019;12(11):100089.
  15. Moreno-Ancillo A, Dominguez-Noche C, Gil-Adrados AC, Cosmes PM. Allergy to banana in a 5-month-old infant. Pediatr Allergy Immunol. 2004;15(3):284-5.
  16. Palacin A, Quirce S, Sanchez-Monge R, Bobolea I, Diaz-Perales A, Martin-Munoz F, et al. Sensitization profiles to purified plant food allergens among pediatric patients with allergy to banana. Pediatr Allergy Immunol. 2011;22(2):186-95.
  17. Dayıoğlu A AS, Nacaroğlu HT, Erdem SB. . The Clinical Spectrum of Reactions Due to Banana Allergy. Bagcilar Med Bull 2020;5(2):60-63. 2020.
  18. Kocacik Uygun DF, Filiz S, Bingol A. An evaluation of banana allergy in children living in the Mediterranean region. Turk J Med Sci. 2018;48(3):469-75.
  19. Imamura T, Kanagawa Y, Ebisawa M. A survey of patients with self-reported severe food allergies in Japan. Pediatr Allergy Immunol. 2008;19(3):270-4.
  20. O'Keefe AW, Ben-Shoshan M. A 4-month-old baby boy presenting with anaphylaxis to a banana: a case report. J Med Case Rep. 2014;8:62.
  21. Hauswirth DW, Burks AW. Banana anaphylaxis with a negative commercial skin test. J Allergy Clin Immunol. 2005;115(3):632-3.
  22. Yilmaz A, Artan R, Uguz A, Arslan G, Akcam M. Severe angioedema caused by banana allergy under tacrolimus immunosuppression. Transplant Proc. 2005;37(10):4251-3.
  23. Inamura H, Kashiwase Y, Morioka J, Kurosawa M. Acute pancreatitis possibly caused by allergy to bananas. J Investig Allergol Clin Immunol. 2005;15(3):222-4.
  24. Receveur-Brechot V, Czjzek M, Barre A, Roussel A, Peumans WJ, Van Damme EJ, et al. Crystal structure at 1.45-A resolution of the major allergen endo-beta-1,3-glucanase of banana as a molecular basis for the latex-fruit syndrome. Proteins. 2006;63(1):235-42.
  25. WHO/IUIS. Allergen Nomenclature “Musa acuminata - All Allergen.”, WHO/IUIS Allergen Nomenclature Sub-Committee. 2019 [December 2nd, 2020. ]. Available from: http://www.allergen.org/search.php?allergenname=mus&searchname=Search.
  26. Sanchez-Monge R, Blanco C, Diaz-Perales A, Collada C, Carrillo T, Aragoncillo C, et al. Isolation and characterization of major banana allergens: identification as fruit class I chitinases. Clin Exp Allergy. 1999;29(5):673-80.
  27. Aleksic I, Popovic M, Dimitrijevic R, Andjelkovic U, Vassilopoulou E, Sinaniotis A, et al. Molecular and immunological characterization of Mus a 5 allergen from banana fruit. Mol Nutr Food Res. 2012;56(3):446-53.
  28. Popescu FD. Cross-reactivity between aeroallergens and food allergens. World J Methodol. 2015;5(2):31-50.
  29. De Martinis M, Sirufo MM, Suppa M, Ginaldi L. New Perspectives in Food Allergy. Int J Mol Sci. 2020;21(4).
  30. Radauer C, Adhami F, Furtler I, Wagner S, Allwardt D, Scala E, et al. Latex-allergic patients sensitized to the major allergen hevein and hevein-like domains of class I chitinases show no increased frequency of latex-associated plant food allergy. Mol Immunol. 2011;48(4):600-9.
  31. Grob M, Reindl J, Vieths S, Wuthrich B, Ballmer-Weber BK. Heterogeneity of banana allergy: characterization of allergens in banana-allergic patients. Ann Allergy Asthma Immunol. 2002;89(5):513-6.
  32. Waserman S, Bégin, P. & Watson, W. IgE-mediated food allergy. . Allergy Asthma Clin Immunol 14, 55. 2018.