Tim, a 6-year-old with a presumed history of peanut allergy, visits his healthcare provider because he’s never been tested for allergen sensitization.
Tim’s healthcare provider conducts a full clinical history and physical examination and decides to test using whole peanut allergen with reflex to components.
Find out how specific IgE testing can show if Tim has a clinical peanut allergy.
These results, together with this patient's case history and symptoms, help confirm the diagnosis.
Test | Type | Tim's Results |
Peanut |
Whole Allergen |
1.6 |
Ara h 1 |
Allergen Component |
<0.1 |
Ara h 2 |
Allergen Component |
<0.1 |
Ara h 3 |
Allergen Component |
<0.1 |
Ara h 8 |
Allergen Component |
2.79 |
Ara h 9 |
Allergen Component |
<0.1 |
Tim’s sensitization to Ara h 8 is associated with localized reactions, like itching and tingling of the mouth and lips, but low risk of systemic reaction. Tim may actually be sensitized to birch pollen, which is cross-reactive and responds similar to Ara h 8. Tim shows no detectable sensitization to three nut storage proteins associated with severe reactions.1-3
REFINED DIAGNOSIS
HEALTHCARE PROVIDER MANAGEMENT PLAN
FOLLOW UP
The people, places, and events depicted in these case studies and photographs do not represent actual patients, nor are they affiliated in any way with Thermo Fisher Scientific.