Identify infectious complications in transplant patients with high specificity
The major postoperative incidents endangering postive outcomes after transplantation are acute rejections and infections. Most mediators of inflammatory events indicate not only immunological reactions but also infections. However, it can be difficult to distinguish between infectious versus non-infectious inflammation.
After transplantation infectious complications significantly increase PCT values while in uncomplicated cases and rejections there is only a slight non-specific PCT increase, depending on the type and extension of intervention. Generally, these unspecific PCT increases reach their maximum one or two days after surgery and then return to normal.8
The use of anti-lymphocyte/thymocyte globuline preparations (or anti CD3) for the treatment of acute rejection after kidney-, heart- and liver-transplantation can induce PCT release for a brief period.14
An infection, but not rejection, causes an early and dramatic increase in PCT and allows the identification of infectious complications in contrast to CRP, which was not suitable to differentiate between both complications.8,15