Case Study
Trevor Van Schooneveld, MD, FACP
Internal Medicine, Infectious Disease
Explore similar topics
Submit your own PCT Case Study and you may be selected to be featured on procalcitonin.com.
*NEWS or National Early Warning Score is a scoring system based on 6 common physiological measurements: respiration rate, oxygen saturation, temperature, blood pressure, heart rate and level of alertness. The score ranges from 0 to 20 points; a level of 5-6 means a patient can be very ill and requires regular monitoring and urgent review.