Case Study
Stopping Empirical Antibiotics in COVID-19 Patients
Neil Powell
Associate Director, Antimicrobial Stewardship, Royal Cornwall Hospital Trust
Consultant Antimicrobial Pharmacist
Learn more about implementing procalcitonin testing
in your hospital.
Explore similar topics
Key Takeaways
Takeaway #1
Takeaway #1
PCT below 0.25 μg/L indicates low likelihood of bacterial infection.
Takeaway #2
Takeaway #2
The combination of a confirmed COVID-19 pneumonitis and low PCT supports decision to stop antibiotics.
Takeaway #3
Takeaway #3
Recent evidence showed that in COVID-19 patients with no evidence of bacterial infection, unnecessary antibiotics might prove harmful to the patient.1
Share your Clinical Experience
Submit your own PCT Case Study and you may be selected to be featured on procalcitonin.com. 
Submit a case study here
Share your experience
*NEWS or National Early Warning Score is a scoring system based on 6 common physiological measurements: respiration rate, oxygen saturation, temperature, blood pressure, heart rate and level of alertness. The score ranges from 0 to 20 points; a level of 5-6 means a patient can be very ill and requires regular monitoring and urgent review.
Related Resources
References
  1. Liu et al. International Immunopharmacology 90 (2021): 107157. https://doi.org/10.1016/j.intimp.2020.107157
Back to Top Arrow