Case Study
Lower Respiratory Tract Infection (LRTI):
Is it Viral or Bacterial?
Shawn Depcinski
PharmD
Antibiotic Stewardship
Learn more about implementing procalcitonin testing
in your hospital.
Explore similar topics
Key Takeaways
Takeaway #1
Takeaway #1
Bacterial co-infection may occur in approximately one-fourth of hospitalized patients with influenza.1
Takeaway #2
Takeaway #2
PCT has a high negative predictive value (NPV) at ≤ 0.25 μg/L, which aids in determining the need for antibiotics in LRTI. When uncertainty exists with an initial negative PCT result, repeating the test within 6 to 24 hours can assist with confirming the initial result, and indicate that antibiotics may no longer be necessary.
Takeaway #3
Takeaway #3
In this case, PCT aided in ruling out bacterial infection, allowing the clinicians to avoid giving unnecessary antibiotics and to continue with the appropriate anti-viral therapy.
Share your Clinical Experience
Submit your own PCT Case Study and you may be selected to be featured on procalcitonin.com. 
Submit a case study here
Share your experience
Related Resources
References
  1. Klein EY, Monteforte B, Gupta A, Jiang W, May L, Hsieh YH, et al. The frequency of influenza and bacterial coinfection: a systematic review and meta-analysis. Influenza Other Respir Viruses. 2016 Sep;10(5):394-403. 
Back to Top Arrow