Case Study
Heart Failure vs. Bacterial Pneumonia
Trevor Van Schooneveld, MD, FACP
Internal Medicine, Infectious Disease
Learn more about implementing procalcitonin testing
in your hospital.
Explore similar topics
Key Takeaways
Takeaway #1
Takeaway #1
This patient had a diagnosis of heart failure secondary to acute myocardial infarction (AMI) two days prior, but the clinical presentation offered multiple possibilities of differential diagnoses.
Takeaway #2
Takeaway #2
PCT gives insight into the presence or absence of systemic bacterial infection.
Takeaway #3
Takeaway #3
Clinicians were able to focus on treating the cardiac issues at hand.
Share your Clinical Experience
Submit your own PCT Case Study and you may be selected to be featured on procalcitonin.com. 
Submit a case study here
Share your experience
*NEWS or National Early Warning Score is a scoring system based on 6 common physiological measurements: respiration rate, oxygen saturation, temperature, blood pressure, heart rate and level of alertness. The score ranges from 0 to 20 points; a level of 5-6 means a patient can be very ill and requires regular monitoring and urgent review.
Related Resources
Back to Top Arrow