Calcitonin is exclusively produced by C-cells of the thyroid gland in response to hormonal stimuli. It plays an important role in the pathway and regulation of calcium and phosphate in bone metabolism and is the main medullary thyroid carcinoma (MTC) tumor marker.4
In healthy individuals, the prohormone PCT is rapidly processed into the mature hormone calcitonin. Therefore, PCT values in non-infected individuals are very low: < 0.1 µg/L.4
In case of bacterial insult, PCT can be induced outside the thyroid gland, in the parenchymal tissue of many organs, and then released into circulation in large amounts4
At first, cytokine-stimulated adherent monocytes release PCT in low quantities (< 2h). While this synthesis is limited, it plays an important role in the initiation of PCT synthesis in storage tissues of humans.4
This PCT burst is initiated in all storage tissues (peak 12-24h). Parenchymal tissue is the most common type of tissue in humans. As a result, extreme concentrations of PCT can be generated (100,000-fold increase in contrast to physiological concentrations).4 The PCT burst continues as long as the stimulus for synthesis exists.4
Additional experiments clarified why PCT is only synthesized during bacterial but not in viral infections. Cells were incubated with IL-1β and INF-γ, inflammatory cytokines known to be released during bacterial and viral infection, respectively. Whereas IL-1ß stimulated the PCT synthesis, the addition of IFN-y blocked this effect.5