48ab - Appl/Samples/Products/Resources/Contact
Multi scale microscopy workflow plot showing microCT, PFIB, and TEM
Plot of volume and resolution that can be analyzed by various instruments
in the multi-scale analysis workflow.
As materials continue to advance, it is becoming increasingly important to not only examine them at ever-higher resolutions but to obtain these observations within the relevant macroscopic context. This necessitates correlating different imaging modes to the same coordinates for truly contextual insight. Measurements must also be obtained quickly enough for practical application in real-world process control and failure analysis environments. Thermo Fisher Scientific offers a complete workflow for the observation of materials, combining correlated imaging at various scales with additional information such as chemical composition.
Multi-scale analysis begins with micro-scale observation with non-destructive spectroscopic techniques. X-ray microtomography (microCT) produces a complete, 3D rendering of the sample through serial X-ray scans. These scans, or 2D tomograms, are digitally combined to form the 3D structure. With Thermo Scientific Heliscan MicroCT, the series of circular scans is replaced by a single, continuous helical scan. This allows for faster scanning at a lower dose, increasing the accuracy and amount of information obtained. MicroCT observations can provide resolution as low as 400 nm, making it an ideal tool for non-destructive surveying of the sample prior to higher resolution characterization.
Once a region of interest is identified, DualBeam (focused ion beam and scanning electron microscopy, FIB-SEM) instrumentation is used for closer surface analysis and sample extraction. (Note that the focused ion beam can consist of a liquid metal ion source (gallium) or a plasma FIB.) The SEM enables nanoscale surface analysis while the FIB/PFIB is used for serial sectioning, or to extract a thin sample lamella for further observation with transmission electron microscopy (TEM). The addition of a femtosecond laser to the PFIB-SEM allows for even more rapid sample preparation, cross-sectioning or serial sectioning. Subsequent TEM analysis provides atomic-scale materials characterization for complete insight into a sample’s elemental and structural composition.
True multi-scale microscopy generates high quality and reliable imaging across all instruments while also accurately aligning them into a complete representation of the sample. With Thermo Scientific automation and data analysis software, the entire multi-scale workflow becomes a guided and routine procedure that can be readily integrated into your process or quality control environment.
Multi scale microscopy workflow plot showing software and hardware
The multi-scale analysis workflow offered by Thermo Fisher Scientific integrating software and hardware.
*Maps is not directly integrated with mCT, it can load processed CT data.
**AutoScript is not available yet on HeliScan and TEM microscopes.
Resources
Video Player is loading.
Current Time 0:00
Duration 0:14
Loaded: 0.00%
Stream Type LIVE
Remaining Time 0:14
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • default, selected
    HeliScan MicroCT analysis used in the correlative study of defects in an oil filter casing made of a glass-fiber-reinforced composite.
    Video Player is loading.
    Current Time 0:00
    Duration 2:38
    Loaded: 0.00%
    Stream Type LIVE
    Remaining Time 2:38
    1x
      • Chapters
      • descriptions off, selected
      • captions off, selected
      • default, selected
      Characterizing material failure of an additively manufactured Inconel 718 part with multi-scale analysis. Performed in collaboration with the University of Manchester.
      Video Player is loading.
      Current Time 0:00
      Duration 0:14
      Loaded: 0.00%
      Stream Type LIVE
      Remaining Time 0:14
      1x
        • Chapters
        • descriptions off, selected
        • captions off, selected
        • default, selected
        HeliScan MicroCT analysis used in the correlative study of defects in an oil filter casing made of a glass-fiber-reinforced composite.
        Video Player is loading.
        Current Time 0:00
        Duration 2:38
        Loaded: 0.00%
        Stream Type LIVE
        Remaining Time 2:38
        1x
          • Chapters
          • descriptions off, selected
          • captions off, selected
          • default, selected
          Characterizing material failure of an additively manufactured Inconel 718 part with multi-scale analysis. Performed in collaboration with the University of Manchester.
          Applications
          Quality control and failure analysis using electron microscopy
          Quality control and failure analysis
          Quality control and assurance are essential in modern industry. We offer a range of EM and spectroscopy tools for multi-scale and multi-modal analysis of defects, allowing you to make reliable and informed decisions for process control and improvement.
          Fundamental Materials Research_R&D_Thumb_274x180_144DPI
          Fundamental Materials Research
          Novel materials are investigated at increasingly smaller scales for maximum control of their physical and chemical properties. Electron microscopy provides researchers with key insight into a wide variety of material characteristics at the micro- to nano-scale.
          Samples
          Battery Research
          Battery development is enabled by multi-scale analysis with microCT, SEM and TEM, Raman spectroscopy, XPS, and digital 3D visualization and analysis. Learn how this approach provides the structural and chemical information needed to build better batteries.
          Learn more ›
          Polymers Research
          Polymer microstructure dictates the material’s bulk characteristics and performance. Electron microscopy enables comprehensive microscale analysis of polymer morphology and composition for R&D and quality control applications.
          Learn more ›
          Metals Research
          Effective production of metals requires precise control of inclusions and precipitates. Our automated tools can perform a variety of tasks critical for metal analysis including; nanoparticle counting, EDS chemical analysis and TEM sample preparation.
          Learn more ›
          Catalysis Research
          Catalysts are critical for a majority of modern industrial processes. Their efficiency depends on the microscopic composition and morphology of the catalytic particles; EM with EDS is ideally suited for studying these properties.
          Learn more ›
          Oil and Gas
          As the demand for oil and gas continues, there is an ongoing need for efficient and effective extraction of hydrocarbons. Thermo Fisher Scientific offers a range of microscopy and spectroscopy solutions for a variety of petroleum science applications.
          Learn more ›
          Geological Research
          Geoscience relies on consistent and accurate multi-scale observation of features within rock samples. SEM-EDS, combined with automation software, enables direct, large-scale analysis of texture and mineral composition for petrology and mineralogy research.
          Learn more ›
          Fibers and Filters
          The diameter, morphology and density of synthetic fibers are key parameters that determine the lifetime and functionality of a filter. Scanning electron microscopy (SEM) is the ideal technique for quickly and easily investigating these features.
          Learn more ›
          Automotive Materials Testing
          Every component in a modern vehicle is designed for safety, efficiency, and performance. Detailed characterization of automotive materials with electron microscopy and spectroscopy informs critical process decisions, product improvements, and new materials.
          Learn more ›
          Products
          Style Sheet for Instrument Cards Original
          Apreo ChemiSEM System
          • Integrated SEM imaging and chemical characterization
          • Enhanced automation to simplify workflows
          • Extended source lifetime and schedulable upgrades
          AutoScript TEM
          • Provides a direct link between research needs and microscope automation
          • Enables improved reproducibility and accuracy
          • Focuses time on the microscope for higher throughput
          FIB-SEM and Laser Ablation
          • All three beams have same coincident point for accurate and repeatable cut placement
          • Millimeter-scale cross sections with up to 15,000x faster material removal than a typical FIB
          • Statistically relevant deep subsurface and 3D data analysis
          Thermo Scientific Helios Hydra plasma focused ion beam scanning electron microscope (DualBeam)
          Helios Hydra DualBeam
          • 4 fast switchable ion species (Xe, Ar, O, N) for optimized PFIB processing of a widest range of materials
          • Ga-free TEM sample preparation
          • Extreme high resolution SEM imaging
          Helios 5 PFIB DualBeam
          • Gallium-free STEM and TEM sample preparation
          • Multi-modal subsurface and 3D information
          • Next-generation 2.5 μA xenon plasma FIB column
          Thermo Scientific Scios 2 plasma focused ion beam scanning electron microscope (DualBeam)
          Scios 2 DualBeam
          • Full support of magnetic and non-conductive samples
          • High throughput subsurface and 3D characterization
          • Advanced ease of use and automation capabilities
          Spectra Ultra
          • New imaging and spectroscopy capabilities on the most beam sensitive materials
          • A leap forward in EDX detection with Ultra-X
          • Column designed to maintain sample integrity.
          Spectra 300
          • Highest-resolution structural and chemical information at the atomic level
          • Flexible high-tension range from 30-300 kV
          • Three lens condenser system
          Spectra 200
          • High-resolution and contrast imaging for accelerating voltages from 30-200 kV
          • Symmetric S-TWIN/X-TWIN objective lens with wide-gap pole piece design of 5.4 mm
          • Sub-Angstrom STEM imaging resolution from 60 kV-200 kV
          Talos F200S TEM
          • Intuitive and easy-to-use automation software
          • Available with Super-X EDS for rapid quantitative chemical analysis
          • High-throughput with simultaneous multi-signal acquisition
          Talos F200i TEM
          • Compact design with X-TWIN objective lens
          • Available with S-FEG, X-FEG, and X-CFEG
          • Flexible and fast EDS options for comprehensive elemental analysis
          Thermo Scientific Talos F200C transmission electron microscope (TEM)
          Talos F200C TEM
          • High-contrast and high-quality TEM and STEM imaging
          • 4k x 4k Ceta CMOS camera options for large FOV and high read-out speeds
          • Large pole piece gap and multiple in situ options
          Talos F200X TEM
          • High-resolution, EDS cleanliness, and quality in 2D as well as 3D
          • X-FEG and X-CFEG available for the highest brightness and energy resolution
          • High accuracy and repeatable results with integrated Thermo Scientific Velox Software
          AutoTEM 5
          • Fully automated in situ S/TEM sample preparation
          • Support of top-down, planar and inverted geometry
          • Highly configurable workflow
          • Easy to use, intuitive user interface
          Thermo Scientific Auto Slice and View 4.0 serial section electron microscopy software
          Auto Slice and View 4.0 Software
          • Automated serial sectioning for DualBeam
          • Multi-modal data acquisition (SEM, EDS, EBSD)
          • On-the-fly editing capabilities
          • Edge based cut placement
          Avizo Software
          Materials Science
          • Support for multi-data/multi-view, multi-channel, time series, very large data
          • Advanced multi-mode 2D/3D automatic registration
          • Artifact reduction algorithms
          Thermo Scientific Maps electron microscopy software
          Maps Software
          • Acquire high-resolution images over large areas
          • Easily find regions of interest
          • Automate image acquisition process
          • Correlate data from different sources
          Style Sheet for Komodo Tabs
          Style Sheet to change H2 style to p with em-h2-header class
          Contact us
          Style Sheet for Support and Service footer
          Style Sheet for Fonts
          Style Sheet for Cards
          Electron microscopy services for
          the materials science
          To ensure optimal system performance, we provide you access to a world-class network of field service experts, technical support, and certified spare parts.