Multiskan FC Microplate Photometer
Wide variety of applications from enzyme kinetic studies to Lowry assays.
Multiskan FC brings features for enhanced usability together with over 40 years of experience with the well-known Multiskan product brand. The Multiskan FC has a 340–850 nm wavelength range, enabling a wide variety of applications from enzyme kinetic studies to Lowry assays. The instrument provides fast and accurate measurements enabling complete 96-well plate reading in less than seven seconds. The instrument is also equipped with linear shaking as standard. A model with an incubator enables incubation temperatures up to 50°C and the capacity to read 384-well plates. SkanIt software supports optimal use of the instrument with a visual workflow, effortless data analysis and exporting capabilities.
Features
In addition to the common features of all Microplate Readers the Multiskan FC offers:
SkanIt software
SkanIt software can be used to control the Multiskan FC and supports optimal use of the instrument’s features with a visual workflow, easy data analysis, and exporting capabilities.
Broad wavelength range
Wavelength range of 340–850 nm for a wide variety of applications such as ELISA immunoassays, protein quantification, endotoxin, cytotoxicity and proliferation assays, enzyme assays and growth curves.
Preinstalled filters
Comes equipped with an eight-position filter wheel with three standard filters, 405 nm, 450 nm and 620 nm. Easy-to install additional filters are available for order.
Proven performance and reliable results
Built in self-diagnostics, IQ/OQ and verification tools make this instrument reliable and trustworthy.
Specifications
Attribute
Description
Technology
Absorbance
Wavelength range (nm)
340–850
Wavelength selection
Filters
Plate format
96 wells
(384 wells optional)
Incubation
Optional
Shaking
Yes
User interface
Color LCD or PC
Robot compatibility
Yes
21 CFR Part 11
Yes, with SkanIt Software for Microplate Readers, Drug Discovery Edition