de69 - Techniques/Samples/Products/Resources/Contact Us
As the dimensions of semiconductor device structures shrink and become more complex, defect localization and failure analysis become more critical, and more challenging. High-density interconnects, wafer-level stacking, flexible electronics, and integral substrates mean that failure-inducing defects have more places to hide. Even worse, these failures can occur at the device packaging stage, resulting in an intolerable loss of yield and an increase in time-to-market.
Advanced analytical tools are essential for the detection of any electrical defects that can negatively influence yield, reliability, or performance. With the right equipment, the time and cost associated with electrical fault isolation can be reduced by quickly extracting comprehensive defect data from the sample.
Thermo Fisher Scientific offers advanced analytical tools for fast physical and electrical failure analysis. Our workflows allow you to localize and characterize subtle electrical issues that affect yield, performance, and reliability. Click through to our technique or product pages for additional information.
Semiconductor defect localization and analysis workflow examples
  West Coast Failure Analysis Seminar: Ask the Experts
   Ask the experts your questions live and in-person June 3-4, 2025!
  Learn more
Style Sheet for Komodo Tabs
Techniques
Optical Fault Isolation
Increasingly complex designs complicate fault and defect isolation in semiconductor manufacturing. Optical fault isolation techniques allow you to analyze the performance of electrically active devices to locate critical defects that cause device failure.
Learn more ›
Thermal Fault Isolation
Uneven distribution of local power dissipation can cause large, localized increases in temperature, leading to device failure. We offer unique solutions for thermal fault isolation with high-sensitivity lock-in infrared thermography (LIT).
Learn more ›
Semiconductor TEM Imaging and Analysis
Thermo Scientific transmission electron microscopes offer high-resolution imaging and analysis of semiconductor devices, enabling manufacturers to calibrate toolsets, diagnose failure mechanisms, and optimize overall process yields.
Learn more ›
Semiconductor Analysis and Imaging
Thermo Fisher Scientific offers scanning electron microscopes for every function of a semiconductor lab, from general imaging tasks to advanced failure analysis techniques requiring precise voltage-contrast measurements.
Learn more ›
Nanoprobing
As device complexity increases, so does the number of places defects have to hide. Nanoprobing provides the precise localization of electrical faults, which is critical for an effective transmission electron microscopy failure analysis workflow.
Learn more ›
Semiconductor Laser Ablation
Laser ablation provides high-throughput milling of semiconductor devices for imaging and analysis with electron microscopy, while still preserving sample integrity. Access large-volume 3D data and optimize milling conditions to best suit your sample type.
Learn more ›
APT Sample Preparation
Atom probe tomography (APT) provides atomic-resolution 3D compositional analysis of materials. Focused ion beam (FIB) microscopy is an essential technique for high-quality, orientation, and site-specific sample preparation for APT characterization.
Learn more ›
Device Delayering
Shrinking feature size, along with advanced design and architecture, results in increasingly challenging failure analysis for semiconductors. Damage-free delayering of devices is a critical technique for the detection of buried electrical faults and failures.
Learn more ›
ESD Compliance Testing
Electrostatic discharge (ESD) can damage small features and structures in semiconductors and integrated circuits. We offer a comprehensive suite of test equipment which verifies that your devices meet targeted ESD compliance standards.
Learn more ›
Optical Fault Isolation
Increasingly complex designs complicate fault and defect isolation in semiconductor manufacturing. Optical fault isolation techniques allow you to analyze the performance of electrically active devices to locate critical defects that cause device failure.
Learn more ›
Thermal Fault Isolation
Uneven distribution of local power dissipation can cause large, localized increases in temperature, leading to device failure. We offer unique solutions for thermal fault isolation with high-sensitivity lock-in infrared thermography (LIT).
Learn more ›
Semiconductor TEM Imaging and Analysis
Thermo Scientific transmission electron microscopes offer high-resolution imaging and analysis of semiconductor devices, enabling manufacturers to calibrate toolsets, diagnose failure mechanisms, and optimize overall process yields.
Learn more ›
Semiconductor Analysis and Imaging
Thermo Fisher Scientific offers scanning electron microscopes for every function of a semiconductor lab, from general imaging tasks to advanced failure analysis techniques requiring precise voltage-contrast measurements.
Learn more ›
Nanoprobing
As device complexity increases, so does the number of places defects have to hide. Nanoprobing provides the precise localization of electrical faults, which is critical for an effective transmission electron microscopy failure analysis workflow.
Learn more ›
Semiconductor Laser Ablation
Laser ablation provides high-throughput milling of semiconductor devices for imaging and analysis with electron microscopy, while still preserving sample integrity. Access large-volume 3D data and optimize milling conditions to best suit your sample type.
Learn more ›
APT Sample Preparation
Atom probe tomography (APT) provides atomic-resolution 3D compositional analysis of materials. Focused ion beam (FIB) microscopy is an essential technique for high-quality, orientation, and site-specific sample preparation for APT characterization.
Learn more ›
Device Delayering
Shrinking feature size, along with advanced design and architecture, results in increasingly challenging failure analysis for semiconductors. Damage-free delayering of devices is a critical technique for the detection of buried electrical faults and failures.
Learn more ›
ESD Compliance Testing
Electrostatic discharge (ESD) can damage small features and structures in semiconductors and integrated circuits. We offer a comprehensive suite of test equipment which verifies that your devices meet targeted ESD compliance standards.
Learn more ›
Samples
Semiconductor Materials and Device Characterization
As semiconductor devices shrink and become more complex, new designs and structures are needed. High-productivity 3D analysis workflows can shorten device development time, maximize yield, and ensure that devices meet the future needs of the industry.
Learn more ›
Products
Style Sheet for Instrument Cards Original
Spectra Ultra
  • New imaging and spectroscopy capabilities on the most beam sensitive materials
  • A leap forward in EDX detection with Ultra-X
  • Column designed to maintain sample integrity.
Helios 5 EXL DualBeam
  • Low-voltage performance for high sample preparation quality
  • Ultra-high-resolution immersion-lens field-emission SEM column
  • Automated handling of 300 mm FOUP with EFEM (GEM300 compliant)
Helios 5 HX/Helios 5 UX/Helios 5 FX DualBeam
  • Fully automated, high-quality, ultra-thin TEM sample preparation
  • High throughput, high resolution subsurface and 3D characterization
  • Rapid nanoprototyping capabilities
Talos F200E TEM
  • High-quality (S)TEM imaging of semiconductor and microelectronic devices
  • Precise, high-speed chemical characterization with EDS
  • Dedicated semiconductor-related applications
Metrios AX TEM
  • Automation options to support quality, consistency, metrology, and reduced OPEX
  • Leverages machine learning for superior autofunctions and feature recognition
  • Workflows for both in-situ and ex-situ lamella preparation
Thermo Scientific Scios 2 plasma focused ion beam scanning electron microscope (DualBeam)
Scios 2 DualBeam
  • Full support of magnetic and non-conductive samples
  • High throughput subsurface and 3D characterization
  • Advanced ease of use and automation capabilities
ExSolve WTP DualBeam
  • Can prepare site-specific, 20 nm thick lamellae on whole wafers up to 300 mm in diameter
  • Addresses needs requiring automated, high-throughput sampling at advanced technology nodes
Thermo Scientific Verios 5 XHR scanning electron microscope (SEM)
Verios 5 XHR SEM
  • Monochromated SEM for sub-nanometer resolution over the full 1 keV to 30 keV energy range
  • Easy access to beam landing energies as low as 20 eV
  • Excellent stability with piezo stage as standard
Thermo Scientific Quattro E scanning electron microscope (SEM)
Quattro ESEM
  • Ultra-versatile high-resolution FEG SEM with unique environmental capability (ESEM)
  • Observe all information from all samples with simultaneous SE and BSE imaging in every mode of operation
Thermo Scientific Prisma E scanning electron microscope (SEM)
Prisma E SEM
  • Entry-level SEM with excellent image quality
  • Easy and quick sample loading and navigation for multiple samples
  • Compatible with a wide range of materials thanks to dedicated vacuum modes
Thermo Scientific Apreo 2 scanning electron microscope (SEM)
Apreo 2 SEM
  • High-performance SEM for all-round nanometer or sub-nanometer resolution
  • In-column T1 backscatter detector for sensitive, TV-rate materials contrast
  • Excellent performance at long working distance (10 mm)
Phenom ProX G6 Desktop SEM
  • High performance desktop SEM with integrated EDS detector
  • Resolution <6 nm (SE) and <8 nm (BSE); magnification up to 350,000x
  • Optional SE detector
Centrios HX Circuit Edit System
  • Cutting-edge circuit editing for sub 7nm advanced semiconductor
  • Unparalleled milling precision and control with low landing energy
  • Advanced navigation and ion beam placement accuracy
Centrios CE
  • High-performance circuit editing for 14-nm and above design rules
  • Superior milling precision and control
  • Advanced navigation and ion beam placement accuracy
MK.4TE ESD and Latch-Up Test System
  • Rapid-relay-based operations—up to 2304 channels
  • Advanced device preconditioning with six separate vector drive levels
  • Fully compliant Latch-Up stimulus and device biasing
ELITE System
  • Electrical defect localization
  • Thermal mapping
  • Advanced packaging analysis
nProber IV
  • Localize transistor and BEOL faults
  • Thermal nanoprobing (-40°C to 150°C)
  • Semi-automated operation
Meridian S System
  • Fault Diagnostic with Active Probe Technology
  • Static laser stimulation (SLS / OBIRCH) and photon emission options
  • Supports both micro-probing and probe card device stimulation
Meridian WS-DP System
  • High-sensitivity, low-noise, low-voltage photon emission detection with broadband DBX or InGaAs camera systems
  • Multi-wavelength laser scanning microscope for scan chain analysis, frequency mapping, transistor probing and isolation of faults
Meridian 7 System
  • Dynamic Optical Fault Isolation for 10nm node and below
  • High resolution visible and Infrared light
  • High-yield sample preparation to 5μm widely available
Meridian IV System
  • High sensitivity extended-wavelength DBX photon emission detection
  • Standard InGaAs photon emission detection
  • Laser Scanning Microscope with multiple wavelength options
Celestron Test System
  • Wafer and package level TLP testing
  • High current TLP pulse generator
  • Can be interfaced with semiautomatic probers
  • Intuitive software for control and report generation
Orion3 Test System
  • Charged device model testing
  • Dual high resolution color cameras
  • Test densities to less than 0.4mm pitch
Pegasus
  • Testing per the latest industry standards
  • True system level ESD 150pF/330Ω network
  • 2 pin connection via wafer probes to any device
AutoTEM 5
  • Fully automated in situ S/TEM sample preparation
  • Support of top-down, planar and inverted geometry
  • Highly configurable workflow
  • Easy to use, intuitive user interface
Thermo Scientific Auto Slice and View 4.0 serial section electron microscopy software
Auto Slice and View 4.0 Software
  • Automated serial sectioning for DualBeam
  • Multi-modal data acquisition (SEM, EDS, EBSD)
  • On-the-fly editing capabilities
  • Edge based cut placement
iFast Software
  • Macro recorder for faster recipe creating
  • Runner for unattended overnight operation
  • Alignment tools: Image recognition and edge finding
Thermo Scientific Inspect 3D tomography software
Inspect 3D Software
  • Image processing tools and filters for cross-correlation
  • Feature tracking for image alignment
  • Algebraic reconstruction technique for iterative projection comparison
NEXS Software
  • Auto-syncs position/magnification between NEXS and Circuit Edit system for a more seamless user experience
  • Connects to most Thermo Scientific tools used for EFA, PFA and Circuit Edit space
Style Sheet to change H2 style to p with em-h2-header class
Contact us
Style Sheet to change H2 style to p with em-h2-header class
Style Sheet for Support and Service footer
Style Sheet for Fonts
Style Sheet for Cards
Electron microscopy services for
semiconductors
To ensure optimal system performance, we provide you access to a world-class network of field service experts, technical support, and certified spare parts.
Learn more ›