protein concentrators of various sizes and the crossflow filtration device
Protein extraction, purification, and cleanup methods frequently produce dilute samples of target biomolecules in buffer matrices incompatible with downstream applications. Pierce protein concentrators are disposable ultrafiltration centrifugal devices used to concentrate, desalt, and buffer exchange biological samples, such as proteins and nucleic acids, readying them for downstream applications with minimal sample handling. Pierce protein concentrators are available in four sizes (0.5, 6, 20, and 100 mL) and offer pH compatibility over a broad pH range (1–9).
The Nanocon™ Crossflow Concentrator is an innovative solution for exosome, lipid nanoparticle (LNP), and virus enrichment. The 30 nM pore size PE membrane is perfect for nanosized particles, and it fits conveniently in a 250 mL centrifuge adapter.
For larger volume sizes, Pierce crossflow filtration cassettes are designed for high purity and recovery of peptides, proteins, and virus/viral particles. They are used to concentrate large supernatant volumes (0.5 L–6 L). Additional accessories are available for use with the crossflow filtration cassettes.
Choose the right protein concentrator
 Protein concentration
 microcentrifuge tube with concentrator
15 ml conical tube with concentrator
50 ml conical tube with concentrator
large tube with concentrator
Sample volume0.1 mL–0.5 mL2 mL–6 mL5 mL–20 mL20 mL–100 mL
See all Protein Concentrators, 3K or 5K MWCO *
See all Protein Concentrators, 10K MWCO
See all Protein Concentrators, 30K MWCO
See all Protein Concentrators, 50K MWCO
See all Protein Concentrators, 100K MWCO
*MWCO = molecular weight cutoff
Choose the right crossflow concentrator
 Nanocon Crossflow ConcentratorCrossflow Filtration Cassettes
Sample Volume40 mL0.5 L–6 L
 See Nanocon Crossflow ConcentratorSee all Crossflow Filtration Cassettes, 3-100 kDa MWCO
Recommended accessories for Pierce Crossflow Filtration Cassettes
Ordering information
Catalog # Name Size Price (USD) Qty
88512 Pierce™ Protein Concentrators PES, 3K MWCO, 0.5 mL, 25 Concentrators Each
130.65
Online Exclusive
148.00
Save 17.35 (12%)
88513 Pierce™ Protein Concentrators PES, 10K MWCO, 0.5 mL, 25 Concentrators Each
129.65
Online Exclusive
148.00
Save 18.35 (12%)
88502 Pierce™ Protein Concentrators PES, 30K MWCO, 0.5 mL, 25 Concentrators Each
128.65
Online Exclusive
148.00
Save 19.35 (13%)
88504 Pierce™ Protein Concentrators PES, 50K MWCO, 0.5 mL, 25 Concentrators Each
137.65
Online Exclusive
148.00
Save 10.35 (7%)
88503 Pierce™ Protein Concentrators PES, 100K MWCO, 0.5 mL, 25 Concentrators Each
133.65
Online Exclusive
148.00
Save 14.35 (10%)
Catalog # Name Size Price (USD) Qty
N003040 Nanocon™ Crossflow Concentrator, 30 nm, 40 mL Each
413.00
88514 Pierce™ Protein Concentrator PES, 3K MWCO, 2-6 mL, 10 Concentrators Each
83.65
Online Exclusive
94.50
Save 10.85 (11%)
88515 Pierce™ Protein Concentrator PES, 3K MWCO, 2-6 mL, 24 Concentrators Each
167.65
Online Exclusive
188.00
Save 20.35 (11%)
88516 Pierce™ Protein Concentrator PES, 10K MWCO, 2-6 mL, 10 Concentrators Each
84.65
Online Exclusive
94.50
Save 9.85 (10%)
88517 Pierce™ Protein Concentrator PES, 10K MWCO, 2-6 mL, 24 Concentrators Each
167.65
Online Exclusive
188.00
Save 20.35 (11%)
88521 Pierce™ Protein Concentrator PES, 30K MWCO, 2-6 mL, 10 Concentrators Each
84.65
Online Exclusive
94.50
Save 9.85 (10%)
88522 Pierce™ Protein Concentrator PES, 30K MWCO, 2-6 mL, 24 Concentrators Each
172.65
Online Exclusive
188.00
Save 15.35 (8%)
88538 Pierce™ Protein Concentrators PES, 50K MWCO, 2–6 mL, 10 Concentrators Each
88.65
Online Exclusive
94.50
Save 5.85 (6%)
88539 Pierce™ Protein Concentrators PES, 50K MWCO, 2–6 mL, 24 Concentrators Each
175.65
Online Exclusive
188.00
Save 12.35 (7%)
88523 Pierce™ Protein Concentrator PES, 100K MWCO, 2-6 mL, 10 Concentrators Each
85.65
Online Exclusive
94.50
Save 8.85 (9%)
88524 Pierce™ Protein Concentrator PES, 100K MWCO, 2-6 mL, 24 Concentrators Each
170.65
Online Exclusive
188.00
Save 17.35 (9%)
Catalog # Name Size Price (USD) Qty
88525 Pierce™ Protein Concentrator PES, 3K MWCO, 5-20 mL, 10 Concentrators Each
167.65
Online Exclusive
187.00
Save 19.35 (10%)
88527 Pierce™ Protein Concentrator PES, 10K MWCO, 5-20 mL, 10 Concentrators Each
165.65
Online Exclusive
187.00
Save 21.35 (11%)
88528 Pierce™ Protein Concentrator PES, 10K MWCO, 5-20 mL, 24 Concentrators Each
328.65
Online Exclusive
375.00
Save 46.35 (12%)
88529 Pierce™ Protein Concentrator PES, 30K MWCO, 5-20 mL, 10 Concentrators Each
166.65
Online Exclusive
187.00
Save 20.35 (11%)
88531 Pierce™ Protein Concentrator PES, 30K MWCO, 5-20 mL, 24 Concentrators Each
330.65
Online Exclusive
375.00
Save 44.35 (12%)
88540 Pierce™ Protein Concentrators PES, 50K MWCO, 5–20 mL, 10 Concentrators Each
159.65
Online Exclusive
187.00
Save 27.35 (15%)
88541 Pierce™ Protein Concentrators PES, 50K MWCO, 5–20 mL, 24 Concentrators Each
329.65
Online Exclusive
375.00
Save 45.35 (12%)
88532 Pierce™ Protein Concentrator PES, 100K MWCO, 5-20 mL, 10 Concentrators Each
170.65
Online Exclusive
187.00
Save 16.35 (9%)
88533 Pierce™ Protein Concentrator PES, 100K MWCO, 5-20 mL, 24 Concentrators Each
336.65
Online Exclusive
375.00
Save 38.35 (10%)
Catalog # Name Size Price (USD) Qty
88534 Pierce™ Protein Concentrator PES, 5K MWCO, 20-100 mL, 4 Concentrators Each
179.65
Online Exclusive
215.00
Save 35.35 (16%)
88535 Pierce™ Protein Concentrator PES, 10K MWCO, 20-100 mL, 4 Concentrators Each
182.65
Online Exclusive
215.00
Save 32.35 (15%)
88536 Pierce™ Protein Concentrator PES, 30K MWCO, 20-100 mL, 4 Concentrators Each
186.65
Online Exclusive
215.00
Save 28.35 (13%)
88542 Pierce™ Protein Concentrators PES, 50K MWCO, 20–100 mL, 4 Concentrators Each
196.65
Online Exclusive
218.00
Save 21.35 (10%)
88537 Pierce™ Protein Concentrator PES, 100K MWCO, 20-100 mL, 4 Concentrators Each
195.65
Online Exclusive
214.00
Save 18.35 (9%)
Product benefits
various sizes of protein concentrators
Protein concentrators
  • Fast—concentrate samples up to 10- to 50-fold in 5–30 minutes (times may vary with different MWCOs)
  • Flexible—suitable for sample volumes ranging from 100 μL to 100 mL, with a wide range of MWCO offerings
  • Efficient—recovery of >90% of target sample while removing contaminants or exchange buffers
  • Convenient—clear markings, wide sample chamber, and removable filtrate chamber make handling simple and easy
  • Instrument compatible—can be used with standard centrifuges with either fixed-angle or swinging-bucket rotors
packaging, tubing, cassette, and accessories of the crossflow filtration kit
Crossflow filtration cassettes
  • Scalable—concentrate sample volumes ranging from 0.5–6 L
  • Fast—many concentration and protein recovery protocols can be completed in 30 or less minutes
  • Versatile—concentration cassettes are available in five different MWCOs and are compatible with buffers up to pH 13
  • Efficient—recovery of >99% protein (with 25 mL buffer rinse)
How-to-videos for each protein concentrator size
Video Player is loading.
Current Time 0:00
Duration 2:56
Loaded: 0.00%
Stream Type LIVE
Remaining Time 2:56
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    0.1–0.5 mL concentrators
    Watch how to concentrate protein solution for sample volumes up to 0.5 mL using the microcentrifuge tube format. Membranes are available with 3K, 10K, 30K, 50K, and 100K MWCO.
    Video Player is loading.
    Current Time 0:00
    Duration 2:52
    Loaded: 0.00%
    Stream Type LIVE
    Remaining Time 2:52
    1x
      • Chapters
      • descriptions off, selected
      • captions off, selected
      • en (Main), selected
      2–6 mL concentrators
      Watch how to concentrate protein solution for sample volumes up to 6 mL using the 15 mL centrifuge tube format. Membranes are available with 3K, 10K, 30K, 50K, or 100K MWCO.
      Video Player is loading.
      Current Time 0:00
      Duration 0:00
      Loaded: 0%
      Stream Type LIVE
      Remaining Time 0:00
      1x
        • Chapters
        • descriptions off, selected
        • captions off, selected
          5–20 mL concentrators
          Watch how to concentrate protein solution for sample volumes up to 20 mL using the 20 mL tube format. Membranes are available with 3K, 10K, 30K, 50K, and 100K MWCO.
          Video Player is loading.
          Current Time 0:00
          Duration 2:57
          Loaded: 0.00%
          Stream Type LIVE
          Remaining Time 2:57
          1x
            • Chapters
            • descriptions off, selected
            • captions off, selected
            • en (Main), selected
            20–100 mL concentrators
            Watch how to concentrate protein solution for sample volumes up to 100 mL using the 250 mL centrifuge bottle format. Membranes are available with 5K, 10K, 30K, 50K, and 100K MWCO.
            Video Player is loading.
            Current Time 0:00
            Duration 3:00
            Loaded: 0.00%
            Stream Type LIVE
            Remaining Time 3:00
            1x
              • Chapters
              • descriptions off, selected
              • captions off, selected
              • en (Main), selected
              0.5–6 L concentrators
              Watch how to concentrate protein solution for sample volumes up to 6 liters using the Crossflow Filtration Cassette. These tangential flow filtration (TFF) cassettes are available in 3K, 10K, 30K, 50K, and 100K MWCO.
              For Research Use Only. Not for use in diagnostic procedures.