Key benefits of upgrading Helios G4 to Helios 5
  • Eliminate slow, tedious, manual processes with SmartAlign Technology, saving up to 4 hours per week
  • Simplify the image tuning procedure with Flash technology and enhance the user experience
  • Thermo Scientific AutoTEM Software enables high-quality S/TEM sample preparation for users of any experience level in less than one hour
  • Faster performance with new Windows 10 microscope PC
  • DualBeam IT infrastructure suited to latest updates
  • Improved cybersecurity and integration to MS OneDrive
Additional upgrades for Helios DualBeam (FIB-SEM)
Femtosecond laser ablation
  • Unprecedented material removal rates with large-volume collection and investigation of large grain size
  • Ideal solution for traditionally difficult-to-section materials like graphite, carbon, and refectory material
TOF-SIMS Detector
  • Detection and mapping of all elements of the periodic table, including light elements such as hydrogen, lithium, boron, and carbon in difficult samples such as low-carbon steels
  • High-sensitivity elemental analysis capable of detecting concentrations on the parts-per-million (ppm) level
AutoScript: Python Scripting environment
  • Powerful Python scripting (API) for DualBeams, allowing repetitive work to be done faster, more accurately, and with fewer mistakes
Correlative Electron Microscopy Software: MAPS
  • Completely automated solution for generating high-quality, multi-scale image of large overview while preserving the context of observations
  • Correlation of images from different microscopes, including imported images from third-party instruments using the integrated Bio-Formats Library
Auto Slice & View
  • Automated acquisition of high-resolution 3D images and analytical maps such as EBSD (electron backscatter diffraction) or EDS (energy dispersive X-ray spectroscopy)
Avizo Software
  • Advanced image processing, quantification, analysis, and reporting
  • 2D and 3D materials characterization
Helios PFIB DualBeam (FIB-SEM) upgrades
Key benefits of upgrading Verios G4 to Verios G5
Key benefits of upgrading Helios G4 PFIB to Helios PFIB 5
  • Eliminate slow, tedious, manual processes with SmartAlign Technology, saving  up to 4 hours per week
  • Simplify the image tuning procedure with Flash technology and enhance the user experience
  • Thermo Scientific AutoTEM Software enables high-quality S/TEM sample preparation for users of any experience level in less than one hour
  • Faster performance with new Windows 10 microscope PC
  • DualBeam IT infrastructure suited to latest updates
  • Improved cybersecurity and integration to MS OneDrive
  • Contamination-free TEM specimen preparation with Extended PFIB voltage range to 500 V
Additional upgrades for Helios PFIB DualBeam (FIB-SEM)
Femtosecond laser ablation
  • Unprecedented material removal rates with large-volume collection and investigation of large grain size
  • Ideal solution for traditionally difficult-to-section materials like graphite, carbon, and refectory material
TOF-SIMS Detector
  • Detection and mapping of all elements of the periodic table, including light elements such as hydrogen, lithium, boron, and carbon in difficult samples such as low-carbon steels
  • High-sensitivity elemental analysis capable of detecting concentrations on the parts-per-million (ppm) level
AutoScript: Python Scripting environment
  • Powerful Python scripting (API) for DualBeams, allowing repetitive work to be done faster, more accurately, and with fewer mistakes
Correlative Electron Microscopy Software: MAPS
  • Completely automated solution for generating high-quality, multi-scale image of large overview while preserving the context of observations
  • Correlation of images from different microscopes, including imported images from third-party instruments using the integrated Bio-Formats Library
Auto Slice & View
  • Automated acquisition of high-resolution 3D images and analytical maps such as EBSD (electron backscatter diffraction) or EDS (energy dispersive X-ray spectroscopy)
Avizo Software
  • Advanced image processing, quantification, analysis, and reporting
  • 2D and 3D materials characterization
Scios 2 DualBeam (FIB-SEM) upgrades
Key benefits of Quattro software upgrade (Windows 10)
Key benefits of Scios 2 software upgrade (Windows 10)
  • Thermo Scientific AutoTEM Software enables high-quality S/TEM sample preparation for users of any experience level in less than one hour
  • Faster performance with new Windows 10 microscope PC
  • DualBeam IT infrastructure suited to latest updates
  • Improved cybersecurity and integration to MS OneDrive
Additional upgrades for Scios DualBeam (FIB-SEM)
TOF-SIMS Detector
  • Detection and mapping of all elements of the periodic table, including light elements such as hydrogen, lithium, boron, and carbon in difficult samples such as low-carbon steels
  • High-sensitivity elemental analysis capable of detecting concentrations on the parts-per-million (ppm) level
AutoScript: Python Scripting environment
  • Powerful Python scripting (API) for DualBeams, allowing repetitive work to be done faster, more accurately, and with fewer mistakes
Correlative Electron Microscopy Software: MAPS
  • Completely automated solution for generating high-quality, multi-scale image of large overview while preserving the context of observations
  • Correlation of images from different microscopes, including imported images from third-party instruments using the integrated Bio-Formats Library
Auto Slice & View
  • Automated acquisition of high-resolution 3D images and analytical maps such as EBSD (electron backscatter diffraction) or EDS (energy dispersive X-ray spectroscopy)
Avizo Software
  • Advanced image processing, quantification, analysis, and reporting
  • 2D and 3D materials characterization
Accessories
Easylift
Easylift
  • Enables precise, site-specific preparation of ultra-thin TEM lamellae
  • Promotes operator confidence for in situ TEM sample lift-outs—critical for one-of-a-kind samples
  • Pairs with iFAST software for consistent, repeatable preparation and lift-out of ultra-thin TEM samples
  • Allows simple “click and drag” movement due to EasyLift’s full integration with Dualbeam xT UI
  • Supported by the Thermo Fisher Scientific expert applications knowledge in TEM lamella prep solutions
 Download datasheet
µHeater
µHeater
  • Ultra-fast heating solution for in situ high resolution imaging
  • Fully integrated
  • Temperatures up to 1200 °C
CleanConnect Sample Transfer System
CleanConnect Sample Transfer System
  • Sample integrity is preserved by the use of inert gas sample transfer
  • Ergonomic and modular design enables uncomplicated sample handling
  • Seamless, automatic integration to a variety of Thermo Scientific SEM/DualBeams
  • Compact design requiring no changes in system footprint or microscope specifications
  • Compatible with most glove box systems and available with optional direct glove box connectivity
 Download datasheet
QuickLoader
QuickLoader
  • Load lock for fast and easy manual transfer of samples
  • Sample cycle: loading sample, pump down till first image and unloading ≤ 2 minutes
  • Fully interlocked for safe and intuitive operation
  • Compact design requiring no changes in system footprint or microscope specifications
  • Field retrofit possible for most Thermo Scientific SEM/DualBeam microscopes
 Download datasheet
Style Sheet for Komodo Tabs
Style Sheet for Global Design System
Contact us
Style Sheet for Support and Service footer
Style Sheet for Fonts
Style Sheet for Cards