Antibody Isotyping and Characterization
Identify & quantify immunoglobulin isotypes
Immunoglobulin (IgG, IgA, IgE, IgM) isotyping and quantitation are important screening and testing steps in rat and mouse monoclonal antibody development and production. Immune responses in ascites fluid must be evaluated and the immunoglobulin class and subclass identities of screened and isolated antibody-producing hybridomas must be determined.
Several different kinds of assay kits are available for antibody isotyping, depending on the needed scale, desired throughput, and available detection equipment. These include ELISA kits, multiplex assays for Luminex systems, rapid test cassettes, and microagglutination titer assays.
ELISA antibody isotyping kits
Invitrogen isotyping ELISA kits are a fast, sensitive and convenient method for quantitation of immunoglobulin isotypes during and after hybridoma development, to assess immune response. These ELISA provide an accurate quantitative result and enable testing of multiple samples at a time.
For additional information about these isotype kits, as well as many single-isotype immunoglobulin ELISA kits for antibody quantitation, see the related page about Immunoglobulin ELISA Kits.
Multiplex antibody isotyping panels for Luminex systems
Invitrogen ProcartaPlex antibody isotyping panels enable quantitative, multiplexed measurement of individual mouse or rat immunoglobulin isotypes from serum, plasma, and cell culture supernatant samples using Luminex magnetic bead technology. Learn more about ProcartaPlex Multiplex Assays.
Dipstick-style rapid antibody isotyping kits
The Pro-Detect Rapid Antibody Isotyping Kit is a single-use, dipstick lateral flow kit for quick, easy determination of the immunoglobulin class, subclass, and light chain type of a mouse, human, or rat antibody. This antibody isotyping assay uses membrane-based strips coated with dye-bound conjugated antibodies that provide a visual, color readout of the monoclonal antibody isotype within ten minutes.
Cassette-style rapid antibody isotyping kits
Thermo Scientific Pierce rapid antibody isotyping kits are single-use, cassette-based kits for 5-minute lateral-flow assay determination of monoclonal antibody class and subclass identity. These antibody isotyping kits use cartridges that provide a color-readout of the monoclonal antibody isotype within five minutes after pipetting a small amount of diluted culture supernatant or ascites fluid sample into the sample well.
Catalog # Name Size Price (USD) Qty
26178 Pierce™ Rapid Antibody Isotyping Kit - Mouse Each
395.00
26179 Pierce™ Rapid Antibody Isotyping Kit plus Kappa and Lambda - Mouse Each
321.65
Online Exclusive
428.00
Save 106.35 (25%)
Easy-Titer antibody quantitation assay kits
Thermo Scientific Easy-Titer antibody assay kits include antibody-sensitized microspheres to measure the specific concentration of antibodies by an easy and rapid microagglutination technique using standard microplates and UV-Vis plate reader (spectrophotometer). The kits are specific for the target immunoglobulin and, unlike total protein assays, can specifically measure the concentration of target antibody in samples (e.g., serum, plasma, culture supernatant) that contain other proteins.
Catalog # Name Size Price (USD) Qty
23300 Easy-Titer™ Mouse IgG Assay Kit Each
572.00
23305 Easy-Titer™ Rabbit IgG Assay Kit Each
594.00
23310 Easy-Titer™ Human IgG (H+L) Assay Kit Each
465.65
Online Exclusive
620.00
Save 154.35 (25%)
23325 Easy-Titer™ Human IgG (gamma chain) Assay Kit Each
504.65
Online Exclusive
672.00
Save 167.35 (25%)
23315 Easy-Titer™ Human IgM Assay Kit Each
662.00
Bulk Quantities and Custom Products
Partner with us to scale-up your process or product development pipeline with bulk quantities of high-quality immunoassay reagents and biochemicals. We offer a variety of compounds, devices, and resins for your large-scale applications.
Complete request form
For Research Use Only. Not for use in diagnostic procedures.