Transfer Buffers for Western Blotting
Premixed transfer buffers are a convenient way to ensure high-quality, consistent gel transfer. We offer premixed western blot transfer buffers that are optimized for each gel chemistry. Use the tabs below to see the recommended transfer buffers and transfer conditions for the gel chemistry being used for western blot wet or semi-dry transfer. Find the suggested transfer buffer recipes for wet transfer below.
Transfer membranesTransfer systems
Recommended transfer buffers for Bis-Tris and Tris-Acetate Systems
Protein sampleGel chemistryTransfer buffer
Broad-range MW (6-400 kDa)Bolt Bis-Tris PlusBolt Bis-Tris transfer buffer
Reduced samples: Antioxidant
Broad-range MW (6-400 kDa)NuPAGE Bis-TrisNuPAGE Bis-Tris transfer buffer
Reduced samples: Antioxidant
High MW (40-500 kDa)NuPAGE Tris-Acetate
Transfer conditions using the Mini Bolt Module (Mini Gel Tank)
Gel typeMembraneTransfer bufferTransfer conditionsExpected current*Run time
Bolt Bis-Tris PlusNitrocellulose
Bis-Tris transfer buffer with 10% methanol and antioxidant for reduced samples
10 V constantStart: 160 mA
End: 60 mA
1 hr
PVDF20 V constantStart: 340 mA
End: 130 mA
1 hr
NuPAGE Bis-TrisNitrocellulose10 V constantStart: 160 mA
End: 60 mA
1 hr
PVDF20 V constantStart: 390 mA
End: 130 mA
1 hr
NuPAGE Tris-AcetateNitrocellulose10 V constantStart: 150 mA
End: 50 mA
1 hr
PVDF20 V constantStart: 380 mA
End: 130 mA
1 hr
* Current readings represent values when running a single gel, and can vary depending upon the power supply being used.
Transfer conditions using the XCell II Blot Module (SureLock Tank)
Gel typeMembraneTransfer buffer*Transfer conditionsExpected current**Run time
Bolt Bis-Tris Plus and NuPAGE Bis-TrisNitrocellulose or PVDFBis-Tris transfer buffer with 10% methanol and antioxidant for reduced samples30 V constantStart: 170 mA
End: 100 mA
1 hr
NuPAGE Tris-AcetateNitrocellulose or PVDFBis-Tris transfer buffer with 10% methanol and antioxidant for reduced samples30 V constantStart: 220 mA
End: 180 mA
1 hr
* Transfer buffer with 10% methanol provides optimal transfer for a single gel in the blot module. If you are transferring 2 gels, increase the methanol content to 20% to ensure efficient transfer.
** Current readings represent values when running a single gel, and can vary depending upon the power supply being used.
Bolt Bis-Tris Plus System
Catalog # Name Size Price (USD) Qty
BT0005 Bolt™ Antioxidant Each
40.65
Online Exclusive
42.18
Save 1.53 (4%)
BT00061 Bolt™ Transfer Buffer (20X), 1L Each
235.65
Online Exclusive
239.00
Save 3.35 (1%)
BT0006 Bolt™ Transfer Buffer (20X), 125mL Each
35.65
Online Exclusive
35.88
NuPAGE System
Catalog # Name Size Price (USD) Qty
NP0005 NuPAGE™ Antioxidant Each
41.30
NP0050 NuPAGE™ MOPS SDS Buffer Kit (for Bis-Tris Gels) Each
133.65
Online Exclusive
139.00
Save 5.35 (4%)
NP00061 NuPAGE™ Transfer Buffer (20X), 1L Each
234.00
NP0006 NuPAGE™ Transfer Buffer (20X), 125mL Each
35.49
Recommended transfer buffers for Tris-Glycine and Tricine Systems
Protein sampleGel chemistryTransfer buffer
Broad-range MW (6-400 kDa)Novex Tris-GlycineTris-Glycine transfer buffer
Low MW (2.5-40 kDa)Novex Tricine
Transfer conditions using the Mini Bolt Module (Mini Gel Tank)
Gel typeMembraneTransfer bufferTransfer conditionsExpected current*Run time
Novex Tris-GlycineNitrocelluloseTris Glycine Transfer Buffer with 20% methanol10 V constantStart: 70 mA
End: 50 mA
1 hr
PVDF20 V constantStart: 160 mA
End: 100 mA
1 hr
Novex TricineNitrocellulose10 V constantStart: 70 mA
End: 60 mA
1 hr
PVDF20 V constantStart: 180 mA
End: 150 mA
1 hr
* Current readings represent values when running a single gel, and can vary depending upon the power supply being used.
Transfer conditions using the XCell II Blot Module (SureLock Tank)
Gel typeMembraneTransfer bufferTransfer conditionsExpected current*Run time
Novex Tris-Glycine and Novex TricineNitrocellulose or PVDFTris Glycine Transfer Buffer with 20% methanol.
1X Transfer Buffer should be pH 8.3 before addition of SDS or methanol. Do not adjust the pH.
25 V constantStart: 100 mA
1-2 hr
* Current readings represent values when running a single gel, and can vary depending upon the power supply being used.
Tris-Glycine and Tricine Systems
Catalog # Name Size Price (USD) Qty
LC3675 Tris-Glycine Transfer Buffer (25X) Each
55.25
28380 BupH™ Tris-Glycine Buffer Packs, 40 Packs Each
170.65
Online Exclusive
192.00
Save 21.35 (11%)
Recommended transfer buffers for Semi-dry Transfer
1-Step Transfer buffer is a high ionic strength formulation which allows for 5-minute to 12-minute protein transfer when used with compatible semi-dry blotting systems. 1-Step Transfer Buffer is compatible with Power Blotter and other protein semi-dry transfer devices, when they are paired with a suitable high-current power supply. Such devices should provide constant high current (1.3 to 5.0 amps) to rapidly transfer proteins via the high ionic strength conditions supplied by the transfer buffer.
Transfer conditions using the Power Blotter System
Gel SizeSurface area (cm2)Constant current (A)Voltage limit (V)Recommended transfer time (min)*
Low MWMixed MWHigh MW
1 mini gel~601.3255710
2 mini gels or 1 midi gel~1202.5255710
3 mini gels~1803.8255710
4 mini gels or 2 midi gels~2405.0255710
* For gels thicker than 1 mm or for homemade gel formations, add 2 minutes of transfer time.
Ordering information
Catalog # Name Size Price (USD) Qty
PB7300 Power Blotter 1-Step™ Transfer Buffer (5X), 1 L Each
337.65
Online Exclusive
352.00
Save 14.35 (4%)
84731X5 Pierce™ 1-Step Transfer Buffer, 5 x 1 L Each
377.65
Online Exclusive
402.00
Save 24.35 (6%)
PB7100 Power Blotter 1-Step™ Transfer Buffer (5X), 250 mL Each
106.65
Online Exclusive
111.00
Save 4.35 (4%)
84731 Pierce™ 1-Step Transfer Buffer, 1 L Each
93.65
Online Exclusive
99.25
Save 5.60 (6%)
Ordering information
Catalog # Name Size Price (USD) Qty
LC3675 Tris-Glycine Transfer Buffer (25X) Each
55.25
BT0005 Bolt™ Antioxidant Each
40.65
Online Exclusive
42.18
Save 1.53 (4%)
84731X5 Pierce™ 1-Step Transfer Buffer, 5 x 1 L Each
377.65
Online Exclusive
402.00
Save 24.35 (6%)
NP0050 NuPAGE™ MOPS SDS Buffer Kit (for Bis-Tris Gels) Each
133.65
Online Exclusive
139.00
Save 5.35 (4%)
NP00061 NuPAGE™ Transfer Buffer (20X), 1L Each
234.00
BT0006 Bolt™ Transfer Buffer (20X), 125mL Each
35.65
Online Exclusive
35.88
NP0005 NuPAGE™ Antioxidant Each
41.30
PB7300 Power Blotter 1-Step™ Transfer Buffer (5X), 1 L Each
337.65
Online Exclusive
352.00
Save 14.35 (4%)
PB7100 Power Blotter 1-Step™ Transfer Buffer (5X), 250 mL Each
106.65
Online Exclusive
111.00
Save 4.35 (4%)
BT00061 Bolt™ Transfer Buffer (20X), 1L Each
235.65
Online Exclusive
239.00
Save 3.35 (1%)
84731 Pierce™ 1-Step Transfer Buffer, 1 L Each
93.65
Online Exclusive
99.25
Save 5.60 (6%)
NP0006 NuPAGE™ Transfer Buffer (20X), 125mL Each
35.49
28380 BupH™ Tris-Glycine Buffer Packs, 40 Packs Each
170.65
Online Exclusive
192.00
Save 21.35 (11%)
Transfer buffer formulations
The following transfer buffer recipes are provided to allow preparation of buffers from scratch. The pH listed for each buffer is for the 1X solution. Do not use acid or base to adjust the pH.
25X Tris-Glycine transfer buffer
  1. Dissolve the following reagents in 400 mL ultrapure water
    ReagentAmountConcentration (1X)
    Tris Base18.2 g12 mM
    Glycine90 g96 mM
  2. Mix well and adjust volume to 500 mL with ultrapure water.
  3. Before western transfer, dilute to 1X with water.
  4. Buffer is stable for 6 months at room temperature.
20X Bis-Tris Transfer buffer
  1. Dissolve the following reagents in 100 mL ultrapure water
    ReagentAmountConcentration (1X)
    Bicine10.2 g25 mM
    Bis-Tris (free base)13.1 g25 mM
    EDTA0.75 g1 mL
  2. Mix well and adjust volume to 125 mL with ultrapure water.
  3. Before western transfer, dilute to 1X with water.
  4. Buffer is stable for 6 months when stored at 4°C.
Recommended transfer buffers for Bis-Tris and Tris-Acetate Systems
Protein sampleGel chemistryTransfer buffer
Broad-range MW (6-400 kDa)Bolt Bis-Tris PlusBolt Bis-Tris transfer buffer
Reduced samples: Antioxidant
Broad-range MW (6-400 kDa)NuPAGE Bis-TrisNuPAGE Bis-Tris transfer buffer
Reduced samples: Antioxidant
High MW (40-500 kDa)NuPAGE Tris-Acetate
Transfer conditions using the Mini Bolt Module (Mini Gel Tank)
Gel typeMembraneTransfer bufferTransfer conditionsExpected current*Run time
Bolt Bis-Tris PlusNitrocellulose
Bis-Tris transfer buffer with 10% methanol and antioxidant for reduced samples
10 V constantStart: 160 mA
End: 60 mA
1 hr
PVDF20 V constantStart: 340 mA
End: 130 mA
1 hr
NuPAGE Bis-TrisNitrocellulose10 V constantStart: 160 mA
End: 60 mA
1 hr
PVDF20 V constantStart: 390 mA
End: 130 mA
1 hr
NuPAGE Tris-AcetateNitrocellulose10 V constantStart: 150 mA
End: 50 mA
1 hr
PVDF20 V constantStart: 380 mA
End: 130 mA
1 hr
* Current readings represent values when running a single gel, and can vary depending upon the power supply being used.
Transfer conditions using the XCell II Blot Module (SureLock Tank)
Gel typeMembraneTransfer buffer*Transfer conditionsExpected current**Run time
Bolt Bis-Tris Plus and NuPAGE Bis-TrisNitrocellulose or PVDFBis-Tris transfer buffer with 10% methanol and antioxidant for reduced samples30 V constantStart: 170 mA
End: 100 mA
1 hr
NuPAGE Tris-AcetateNitrocellulose or PVDFBis-Tris transfer buffer with 10% methanol and antioxidant for reduced samples30 V constantStart: 220 mA
End: 180 mA
1 hr
* Transfer buffer with 10% methanol provides optimal transfer for a single gel in the blot module. If you are transferring 2 gels, increase the methanol content to 20% to ensure efficient transfer.
** Current readings represent values when running a single gel, and can vary depending upon the power supply being used.
Bolt Bis-Tris Plus System
Catalog # Name Size Price (USD) Qty
BT0005 Bolt™ Antioxidant Each
40.65
Online Exclusive
42.18
Save 1.53 (4%)
BT00061 Bolt™ Transfer Buffer (20X), 1L Each
235.65
Online Exclusive
239.00
Save 3.35 (1%)
BT0006 Bolt™ Transfer Buffer (20X), 125mL Each
35.65
Online Exclusive
35.88
NuPAGE System
Catalog # Name Size Price (USD) Qty
NP0005 NuPAGE™ Antioxidant Each
41.30
NP0050 NuPAGE™ MOPS SDS Buffer Kit (for Bis-Tris Gels) Each
133.65
Online Exclusive
139.00
Save 5.35 (4%)
NP00061 NuPAGE™ Transfer Buffer (20X), 1L Each
234.00
NP0006 NuPAGE™ Transfer Buffer (20X), 125mL Each
35.49
Recommended transfer buffers for Tris-Glycine and Tricine Systems
Protein sampleGel chemistryTransfer buffer
Broad-range MW (6-400 kDa)Novex Tris-GlycineTris-Glycine transfer buffer
Low MW (2.5-40 kDa)Novex Tricine
Transfer conditions using the Mini Bolt Module (Mini Gel Tank)
Gel typeMembraneTransfer bufferTransfer conditionsExpected current*Run time
Novex Tris-GlycineNitrocelluloseTris Glycine Transfer Buffer with 20% methanol10 V constantStart: 70 mA
End: 50 mA
1 hr
PVDF20 V constantStart: 160 mA
End: 100 mA
1 hr
Novex TricineNitrocellulose10 V constantStart: 70 mA
End: 60 mA
1 hr
PVDF20 V constantStart: 180 mA
End: 150 mA
1 hr
* Current readings represent values when running a single gel, and can vary depending upon the power supply being used.
Transfer conditions using the XCell II Blot Module (SureLock Tank)
Gel typeMembraneTransfer bufferTransfer conditionsExpected current*Run time
Novex Tris-Glycine and Novex TricineNitrocellulose or PVDFTris Glycine Transfer Buffer with 20% methanol.
1X Transfer Buffer should be pH 8.3 before addition of SDS or methanol. Do not adjust the pH.
25 V constantStart: 100 mA
1-2 hr
* Current readings represent values when running a single gel, and can vary depending upon the power supply being used.
Tris-Glycine and Tricine Systems
Catalog # Name Size Price (USD) Qty
LC3675 Tris-Glycine Transfer Buffer (25X) Each
55.25
28380 BupH™ Tris-Glycine Buffer Packs, 40 Packs Each
170.65
Online Exclusive
192.00
Save 21.35 (11%)
Recommended transfer buffers for Semi-dry Transfer
1-Step Transfer buffer is a high ionic strength formulation which allows for 5-minute to 12-minute protein transfer when used with compatible semi-dry blotting systems. 1-Step Transfer Buffer is compatible with Power Blotter and other protein semi-dry transfer devices, when they are paired with a suitable high-current power supply. Such devices should provide constant high current (1.3 to 5.0 amps) to rapidly transfer proteins via the high ionic strength conditions supplied by the transfer buffer.
Transfer conditions using the Power Blotter System
Gel SizeSurface area (cm2)Constant current (A)Voltage limit (V)Recommended transfer time (min)*
Low MWMixed MWHigh MW
1 mini gel~601.3255710
2 mini gels or 1 midi gel~1202.5255710
3 mini gels~1803.8255710
4 mini gels or 2 midi gels~2405.0255710
* For gels thicker than 1 mm or for homemade gel formations, add 2 minutes of transfer time.
Ordering information
Catalog # Name Size Price (USD) Qty
PB7300 Power Blotter 1-Step™ Transfer Buffer (5X), 1 L Each
337.65
Online Exclusive
352.00
Save 14.35 (4%)
84731X5 Pierce™ 1-Step Transfer Buffer, 5 x 1 L Each
377.65
Online Exclusive
402.00
Save 24.35 (6%)
PB7100 Power Blotter 1-Step™ Transfer Buffer (5X), 250 mL Each
106.65
Online Exclusive
111.00
Save 4.35 (4%)
84731 Pierce™ 1-Step Transfer Buffer, 1 L Each
93.65
Online Exclusive
99.25
Save 5.60 (6%)
Ordering information
Catalog # Name Size Price (USD) Qty
LC3675 Tris-Glycine Transfer Buffer (25X) Each
55.25
BT0005 Bolt™ Antioxidant Each
40.65
Online Exclusive
42.18
Save 1.53 (4%)
84731X5 Pierce™ 1-Step Transfer Buffer, 5 x 1 L Each
377.65
Online Exclusive
402.00
Save 24.35 (6%)
NP0050 NuPAGE™ MOPS SDS Buffer Kit (for Bis-Tris Gels) Each
133.65
Online Exclusive
139.00
Save 5.35 (4%)
NP00061 NuPAGE™ Transfer Buffer (20X), 1L Each
234.00
BT0006 Bolt™ Transfer Buffer (20X), 125mL Each
35.65
Online Exclusive
35.88
NP0005 NuPAGE™ Antioxidant Each
41.30
PB7300 Power Blotter 1-Step™ Transfer Buffer (5X), 1 L Each
337.65
Online Exclusive
352.00
Save 14.35 (4%)
PB7100 Power Blotter 1-Step™ Transfer Buffer (5X), 250 mL Each
106.65
Online Exclusive
111.00
Save 4.35 (4%)
BT00061 Bolt™ Transfer Buffer (20X), 1L Each
235.65
Online Exclusive
239.00
Save 3.35 (1%)
84731 Pierce™ 1-Step Transfer Buffer, 1 L Each
93.65
Online Exclusive
99.25
Save 5.60 (6%)
NP0006 NuPAGE™ Transfer Buffer (20X), 125mL Each
35.49
28380 BupH™ Tris-Glycine Buffer Packs, 40 Packs Each
170.65
Online Exclusive
192.00
Save 21.35 (11%)
Transfer buffer formulations
The following transfer buffer recipes are provided to allow preparation of buffers from scratch. The pH listed for each buffer is for the 1X solution. Do not use acid or base to adjust the pH.
25X Tris-Glycine transfer buffer
  1. Dissolve the following reagents in 400 mL ultrapure water
    ReagentAmountConcentration (1X)
    Tris Base18.2 g12 mM
    Glycine90 g96 mM
  2. Mix well and adjust volume to 500 mL with ultrapure water.
  3. Before western transfer, dilute to 1X with water.
  4. Buffer is stable for 6 months at room temperature.
20X Bis-Tris Transfer buffer
  1. Dissolve the following reagents in 100 mL ultrapure water
    ReagentAmountConcentration (1X)
    Bicine10.2 g25 mM
    Bis-Tris (free base)13.1 g25 mM
    EDTA0.75 g1 mL
  2. Mix well and adjust volume to 125 mL with ultrapure water.
  3. Before western transfer, dilute to 1X with water.
  4. Buffer is stable for 6 months when stored at 4°C.
For Research Use Only. Not for use in diagnostic procedures.