de69 - Techniques/Samples/Products/Resources/Contact Us
With growing complexity and ever higher standards of reliability, quality control and assurance of manufactured parts have become increasingly more vital in modern industry. A critical aspect of quality regulation is failure analysis, which provides insight into the root cause of the component/material failure, establishing metrics for quality control during the manufacturing process, and enforcing 3rd-party quality requirements. Since component failure is often a direct result of numerous small, microscopic defects, their multi-scale observation and quantification is the only way to obtain the accurate characterization needed for root cause determination.
Thermo Fisher Scientific offers a range of tools for monitoring consistency through the holistic identification and study of defects, faults, and failures. For the analysis of large volumes, X-ray microtomography (microCT) is a non-destructive technique that generates 3D reconstructions of samples with micrometer resolution. Essentially identical to familiar hospital CAT scan technology, microCT provides a practical overview of the material for defect localization and isolation. These can subsequently be extracted and analyzed further by higher-resolution techniques, such as electron microscopy (EM).
With EM, structural details down to the nanometer scale become available. This allows for precise characterization of microscopic defects or nanoscale deviations from process specifications that could not be observed with other tools. With this information in hand, engineers and researchers can enact quality improvements in the earliest stages of defect formation.
Not only does electron microscopy offer unparalleled structural detail, but the technique also has the added benefit of elemental analysis. This method, called energy-dispersive X-ray spectroscopy (EDS, EDX, or XEDS), is made possible by the X-rays emitted from the sample surface during electron bombardment. The X-ray spectra are characteristic of the material they originate from, whereas the intensity of the peaks corresponds to concentration. This signal can be linked to a position on the EM image for elemental insight of observed defects. In fact, Thermo Scientific ChemiSEM Technology even features live EDX analysis, which automatically colors greyscale electron micrographs, granting instant chemical context for observed faults and defects.
Style Sheet for Komodo Tabs
Resources
Style Sheet to change H2 style to p with em-h2-header class
Samples
Battery Research
Battery development is enabled by multi-scale analysis with microCT, SEM and TEM, Raman spectroscopy, XPS, and digital 3D visualization and analysis. Learn how this approach provides the structural and chemical information needed to build better batteries.
Learn more ›
Nanoparticles
Materials have fundamentally different properties at the nanoscale than at the macroscale. To study them, S/TEM instrumentation can be combined with energy dispersive X-ray spectroscopy to obtain nanometer, or even sub-nanometer, resolution data.
Learn more ›
Metals Research and Characterization
Effective production of metals requires precise control of inclusions and precipitates. Our automated tools can perform a variety of tasks critical for metal analysis including; nanoparticle counting, EDS chemical analysis and TEM sample preparation.
Learn more ›
Catalysis Research
Catalysts are critical for a majority of modern industrial processes. Their efficiency depends on the microscopic composition and morphology of the catalytic particles; EM with EDS is ideally suited for studying these properties.
Learn more ›
Polymers Research
Polymer microstructure dictates the material’s bulk characteristics and performance. Electron microscopy enables comprehensive microscale analysis of polymer morphology and composition for R&D and quality control applications.
Learn more ›
2D Materials
Novel materials research is increasingly interested in the structure of low-dimensional materials. Scanning transmission electron microscopy with probe correction and monochromation allows for high-resolution two-dimensional materials imaging.
Learn more ›
Automotive Materials Testing
Every component in a modern vehicle is designed for safety, efficiency, and performance. Detailed characterization of automotive materials with electron microscopy and spectroscopy informs critical process decisions, product improvements, and new materials.
Learn more ›
Techniques
(S)TEM Sample Preparation
DualBeam microscopes enable the preparation of high-quality, ultra-thin samples for (S)TEM analysis. Thanks to advanced automation, users with any experience level can obtain expert-level results for a wide range of materials.
Learn more ›
3D Materials Characterization
Development of materials often requires multi-scale 3D characterization. DualBeam instruments enable serial sectioning of large volumes and subsequent SEM imaging at nanometer scale, which can be processed into high-quality 3D reconstructions of the sample.
Learn more ›
EDS Elemental Analysis
Thermo Scientific Phenom Elemental Mapping Software provides fast and reliable information on the distribution of chemical elements within a sample.
Learn more ›
3D EDS Tomography
Modern materials research is increasingly reliant on nanoscale analysis in three dimensions. 3D characterization, including compositional data for full chemical and structural context, is possible with 3D EM and energy dispersive X-ray spectroscopy.
Learn more ›
EDS Analysis with ChemiSEM Technology
Energy dispersive X-ray spectroscopy for materials characterization.
Learn more ›
Cross-sectioning
Cross sectioning provides extra insight by revealing sub-surface information. DualBeam instruments feature superior focused ion beam columns for high-quality cross sectioning. With automation, unattended high-throughput processing of samples is possible.
Learn more ›
In Situ experimentation
Direct, real-time observation of microstructural changes with electron microscopy is necessary to understand the underlying principles of dynamic processes such as recrystallization, grain growth, and phase transformation during heating, cooling, and wetting.
Learn more ›
Particle analysis
Particle analysis plays a vital role in nanomaterials research and quality control. The nanometer-scale resolution and superior imaging of electron microscopy can be combined with specialized software for rapid characterization of powders and particles.
Learn more ›
Cathodoluminescence
Cathodoluminescence (CL) describes the emission of light from a material when it is excited by an electron beam. This signal, captured by a specialized CL detector, carries information on the sample’s composition, crystal defects, or photonic properties.
Learn more ›
Multi-scale analysis
Novel materials must be analyzed at ever higher resolution while retaining the larger context of the sample. Multi-scale analysis allows for the correlation of various imaging tools and modalities such as X-ray microCT, DualBeam, Laser PFIB, SEM and TEM.
Learn more ›
Automated Particle Workflow
The Automated NanoParticle Workflow (APW) is a transmission electron microscope workflow for nanoparticle analysis, offering large area, high resolution imaging and data acquisition at the nanoscale, with on-the-fly processing.
Learn more ›
(S)TEM Sample Preparation
DualBeam microscopes enable the preparation of high-quality, ultra-thin samples for (S)TEM analysis. Thanks to advanced automation, users with any experience level can obtain expert-level results for a wide range of materials.
Learn more ›
3D Materials Characterization
Development of materials often requires multi-scale 3D characterization. DualBeam instruments enable serial sectioning of large volumes and subsequent SEM imaging at nanometer scale, which can be processed into high-quality 3D reconstructions of the sample.
Learn more ›
EDS Elemental Analysis
Thermo Scientific Phenom Elemental Mapping Software provides fast and reliable information on the distribution of chemical elements within a sample.
Learn more ›
3D EDS Tomography
Modern materials research is increasingly reliant on nanoscale analysis in three dimensions. 3D characterization, including compositional data for full chemical and structural context, is possible with 3D EM and energy dispersive X-ray spectroscopy.
Learn more ›
EDS Analysis with ChemiSEM Technology
Energy dispersive X-ray spectroscopy for materials characterization.
Learn more ›
Cross-sectioning
Cross sectioning provides extra insight by revealing sub-surface information. DualBeam instruments feature superior focused ion beam columns for high-quality cross sectioning. With automation, unattended high-throughput processing of samples is possible.
Learn more ›
In Situ experimentation
Direct, real-time observation of microstructural changes with electron microscopy is necessary to understand the underlying principles of dynamic processes such as recrystallization, grain growth, and phase transformation during heating, cooling, and wetting.
Learn more ›
Particle analysis
Particle analysis plays a vital role in nanomaterials research and quality control. The nanometer-scale resolution and superior imaging of electron microscopy can be combined with specialized software for rapid characterization of powders and particles.
Learn more ›
Cathodoluminescence
Cathodoluminescence (CL) describes the emission of light from a material when it is excited by an electron beam. This signal, captured by a specialized CL detector, carries information on the sample’s composition, crystal defects, or photonic properties.
Learn more ›
Multi-scale analysis
Novel materials must be analyzed at ever higher resolution while retaining the larger context of the sample. Multi-scale analysis allows for the correlation of various imaging tools and modalities such as X-ray microCT, DualBeam, Laser PFIB, SEM and TEM.
Learn more ›
Automated Particle Workflow
The Automated NanoParticle Workflow (APW) is a transmission electron microscope workflow for nanoparticle analysis, offering large area, high resolution imaging and data acquisition at the nanoscale, with on-the-fly processing.
Learn more ›
Products
Style Sheet for Instrument Cards Original
AutoScript TEM
  • Provides a direct link between research needs and microscope automation
  • Enables improved reproducibility and accuracy
  • Focuses time on the microscope for higher throughput
FIB-SEM and Laser Ablation
  • All three beams have same coincident point for accurate and repeatable cut placement
  • Millimeter-scale cross sections with up to 15,000x faster material removal than a typical FIB
  • Statistically relevant deep subsurface and 3D data analysis
Thermo Scientific Helios Hydra plasma focused ion beam scanning electron microscope (DualBeam)
Helios Hydra DualBeam
  • 4 fast switchable ion species (Xe, Ar, O, N) for optimized PFIB processing of a widest range of materials
  • Ga-free TEM sample preparation
  • Extreme high resolution SEM imaging
Helios 5 HX/Helios 5 UX/Helios 5 FX DualBeam
  • Fully automated, high-quality, ultra-thin TEM sample preparation
  • High throughput, high resolution subsurface and 3D characterization
  • Rapid nanoprototyping capabilities
Helios 5 PFIB DualBeam
  • Gallium-free STEM and TEM sample preparation
  • Multi-modal subsurface and 3D information
  • Next-generation 2.5 μA xenon plasma FIB column
Spectra Ultra
  • New imaging and spectroscopy capabilities on the most beam sensitive materials
  • A leap forward in EDX detection with Ultra-X
  • Column designed to maintain sample integrity.
Talos F200S TEM
  • Intuitive and easy-to-use automation software
  • Available with Super-X EDS for rapid quantitative chemical analysis
  • High-throughput with simultaneous multi-signal acquisition
Talos F200X TEM
  • High-resolution, EDS cleanliness, and quality in 2D as well as 3D
  • X-FEG and X-CFEG available for the highest brightness and energy resolution
  • High accuracy and repeatable results with integrated Thermo Scientific Velox Software
Thermo Scientific Talos F200C transmission electron microscope (TEM)
Talos F200C TEM
  • High-contrast and high-quality TEM and STEM imaging
  • 4k x 4k Ceta CMOS camera options for large FOV and high read-out speeds
  • Large pole piece gap and multiple in situ options
Talos F200i TEM
  • Compact design with X-TWIN objective lens
  • Available with S-FEG, X-FEG, and X-CFEG
  • Flexible and fast EDS options for comprehensive elemental analysis
Axia ChemiSEM
  • Live quantitative elemental mapping
  • High fidelity scanning electron microscopy imaging
  • Flexible and easy to use, even for novice users
  • Easy maintenance
VolumeScope 2 SEM
  • Isotropic 3D data from large volumes
  • High contrast and resolution in high and low vacuum modes
  • Simple switch between normal SEM use and serial block-face imaging
Thermo Scientific Apreo 2 scanning electron microscope (SEM)
Apreo 2 SEM
  • High-performance SEM for all-round nanometer or sub-nanometer resolution
  • In-column T1 backscatter detector for sensitive, TV-rate materials contrast
  • Excellent performance at long working distance (10 mm)
Phenom ParticleX Battery Desktop SEM
  • Versatile solution for high-quality, in-house analysis
  • Automated system and analysis of multiple samples
  • Testing 10x faster
Phenom ParticleX Steel Desktop SEM
  • SEM and EDS integrated
  • Ease of use
  • Sub-micrometer inclusions
Phenom XL G2 Desktop SEM
  • For large samples (100x100 mm) and ideal for automation
  • <10 nm resolution and up to 200,000x magnification; 4.8 kV up to 20 kV acceleration voltage
  • Optional fully integrated EDS and BSE detector
Phenom Pharos G2 Desktop FEG-SEM
  • FEG source with 1 – 20 kV acceleration voltage range
  • <2.0 nm (SE) and 3.0 nm (BSE) resolution @ 20 kV
  • Optional fully integrated EDS and SE detector
Phenom ParticleX TC Desktop SEM
  • Versatile desktop SEM with automation software for Technical Cleanliness
  • Resolution <10 nm; magnification up to 200,000x
  • Optional SE detector
Phenom ParticleX AM Desktop SEM
  • Versatile desktop SEM with automation software for Additive Manufacturing
  • Resolution <10 nm; magnification up to 200,000x
  • Optional SE detector
Thermo Scientific Maps electron microscopy software
Maps Software
  • Acquire high-resolution images over large areas
  • Easily find regions of interest
  • Automate image acquisition process
  • Correlate data from different sources
Avizo Software
Materials Science
  • Support for multi-data/multi-view, multi-channel, time series, very large data
  • Advanced multi-mode 2D/3D automatic registration
  • Artifact reduction algorithms
Elemental Mapping
  • Fast and reliable information on the distribution of elements within the sample or the selected line
  • Easily exported and reported results
FiberMetric
  • Save time by automated measurements
  • Fast and automated collection of all statistical data
  • View and measure micro and nano fibers with unmatched accuracy
Velox
  • An experiments panel on the left side of the processing window.
  • Live quantitative mapping
  • Interactive detector layout interface for reproducible experiment control and setup
PoroMetric
  • Correlate pore features such as area, aspect ratio, major and minor axis
  • Acquire images directly from the Desktop SEM
  • Statistical data with high-quality images
ParticleMetric
  • Integrated software in ProSuite for online and offline analysis
  • Correlating particle features such as diameter, circularity, aspect ratio and convexity
  • Creating image datasets with Automated Image Mapping
Contact us
Style Sheet for Support and Service footer
Style Sheet for Fonts
Style Sheet for Cards
Electron microscopy services for
the materials science
To ensure optimal system performance, we provide you access to a world-class network of field service experts, technical support, and certified spare parts.