Optimize protein integrity and yields during workflows
(See a list of the products featured in this article.)
Inside a cell, proteins are commonly separated from proteolytic enzymes through differential localization. Disruption of cellular and tissue architecture during protein extraction distorts the in vivo state by making all proteins potentially accessible for degradation or modification by endogenous proteases and phosphatases. Unregulated, these activities can reduce protein yield and function in a sample. To minimize these losses, protease and phosphatase inhibitors can be added during cell lysis to prevent degradation of extracted proteins and preserve protein structure, function, and yields during sample preparation.
Protease and phosphatase inhibitors
Protease and phosphatase inhibitor cocktails and tablets are ideal for protecting proteins during extraction procedures or lysate preparation using primary cells, cultured mammalian cells, animal tissues, plant tissues, yeast cells, or bacterial cells. Formulations are packaged in multiple sizes, and EDTA-free versions are available for divalent cation–sensitive assays. Thermo Scientific Pierce inhibitor mini tablets have been reformulated to dissolve quickly into a clear solution and are fully compatible with all Pierce protein assays. Figures 1 and 2 provide representative examples of the effectiveness of Pierce Protease and Phosphatase Inhibitor Mini Tablets.
Performance comparison between three commercially available protease inhibitor tablets
Figure 1. Performance comparison between three commercially available protease inhibitor tablets. Pancreatic extract (100 μL, 0.5 μg/μL) was incubated with cleavable fluorogenic substrates for trypsin and cysteine proteases, metalloproteases, and cathepsins in the presence of Thermo Scientific Pierce Protease Inhibitor Mini Tablets, Roche cOmplete Protease Inhibitor Tablets, and Sigma-Aldrich SIGMAFAST Protease Inhibitor Cocktail Tablets, with and without EDTA. Reactions were incubated for 1 hr at 37ºC, and fluorescence was determined at the appropriate detection emission wavelengths on a Thermo Scientific Varioskan Flash microplate reader. Percent protease inhibition is shown for each protease inhibitor formulation.
Inhibition of phosphatase activity in human colorectal carcinoma cell lysate
Figure 2. Inhibition of phosphatase activity in human colorectal carcinoma cell lysate. HCT116 cells were serum-starved and treated with epidermal growth factor (EGF) for 15 min or left untreated as controls (data not shown). Cell lysates were prepared in Thermo Scientific Pierce IP Lysis Buffer with or without Thermo Scientific Pierce Protease and Phosphatase Inhibitor Mini Tablets, EDTA-Free and incubated with anti–phosphotyrosine antibody overnight at 4ºC. The immunoprecipitated complex was incubated with Thermo Scientific Pierce Protein A/G Magnetic Beads for 1 hr at room temperature. After washing the beads, a low-pH elution was performed, and this eluate was run on a gel, transferred to a membrane, and probed with anti– EGF receptor (EGFR) antibody prior to chemiluminescence western blot detection.
Download the free Protein Preparation Handbook
The Protein Preparation Handbook provides technical information on our broad portfolio of reagents and tools for protein extraction, cleanup, immunoprecipitation, and purification, including the inhibitor tablets discussed here. Practical information, selection guides, and relevant data are included to help improve protein yield, facilitate downstream analysis, and optimize workflows.
Catalog # Name Size Price (USD) Qty
87787 Pierce™ IP Lysis Buffer, 100 mL Each
114.65
Online Exclusive
126.00
Save 11.35 (9%)
87788 Pierce™ IP Lysis Buffer, 250 mL Each
243.65
Online Exclusive
269.00
Save 25.35 (9%)
A32953 Pierce Protease Inhibitor Mini Tablets Each
172.65
Online Exclusive
192.00
Save 19.35 (10%)
A32963 Pierce Protease Inhibitor Tablets Each
359.65
Online Exclusive
395.00
Save 35.35 (9%)
A32955 Pierce™ Protease Inhibitor Mini Tablets, EDTA-free Each
164.65
Online Exclusive
192.00
Save 27.35 (14%)
A32965 Pierce™ Protease Inhibitor Tablets, EDTA-free, 20 Tablets Each
340.65
Online Exclusive
393.00
Save 52.35 (13%)
A32957 Pierce™ Phosphatase Inhibitor Mini Tablets, 20 Tablets Each
225.65
Online Exclusive
245.00
Save 19.35 (8%)
A32959 Pierce Protease and Phosphatase Inhibitor Mini Tablets Each
306.65
Online Exclusive
333.00
Save 26.35 (8%)
A32961 Pierce Protease and Phosphatase Inhibitor Mini Tablets,EDTA-free Each
301.65
Online Exclusive
333.00
Save 31.35 (9%)
Learn more about
Article download
Download a printer-friendly version of this article.
Download now
Not for resale. Super Bright Polymer Dyes are sold under license from Becton, Dickinson and Company.
For Research Use Only. Not for use in diagnostic procedures.