Stille cross-coupling reaction—a transition metal-catalyzed coupling reaction
In 1976, the first palladium-catalyzed cross-coupling of organotin compounds was accomplished by C. Eaborn, et al. The next year, M. Kosugi and T. Migita described the transition metal-catalyzed cross-coupling of organotin compounds with aryl halides and acid chlorides. Following this, in 1978, J.K. Stille used organotin compounds to synthesize ketones using milder reaction conditions than those used by Kosugi, but giving improved yields. In the early 1980s, Stille continued to develop and improve on his methodology, and today the palladium-catalyzed coupling reaction between an organostannane and an organic electrophile to form carbon-carbon bonds is known as the Stille cross-coupling reaction.
Despite the toxicity of the tin compounds, the Stille reaction has developed into one of the most important reactions in organic synthesis. The success of the Stille coupling derives primarily from its ability of the tin precursors to tolerate a wide variety of functional groups while also lacking sensitivity to air and moisture, unlike other reactive organometallic compounds.
The mild reaction conditions of the method are compatible with many types of functional groups, including amines, amides, esters, carboxylic acids, ketones, hydroxyl, and formyl groups, to name a few.
Among the many uses of the Stille cross-coupling reaction is the total synthesis of natural products, including the manzamine alkaloid ircinal A, and another alkaloid, quadrigemine C. 
Review available Thermo Scientific products for the Stille cross-coupling reaction:
Katalognr. Produkt Größe Preis (EUR) Menge
A10746.30 Tri-n-butyltin chloride, 96% Each
73,10
Exklusiv online
86,00
Erparnis 12,90 (15%)
A10746.18 Tri-n-butyltin chloride, 96% Each
23,38
Exklusiv online
27,50
Erparnis 4,12 (15%)
B24911.03 2-(Tri-n-butylstannyl)pyridine, tech. 80% Each
39,02
Exklusiv online
45,90
Erparnis 6,88 (15%)
B24911.14 2-(Tri-n-butylstannyl)pyridine, tech. 80% Each
424,15
Exklusiv online
499,00
Erparnis 74,85 (15%)
H51435.03 4-(Tri-n-butylstannyl)pyridine, 96% Each
79,48
Exklusiv online
93,50
Erparnis 14,02 (15%)
B24911.06 2-(Tri-n-butylstannyl)pyridine, tech. 80% Each
122,40
Exklusiv online
144,00
Erparnis 21,60 (15%)
H51370.MD 2-(Tri-n-butylstannyl)pyrazine, 95% Each
41,82
Exklusiv online
49,20
Erparnis 7,38 (15%)
H51435.06 4-(Tri-n-butylstannyl)pyridine, 96% Each
271,15
Exklusiv online
319,00
Erparnis 47,85 (15%)
H55904.AA 2-(Tri-n-butylstannyl)thiophene, 97% Each
34,68
Exklusiv online
40,80
Erparnis 6,12 (15%)
H51458.03 5-(Tri-n-butylstannyl)pyrimidine, 96% Each
181,05
Exklusiv online
213,00
Erparnis 31,95 (15%)
A10746.0B Tri-n-butyltin chloride, 96% Each
208,25
Exklusiv online
245,00
Erparnis 36,75 (15%)
H55904.AC 2-(Tri-n-butylstannyl)thiophene, 97% Each
104,55
Exklusiv online
123,00
Erparnis 18,45 (15%)
H55904.AE 2-(Tri-n-butylstannyl)thiophene, 97% Each
307,70
Exklusiv online
362,00
Erparnis 54,30 (15%)
H51370.03 2-(Tri-n-butylstannyl)pyrazine, 95% Each
120,70
Exklusiv online
142,00
Erparnis 21,30 (15%)
H51458.MD 5-(Tri-n-butylstannyl)pyrimidine, 96% Each
58,48
Exklusiv online
68,80
Erparnis 10,32 (15%)
Vote for your favorite named reaction
Learn more about the most popular named reactions based on feedback from our customers. Do you have a favorite reaction? Let us know!
Vote now