Search Thermo Fisher Scientific
Solve your separation challenges with our HPLC and UHPLC columns for small molecules. Our popular product lines of Hypersil GOLD and Accucore columns are designed for a wide range of applications and offer the following features:
From the most commonly-used C18 phases, to innovative chemistries, we have the right column to fulfill your chromatography needs
Fully porous for simple samples
Solid core for high resolution
Your first choice of columns for method development
Hydrophobic Column Phases | Reversed phase chemistry for separation via hydrophobic interactions. Stationary phases with longer carbon chains provide longer retention times and greater hydrophobicity.
|
Polar-Modified C18 Column Phases | Good for the separation of acidic and basic compounds via reversed phase chromatography using an aqueous mobile phase.
|
Phenyl-Based Column Phases | Orthogonal selectivity to traditional hydrophobic phases that utilizes aromatic interactions to achieve challenging separations of aromatic and polar compounds. Phenyl, PFP |
Ion-Exchange Column Phases | Ionic and polar compounds are retained via hydrophilic and ionic interactions with a charged stationary phase. Retained compounds can be eluted by changing the solvent conditions of the column.
|
Hydrophilic Columns | Ideal for retaining and separating polar compounds based on polarity differences and affinity for a polar stationary phase. HILIC, Silica |
Solid core silica particles for high-resolution separation at any pressure
Hydrophobic Column Phases | Reversed phase chemistry for separation via hydrophobic interactions. Stationary phases with longer carbon chains provide longer retention times and greater hydrophobicity. C18, C8, C4 |
Specialty Hydrophobic Column Phases | Alternative selectivity to C18 for hydrophobic retention and the geometric separation of structural isomers. RP-MS, C30 |
Phenyl-Based Column Phases | Orthogonal selectivity to traditional hydrophobic phases that utilizes aromatic interactions to achieve challenging separations of aromatic and polar compounds. Biphenyl, Phenyl-Hexyl, PFP |
Polar-Modified C18 Column Phases | Good for the separation of acidic and basic compounds via reversed phase chromatography using an aqueous mobile phase Polar Premium, aQ |
HILIC Column Phases | Ideal for retaining and separating polar compounds based on polarity differences and affinity for a polar stationary phase. HILIC, Amide HILIC |
Phase Name | 1.5 μm | 2.6 μm | 4 μm | Pore Size (Å) | Surface Area (m²/g) | Carbon Load (%) | USP |
— | — | 80 | 130 | 9 | L1 | ||
— | — | 80 | 130 | 6.5 | L1 | ||
— | — | 80 | 130 | 7 | L1 | ||
— | — | 80 | 130 | 9 | L7 | ||
— | — | 80 | 130 | 4 | L7 | ||
— | — | 150 | 80 | 2 | L26 | ||
— | — | 80 | 130 | 9 | L1 | ||
— | — | 80 | 130 | 8 | L60 | ||
— | — | 80 | 130 | 7 | L1 | ||
— | — | 80 | 130 | 5 | L62 | ||
— | — | 80 | 130 | 6 | L11 | ||
— | — | 80 | 130 | 5 | L11 | ||
— | — | 80 | 130 | 5 | L43 | ||
— | — | 80 | 130 | 6 | – | ||
— | — | 80 | 130 | – | L3 | ||
— | — | 150 | 80 | – | – | ||
— | — | 80 | 130 | – | – |
|
|
|
A great resource for getting familiar with and comparing (U)HPLC columns, our Premier Columns Guide contains:
Download premier columns guide
* Required field