Search Thermo Fisher Scientific
The Thermo Scientific 43iQ Sulfur Dioxide (SO2) gas analyzer utilizes pulsed fluorescence technology to measure the amount of sulfur dioxide in the air. The pulsing of the UV source lamp serves to increase the optical intensity whereby a greater UV energy throughput and lower detectable SO2 concentration are realized. Reflective bandpass filters, as compared to commonly used transmission filters, are less subject to photochemical degradation and more selective in wavelength isolation. This results in both increased detection specificity and long term stability.
The iQ Series Instrument Platform is a smart environmental monitoring solution for ambient and source gas analysis that affords greater control over instrument performance and data availability.
Learn more about the 43iQ Sulfur Dioxide Analyzer and the complete line of iQ Series Gas Analyzers.